Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92456893-92457432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPN
Entrez Gene ID
54829
Gene Name
asporin
Gene Aliases
OS3, PLAP-1, PLAP1, SLRR1C
UniGene
Hs.435655
Gene Chromosome Location
Chr.9: 92456205-92482562 on Build GRCh38
Transcripts
NM_001193335.1 NM_017680.4 NM_001012267.2 NM_001286969.1 XM_011518689.1

Gene Symbol
CENPP
Entrez Gene ID
401541
Gene Name
centromere protein P
Gene Aliases
CENP-P
UniGene
Hs.713775
Gene Chromosome Location
Chr.9: 92325457-92615164 on Build GRCh38
Transcripts
NM_001193335.1 NM_017680.4 NM_001012267.2 NM_001286969.1 XM_011518689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1111118 c.1093C>T p.R365C Chr.9: 92457338-92457338 on Build GRCh38
3734267 c.1012A>C p.K338Q Chr.9: 92457419-92457419 on Build GRCh38
1463585 c.1099_1100delGT p.V367fs*>14 Chr.9: 92457331-92457332 on Build GRCh38
1111119 c.1025C>A p.S342Y Chr.9: 92457406-92457406 on Build GRCh38
6498926 c.1101T>G p.V367V Chr.9: 92457330-92457330 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746445338 Chr.9: 92457259-92457259 on Build GRCh38
rs542492351 Chr.9: 92456978-92456978 on Build GRCh38
rs138321845 Chr.9: 92457277-92457277 on Build GRCh38
rs4744132 Chr.9: 92457315-92457315 on Build GRCh38
rs190218000 Chr.9: 92456993-92456993 on Build GRCh38
rs148676537 Chr.9: 92457372-92457372 on Build GRCh38
rs748686870 Chr.9: 92457398-92457398 on Build GRCh38
rs1020927708 Chr.9: 92456992-92456992 on Build GRCh38
rs866428258 Chr.9: 92457361-92457361 on Build GRCh38
rs894682780 Chr.9: 92456930-92456930 on Build GRCh38
rs748649282 Chr.9: 92457283-92457283 on Build GRCh38
rs929525388 Chr.9: 92457108-92457108 on Build GRCh38
rs542221018 Chr.9: 92457344-92457344 on Build GRCh38
rs779693811 Chr.9: 92457242-92457242 on Build GRCh38
rs761721603 Chr.9: 92457346-92457346 on Build GRCh38
rs1020594690 Chr.9: 92457060-92457060 on Build GRCh38
rs773519082 Chr.9: 92457292-92457292 on Build GRCh38
rs926238139 Chr.9: 92457094-92457094 on Build GRCh38
rs888945026 Chr.9: 92456937-92456937 on Build GRCh38
rs992096621 Chr.9: 92457130-92457130 on Build GRCh38
rs965812945 Chr.9: 92457004-92457004 on Build GRCh38
rs1036406249 Chr.9: 92457226-92457226 on Build GRCh38
rs758043075 Chr.9: 92457239-92457239 on Build GRCh38
rs774720272 Chr.9: 92457309-92457309 on Build GRCh38
rs868522113 Chr.9: 92457264-92457264 on Build GRCh38
rs1039221346 Chr.9: 92457171-92457171 on Build GRCh38
rs1049457881 Chr.9: 92456933-92456933 on Build GRCh38
rs917430235 Chr.9: 92457198-92457198 on Build GRCh38
rs893863130 Chr.9: 92457187-92457187 on Build GRCh38
rs756639577 Chr.9: 92457401-92457401 on Build GRCh38
rs559223233 Chr.9: 92457129-92457129 on Build GRCh38
rs1018072304 Chr.9: 92456950-92456950 on Build GRCh38
rs368817815 Chr.9: 92457173-92457173 on Build GRCh38
rs771223864 Chr.9: 92457301-92457301 on Build GRCh38
rs918954979 Chr.9: 92457105-92457105 on Build GRCh38
rs560931002 Chr.9: 92457406-92457406 on Build GRCh38
rs749516419 Chr.9: 92457300-92457300 on Build GRCh38
rs3174352 Chr.9: 92456966-92456966 on Build GRCh38
rs566084628 Chr.9: 92456925-92456925 on Build GRCh38
rs765066997 Chr.9: 92457347-92457347 on Build GRCh38
rs572792291 Chr.9: 92457061-92457061 on Build GRCh38
rs991344139 Chr.9: 92457032-92457032 on Build GRCh38
rs943494361 Chr.9: 92457159-92457161 on Build GRCh38
rs770166448 Chr.9: 92457284-92457284 on Build GRCh38
rs758217255 Chr.9: 92457370-92457370 on Build GRCh38
rs960177829 Chr.9: 92457018-92457018 on Build GRCh38
rs545140869 Chr.9: 92457274-92457274 on Build GRCh38
rs7043809 Chr.9: 92457336-92457336 on Build GRCh38
rs539889035 Chr.9: 92457210-92457210 on Build GRCh38
rs754675611 Chr.9: 92457276-92457276 on Build GRCh38
rs776690193 Chr.9: 92457342-92457342 on Build GRCh38
rs17591776 Chr.9: 92457037-92457037 on Build GRCh38
rs371868297 Chr.9: 92457374-92457374 on Build GRCh38
rs750126730 Chr.9: 92457359-92457359 on Build GRCh38
rs537736130 Chr.9: 92457079-92457079 on Build GRCh38
rs577499813 Chr.9: 92457216-92457216 on Build GRCh38
rs3174351 Chr.9: 92456953-92456953 on Build GRCh38
rs767729849 Chr.9: 92457337-92457337 on Build GRCh38
rs751232189 Chr.9: 92457373-92457373 on Build GRCh38

View Full Product Details