Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 93126307-93126804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NINJ1
Entrez Gene ID
4814
Gene Name
ninjurin 1
Gene Aliases
NIN1, NINJURIN
UniGene
Hs.494457
Gene Chromosome Location
Chr.9: 93121489-93134304 on Build GRCh38
Transcripts
NM_004148.3 XM_011518716.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6453669 c.254C>G p.S85C Chr.9: 93126460-93126460 on Build GRCh38
6348929 c.117C>A p.N39K Chr.9: 93126597-93126597 on Build GRCh38
6726281 c.199G>A p.V67I Chr.9: 93126515-93126515 on Build GRCh38
6726280 c.182C>T p.A61V Chr.9: 93126532-93126532 on Build GRCh38
6236443 c.79G>A p.A27T Chr.9: 93126635-93126635 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202012057 Chr.9: 93126637-93126637 on Build GRCh38
rs770810135 Chr.9: 93126519-93126519 on Build GRCh38
rs375893056 Chr.9: 93126492-93126492 on Build GRCh38
rs758840752 Chr.9: 93126484-93126484 on Build GRCh38
rs924785674 Chr.9: 93126668-93126668 on Build GRCh38
rs745548269 Chr.9: 93126531-93126531 on Build GRCh38
rs771785987 Chr.9: 93126532-93126532 on Build GRCh38
rs780756237 Chr.9: 93126689-93126689 on Build GRCh38
rs901514888 Chr.9: 93126525-93126525 on Build GRCh38
rs765652528 Chr.9: 93126552-93126552 on Build GRCh38
rs371520758 Chr.9: 93126406-93126406 on Build GRCh38
rs780083241 Chr.9: 93126419-93126419 on Build GRCh38
rs930200752 Chr.9: 93126682-93126682 on Build GRCh38
rs761838237 Chr.9: 93126676-93126677 on Build GRCh38
rs975426989 Chr.9: 93126427-93126427 on Build GRCh38
rs758621940 Chr.9: 93126567-93126567 on Build GRCh38
rs961922537 Chr.9: 93126363-93126363 on Build GRCh38
rs140825165 Chr.9: 93126420-93126420 on Build GRCh38
rs371691982 Chr.9: 93126624-93126624 on Build GRCh38
rs755107685 Chr.9: 93126656-93126656 on Build GRCh38
rs200780440 Chr.9: 93126614-93126614 on Build GRCh38
rs769630940 Chr.9: 93126513-93126513 on Build GRCh38
rs183379364 Chr.9: 93126342-93126342 on Build GRCh38
rs536006000 Chr.9: 93126632-93126632 on Build GRCh38
rs147901839 Chr.9: 93126534-93126534 on Build GRCh38
rs768909429 Chr.9: 93126509-93126509 on Build GRCh38
rs1008103245 Chr.9: 93126348-93126349 on Build GRCh38
rs547110787 Chr.9: 93126609-93126609 on Build GRCh38
rs759242044 Chr.9: 93126444-93126444 on Build GRCh38
rs554253795 Chr.9: 93126638-93126638 on Build GRCh38
rs770617886 Chr.9: 93126438-93126438 on Build GRCh38
rs577144241 Chr.9: 93126354-93126354 on Build GRCh38
rs781111372 Chr.9: 93126574-93126574 on Build GRCh38
rs947622622 Chr.9: 93126424-93126424 on Build GRCh38
rs1050499908 Chr.9: 93126718-93126718 on Build GRCh38
rs368328185 Chr.9: 93126515-93126515 on Build GRCh38
rs140126462 Chr.9: 93126535-93126535 on Build GRCh38
rs558793520 Chr.9: 93126687-93126687 on Build GRCh38
rs772176567 Chr.9: 93126415-93126415 on Build GRCh38
rs773970689 Chr.9: 93126520-93126520 on Build GRCh38
rs200628483 Chr.9: 93126512-93126512 on Build GRCh38
rs79209251 Chr.9: 93126548-93126548 on Build GRCh38
rs922846658 Chr.9: 93126683-93126683 on Build GRCh38
rs752654951 Chr.9: 93126667-93126667 on Build GRCh38
rs188398780 Chr.9: 93126677-93126677 on Build GRCh38
rs764606426 Chr.9: 93126489-93126489 on Build GRCh38
rs752216843 Chr.9: 93126462-93126462 on Build GRCh38
rs953241517 Chr.9: 93126345-93126345 on Build GRCh38
rs574864844 Chr.9: 93126737-93126737 on Build GRCh38
rs773535459 Chr.9: 93126775-93126775 on Build GRCh38
rs368098029 Chr.9: 93126550-93126550 on Build GRCh38
rs867491971 Chr.9: 93126732-93126732 on Build GRCh38
rs768150552 Chr.9: 93126688-93126688 on Build GRCh38
rs757233765 Chr.9: 93126675-93126675 on Build GRCh38
rs749272000 Chr.9: 93126575-93126575 on Build GRCh38
rs779257590 Chr.9: 93126403-93126403 on Build GRCh38
rs551321471 Chr.9: 93126458-93126458 on Build GRCh38
rs1016491217 Chr.9: 93126558-93126558 on Build GRCh38
rs200089902 Chr.9: 93126678-93126678 on Build GRCh38
rs762750806 Chr.9: 93126434-93126434 on Build GRCh38
rs559535977 Chr.9: 93126734-93126734 on Build GRCh38
rs745603326 Chr.9: 93126591-93126591 on Build GRCh38
rs541396699 Chr.9: 93126719-93126719 on Build GRCh38
rs766725739 Chr.9: 93126646-93126646 on Build GRCh38
rs538594312 Chr.9: 93126576-93126576 on Build GRCh38
rs750743421 Chr.9: 93126554-93126554 on Build GRCh38
rs771983861 Chr.9: 93126599-93126599 on Build GRCh38
rs750690017 Chr.9: 93126507-93126507 on Build GRCh38
rs762411363 Chr.9: 93126636-93126636 on Build GRCh38
rs972064028 Chr.9: 93126373-93126373 on Build GRCh38
rs779111240 Chr.9: 93126506-93126506 on Build GRCh38
rs780136552 Chr.9: 93126508-93126508 on Build GRCh38
rs948227853 Chr.9: 93126498-93126498 on Build GRCh38
rs757437474 Chr.9: 93126395-93126395 on Build GRCh38
rs12235416 Chr.9: 93126385-93126385 on Build GRCh38
rs754291134 Chr.9: 93126491-93126491 on Build GRCh38
rs777230951 Chr.9: 93126549-93126549 on Build GRCh38
rs571472744 Chr.9: 93126572-93126572 on Build GRCh38
rs144669023 Chr.9: 93126533-93126533 on Build GRCh38
rs993390258 Chr.9: 93126571-93126571 on Build GRCh38
rs899712629 Chr.9: 93126776-93126776 on Build GRCh38
rs145421928 Chr.9: 93126590-93126590 on Build GRCh38
rs35551956 Chr.9: 93126348-93126348 on Build GRCh38
rs767667390 Chr.9: 93126666-93126666 on Build GRCh38
rs150031694 Chr.9: 93126570-93126570 on Build GRCh38
rs772885711 Chr.9: 93126430-93126430 on Build GRCh38
rs372269819 Chr.9: 93126516-93126516 on Build GRCh38
rs540282147 Chr.9: 93126352-93126352 on Build GRCh38
rs751829505 Chr.9: 93126649-93126649 on Build GRCh38
rs933007138 Chr.9: 93126701-93126701 on Build GRCh38
rs577503592 Chr.9: 93126702-93126702 on Build GRCh38
rs774466422 Chr.9: 93126677-93126677 on Build GRCh38
rs201900581 Chr.9: 93126476-93126476 on Build GRCh38
rs565241448 Chr.9: 93126353-93126353 on Build GRCh38
rs990410657 Chr.9: 93126370-93126370 on Build GRCh38

View Full Product Details