Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 96301895-96302391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B3
Entrez Gene ID
3293
Gene Name
hydroxysteroid 17-beta dehydrogenase 3
Gene Aliases
EDH17B3, SDR12C2
UniGene
Hs.477
Gene Chromosome Location
Chr.9: 96235306-96313987 on Build GRCh38
Transcripts
NM_000197.1 XM_011518618.2 XM_011518619.2 XM_017014671.1 XM_017014672.1 XM_017014673.1 XM_017014674.1 XM_017014675.1 XM_017014676.1 XM_017014677.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3413859 c.64G>A p.A22T Chr.9: 96302041-96302041 on Build GRCh38
5876631 c.31C>T p.L11F Chr.9: 96302074-96302074 on Build GRCh38
3659833 c.15G>A p.L5L Chr.9: 96302090-96302090 on Build GRCh38
3927872 c.108G>A p.W36* Chr.9: 96301997-96301997 on Build GRCh38
7225425 c.91G>A p.V31I Chr.9: 96302014-96302014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35188473 Chr.9: 96302363-96302363 on Build GRCh38
rs771557138 Chr.9: 96302134-96302134 on Build GRCh38
rs763883369 Chr.9: 96302069-96302069 on Build GRCh38
rs139084702 Chr.9: 96301972-96301972 on Build GRCh38
rs530025013 Chr.9: 96301934-96301934 on Build GRCh38
rs114520006 Chr.9: 96302032-96302032 on Build GRCh38
rs377018679 Chr.9: 96302095-96302095 on Build GRCh38
rs753428378 Chr.9: 96302344-96302344 on Build GRCh38
rs376457868 Chr.9: 96302153-96302153 on Build GRCh38
rs749816800 Chr.9: 96302015-96302015 on Build GRCh38
rs770504476 Chr.9: 96302017-96302017 on Build GRCh38
rs199678489 Chr.9: 96302149-96302149 on Build GRCh38
rs577299245 Chr.9: 96302165-96302165 on Build GRCh38
rs28932190 Chr.9: 96302336-96302336 on Build GRCh38
rs199704576 Chr.9: 96302131-96302131 on Build GRCh38
rs756479113 Chr.9: 96302123-96302123 on Build GRCh38
rs747173183 Chr.9: 96301984-96301984 on Build GRCh38
rs767254534 Chr.9: 96301953-96301953 on Build GRCh38
rs186025805 Chr.9: 96302366-96302366 on Build GRCh38
rs760649604 Chr.9: 96301927-96301928 on Build GRCh38
rs200708778 Chr.9: 96302110-96302110 on Build GRCh38
rs774078322 Chr.9: 96301938-96301938 on Build GRCh38
rs775234559 Chr.9: 96302040-96302040 on Build GRCh38
rs776847871 Chr.9: 96301925-96301925 on Build GRCh38
rs760426847 Chr.9: 96302045-96302045 on Build GRCh38
rs928710667 Chr.9: 96302234-96302234 on Build GRCh38
rs2066474 Chr.9: 96302143-96302143 on Build GRCh38
rs751562442 Chr.9: 96302105-96302105 on Build GRCh38
rs770305727 Chr.9: 96302010-96302010 on Build GRCh38
rs752426255 Chr.9: 96301955-96301955 on Build GRCh38
rs769008684 Chr.9: 96301924-96301924 on Build GRCh38
rs537079694 Chr.9: 96301971-96301971 on Build GRCh38
rs1049882146 Chr.9: 96302112-96302112 on Build GRCh38
rs755152747 Chr.9: 96301992-96301992 on Build GRCh38
rs763632725 Chr.9: 96302335-96302335 on Build GRCh38
rs981462321 Chr.9: 96302208-96302208 on Build GRCh38
rs199734603 Chr.9: 96301946-96301946 on Build GRCh38
rs748613560 Chr.9: 96301926-96301926 on Build GRCh38
rs1049054724 Chr.9: 96301928-96301928 on Build GRCh38
rs974553626 Chr.9: 96302142-96302142 on Build GRCh38
rs759418931 Chr.9: 96302138-96302138 on Build GRCh38
rs369072039 Chr.9: 96302318-96302318 on Build GRCh38
rs2066480 Chr.9: 96302014-96302014 on Build GRCh38
rs780124210 Chr.9: 96301979-96301979 on Build GRCh38
rs907829272 Chr.9: 96302152-96302152 on Build GRCh38
rs201767176 Chr.9: 96301937-96301937 on Build GRCh38
rs563753328 Chr.9: 96302190-96302192 on Build GRCh38
rs1055887604 Chr.9: 96302280-96302280 on Build GRCh38
rs1001410448 Chr.9: 96302178-96302178 on Build GRCh38
rs937449922 Chr.9: 96302263-96302263 on Build GRCh38
rs991758568 Chr.9: 96302246-96302246 on Build GRCh38
rs756803511 Chr.9: 96302172-96302172 on Build GRCh38
rs774942837 Chr.9: 96302147-96302147 on Build GRCh38
rs770393873 Chr.9: 96301929-96301929 on Build GRCh38
rs371391168 Chr.9: 96301930-96301930 on Build GRCh38
rs2066476 Chr.9: 96302133-96302133 on Build GRCh38
rs765227695 Chr.9: 96302103-96302103 on Build GRCh38
rs191153391 Chr.9: 96301966-96301966 on Build GRCh38
rs781518300 Chr.9: 96301996-96301996 on Build GRCh38
rs907127544 Chr.9: 96302168-96302168 on Build GRCh38
rs760336657 Chr.9: 96302154-96302154 on Build GRCh38
rs116530138 Chr.9: 96302033-96302033 on Build GRCh38
rs868469733 Chr.9: 96302055-96302055 on Build GRCh38
rs955572090 Chr.9: 96302360-96302360 on Build GRCh38
rs201302838 Chr.9: 96302039-96302039 on Build GRCh38
rs565374723 Chr.9: 96302330-96302330 on Build GRCh38
rs2066484 Chr.9: 96302114-96302114 on Build GRCh38
rs778294421 Chr.9: 96302128-96302128 on Build GRCh38
rs8190482 Chr.9: 96302357-96302357 on Build GRCh38
rs34037771 Chr.9: 96302303-96302303 on Build GRCh38
rs139582148 Chr.9: 96302096-96302096 on Build GRCh38
rs985119989 Chr.9: 96302150-96302150 on Build GRCh38
rs779510189 Chr.9: 96302137-96302137 on Build GRCh38
rs8190483 Chr.9: 96301921-96301922 on Build GRCh38
rs373446255 Chr.9: 96302121-96302121 on Build GRCh38
rs781387334 Chr.9: 96302044-96302044 on Build GRCh38
rs1006028647 Chr.9: 96302215-96302215 on Build GRCh38
rs377243298 Chr.9: 96301931-96301931 on Build GRCh38

View Full Product Details