Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98201288-98201804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D2
Entrez Gene ID
55357
Gene Name
TBC1 domain family member 2
Gene Aliases
PARIS-1, PARIS1, TBC1D2A
UniGene
Hs.371016
Gene Chromosome Location
Chr.9: 98198998-98255721 on Build GRCh38
Transcripts
NM_001267571.1 NM_001267572.1 NM_018421.3 NR_051978.1 XM_011518843.1 XM_017014899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1228579 c.2362G>A p.V788I Chr.9: 98201574-98201574 on Build GRCh38
6893777 c.1005G>A p.V335V Chr.9: 98201551-98201551 on Build GRCh38
5875221 c.1013C>T p.A338V Chr.9: 98201543-98201543 on Build GRCh38
4870016 c.1012G>A p.A338T Chr.9: 98201544-98201544 on Build GRCh38
5475207 c.2304G>A p.S768S Chr.9: 98201632-98201632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376157763 Chr.9: 98201530-98201530 on Build GRCh38
rs768083057 Chr.9: 98201424-98201424 on Build GRCh38
rs372529997 Chr.9: 98201494-98201494 on Build GRCh38
rs969006255 Chr.9: 98201386-98201386 on Build GRCh38
rs774979152 Chr.9: 98201691-98201691 on Build GRCh38
rs779463392 Chr.9: 98201517-98201517 on Build GRCh38
rs958332674 Chr.9: 98201779-98201779 on Build GRCh38
rs79987207 Chr.9: 98201695-98201695 on Build GRCh38
rs769767410 Chr.9: 98201537-98201537 on Build GRCh38
rs544237103 Chr.9: 98201353-98201354 on Build GRCh38
rs768315511 Chr.9: 98201474-98201474 on Build GRCh38
rs933449740 Chr.9: 98201455-98201455 on Build GRCh38
rs777686317 Chr.9: 98201538-98201538 on Build GRCh38
rs752211136 Chr.9: 98201701-98201701 on Build GRCh38
rs967189414 Chr.9: 98201694-98201694 on Build GRCh38
rs981026551 Chr.9: 98201394-98201394 on Build GRCh38
rs752444063 Chr.9: 98201609-98201609 on Build GRCh38
rs772091155 Chr.9: 98201490-98201490 on Build GRCh38
rs148540803 Chr.9: 98201593-98201593 on Build GRCh38
rs534913208 Chr.9: 98201632-98201632 on Build GRCh38
rs768618403 Chr.9: 98201567-98201567 on Build GRCh38
rs957053268 Chr.9: 98201754-98201754 on Build GRCh38
rs764329012 Chr.9: 98201443-98201443 on Build GRCh38
rs1031928755 Chr.9: 98201758-98201758 on Build GRCh38
rs200433647 Chr.9: 98201513-98201513 on Build GRCh38
rs763310349 Chr.9: 98201683-98201683 on Build GRCh38
rs3739668 Chr.9: 98201371-98201371 on Build GRCh38
rs925530570 Chr.9: 98201419-98201419 on Build GRCh38
rs77093962 Chr.9: 98201677-98201677 on Build GRCh38
rs767174002 Chr.9: 98201698-98201698 on Build GRCh38
rs778978870 Chr.9: 98201644-98201644 on Build GRCh38
rs779329583 Chr.9: 98201457-98201457 on Build GRCh38
rs781476687 Chr.9: 98201461-98201461 on Build GRCh38
rs980411927 Chr.9: 98201757-98201757 on Build GRCh38
rs1017829032 Chr.9: 98201391-98201391 on Build GRCh38
rs374539701 Chr.9: 98201655-98201655 on Build GRCh38
rs751032477 Chr.9: 98201523-98201523 on Build GRCh38
rs753972506 Chr.9: 98201584-98201584 on Build GRCh38
rs143801854 Chr.9: 98201491-98201491 on Build GRCh38
rs897891632 Chr.9: 98201338-98201338 on Build GRCh38
rs773526544 Chr.9: 98201577-98201577 on Build GRCh38
rs747893061 Chr.9: 98201475-98201475 on Build GRCh38
rs754210362 Chr.9: 98201502-98201502 on Build GRCh38
rs752901826 Chr.9: 98201450-98201450 on Build GRCh38
rs988768584 Chr.9: 98201782-98201782 on Build GRCh38
rs966217393 Chr.9: 98201393-98201393 on Build GRCh38
rs762968935 Chr.9: 98201486-98201486 on Build GRCh38
rs757778445 Chr.9: 98201449-98201449 on Build GRCh38
rs757964172 Chr.9: 98201462-98201462 on Build GRCh38
rs371036180 Chr.9: 98201687-98201687 on Build GRCh38
rs770857502 Chr.9: 98201543-98201543 on Build GRCh38
rs373038046 Chr.9: 98201437-98201437 on Build GRCh38
rs767523389 Chr.9: 98201575-98201575 on Build GRCh38
rs371626516 Chr.9: 98201589-98201589 on Build GRCh38
rs745782199 Chr.9: 98201654-98201654 on Build GRCh38
rs776900208 Chr.9: 98201574-98201574 on Build GRCh38
rs767120830 Chr.9: 98201600-98201600 on Build GRCh38
rs142304145 Chr.9: 98201533-98201533 on Build GRCh38
rs748401585 Chr.9: 98201663-98201663 on Build GRCh38
rs775450223 Chr.9: 98201548-98201548 on Build GRCh38
rs969058558 Chr.9: 98201711-98201711 on Build GRCh38
rs750927558 Chr.9: 98201458-98201458 on Build GRCh38
rs369104720 Chr.9: 98201482-98201482 on Build GRCh38
rs917610195 Chr.9: 98201385-98201385 on Build GRCh38
rs149010653 Chr.9: 98201483-98201483 on Build GRCh38
rs754582382 Chr.9: 98201595-98201595 on Build GRCh38
rs746681152 Chr.9: 98201471-98201471 on Build GRCh38
rs771262049 Chr.9: 98201449-98201450 on Build GRCh38
rs769954649 Chr.9: 98201676-98201676 on Build GRCh38
rs755793313 Chr.9: 98201623-98201623 on Build GRCh38
rs574780559 Chr.9: 98201420-98201420 on Build GRCh38
rs376011301 Chr.9: 98201470-98201470 on Build GRCh38
rs758873348 Chr.9: 98201514-98201514 on Build GRCh38
rs202081816 Chr.9: 98201633-98201633 on Build GRCh38
rs760745254 Chr.9: 98201554-98201554 on Build GRCh38
rs111337889 Chr.9: 98201312-98201312 on Build GRCh38
rs766851660 Chr.9: 98201590-98201590 on Build GRCh38
rs777465752 Chr.9: 98201532-98201532 on Build GRCh38
rs754186532 Chr.9: 98201447-98201447 on Build GRCh38
rs1045811916 Chr.9: 98201334-98201334 on Build GRCh38
rs147981592 Chr.9: 98201496-98201496 on Build GRCh38
rs1028067410 Chr.9: 98201354-98201354 on Build GRCh38
rs914033099 Chr.9: 98201448-98201448 on Build GRCh38
rs1039314473 Chr.9: 98201614-98201614 on Build GRCh38
rs777739277 Chr.9: 98201629-98201629 on Build GRCh38
rs145936813 Chr.9: 98201542-98201542 on Build GRCh38

View Full Product Details