Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 100252044-100252549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INVS
Entrez Gene ID
27130
Gene Name
inversin
Gene Aliases
INV, NPH2, NPHP2
UniGene
Hs.558477
Gene Chromosome Location
Chr.9: 100099185-100302175 on Build GRCh38
Transcripts
NM_001318381.1 NM_001318382.1 NM_014425.4 NR_134606.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3432705 c.1100C>A p.S367Y Chr.9: 100252304-100252304 on Build GRCh38
3432706 c.812C>A p.S271Y Chr.9: 100252304-100252304 on Build GRCh38
5415757 c.942T>C p.I314I Chr.9: 100252434-100252434 on Build GRCh38
116164 c.1108G>T p.V370F Chr.9: 100252312-100252312 on Build GRCh38
4692787 c.1117G>A p.V373M Chr.9: 100252321-100252321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370344619 Chr.9: 100252247-100252247 on Build GRCh38
rs762196389 Chr.9: 100252441-100252441 on Build GRCh38
rs1037513621 Chr.9: 100252479-100252479 on Build GRCh38
rs200546215 Chr.9: 100252315-100252315 on Build GRCh38
rs187066093 Chr.9: 100252507-100252507 on Build GRCh38
rs759162487 Chr.9: 100252321-100252321 on Build GRCh38
rs960177449 Chr.9: 100252500-100252500 on Build GRCh38
rs368746630 Chr.9: 100252423-100252423 on Build GRCh38
rs201219699 Chr.9: 100252257-100252257 on Build GRCh38
rs747812451 Chr.9: 100252379-100252379 on Build GRCh38
rs373264811 Chr.9: 100252270-100252270 on Build GRCh38
rs770939282 Chr.9: 100252404-100252404 on Build GRCh38
rs183072942 Chr.9: 100252075-100252075 on Build GRCh38
rs750752575 Chr.9: 100252323-100252323 on Build GRCh38
rs746571523 Chr.9: 100252343-100252343 on Build GRCh38
rs375376581 Chr.9: 100252466-100252466 on Build GRCh38
rs912526774 Chr.9: 100252450-100252450 on Build GRCh38
rs1036079928 Chr.9: 100252475-100252475 on Build GRCh38
rs907686265 Chr.9: 100252152-100252152 on Build GRCh38
rs111876971 Chr.9: 100252254-100252254 on Build GRCh38
rs377068989 Chr.9: 100252234-100252234 on Build GRCh38
rs749012941 Chr.9: 100252393-100252393 on Build GRCh38
rs759466960 Chr.9: 100252417-100252417 on Build GRCh38
rs555064910 Chr.9: 100252215-100252215 on Build GRCh38
rs207470405 Chr.9: 100252407-100252407 on Build GRCh38
rs773049314 Chr.9: 100252390-100252390 on Build GRCh38
rs770533497 Chr.9: 100252092-100252092 on Build GRCh38
rs775686671 Chr.9: 100252308-100252308 on Build GRCh38
rs983325567 Chr.9: 100252126-100252126 on Build GRCh38
rs766212732 Chr.9: 100252307-100252307 on Build GRCh38
rs78126129 Chr.9: 100252138-100252138 on Build GRCh38
rs780632454 Chr.9: 100252361-100252361 on Build GRCh38
rs760637052 Chr.9: 100252311-100252311 on Build GRCh38
rs371389112 Chr.9: 100252480-100252480 on Build GRCh38
rs772924698 Chr.9: 100252274-100252274 on Build GRCh38
rs754479299 Chr.9: 100252344-100252344 on Build GRCh38
rs377720510 Chr.9: 100252465-100252465 on Build GRCh38
rs750852175 Chr.9: 100252456-100252456 on Build GRCh38
rs746490671 Chr.9: 100252245-100252245 on Build GRCh38
rs369363271 Chr.9: 100252261-100252261 on Build GRCh38
rs897310804 Chr.9: 100252108-100252108 on Build GRCh38
rs1053326550 Chr.9: 100252369-100252369 on Build GRCh38
rs774183455 Chr.9: 100252409-100252409 on Build GRCh38
rs780406236 Chr.9: 100252335-100252335 on Build GRCh38
rs757492226 Chr.9: 100252320-100252320 on Build GRCh38
rs779684451 Chr.9: 100252233-100252233 on Build GRCh38
rs985435686 Chr.9: 100252440-100252440 on Build GRCh38
rs1032763941 Chr.9: 100252265-100252265 on Build GRCh38
rs764227386 Chr.9: 100252313-100252313 on Build GRCh38
rs963359158 Chr.9: 100252190-100252190 on Build GRCh38
rs995804715 Chr.9: 100252127-100252127 on Build GRCh38
rs765546333 Chr.9: 100252451-100252451 on Build GRCh38
rs1037206509 Chr.9: 100252083-100252083 on Build GRCh38
rs538746047 Chr.9: 100252384-100252384 on Build GRCh38
rs369133866 Chr.9: 100252189-100252189 on Build GRCh38
rs914508910 Chr.9: 100252406-100252406 on Build GRCh38
rs1001553325 Chr.9: 100252467-100252467 on Build GRCh38
rs536204078 Chr.9: 100252232-100252232 on Build GRCh38
rs973291854 Chr.9: 100252207-100252207 on Build GRCh38
rs763013060 Chr.9: 100252303-100252303 on Build GRCh38
rs140255233 Chr.9: 100252326-100252326 on Build GRCh38
rs774076846 Chr.9: 100252175-100252175 on Build GRCh38
rs373472252 Chr.9: 100252485-100252493 on Build GRCh38
rs374662619 Chr.9: 100252424-100252424 on Build GRCh38
rs543051666 Chr.9: 100252133-100252133 on Build GRCh38

View Full Product Details