Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 74947103-74947600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf40
Entrez Gene ID
55071
Gene Name
chromosome 9 open reading frame 40
Gene Aliases
-
UniGene
Hs.532296
Gene Chromosome Location
Chr.9: 74946583-74952886 on Build GRCh38
Transcripts
NM_017998.2

SNP Detail

SNP ID Chromosome Location
rs1016402384 Chr.9: 74947129-74947129 on Build GRCh38
rs145070454 Chr.9: 74947325-74947325 on Build GRCh38
rs919016508 Chr.9: 74947177-74947177 on Build GRCh38
rs988904373 Chr.9: 74947183-74947184 on Build GRCh38
rs868048218 Chr.9: 74947431-74947431 on Build GRCh38
rs968636229 Chr.9: 74947220-74947220 on Build GRCh38
rs144601043 Chr.9: 74947565-74947565 on Build GRCh38
rs35706355 Chr.9: 74947408-74947409 on Build GRCh38
rs545065609 Chr.9: 74947343-74947343 on Build GRCh38
rs950486994 Chr.9: 74947501-74947501 on Build GRCh38
rs777916094 Chr.9: 74947204-74947204 on Build GRCh38
rs1033169025 Chr.9: 74947414-74947414 on Build GRCh38
rs553542472 Chr.9: 74947463-74947463 on Build GRCh38
rs978651921 Chr.9: 74947252-74947252 on Build GRCh38
rs184061836 Chr.9: 74947405-74947405 on Build GRCh38
rs746789992 Chr.9: 74947320-74947320 on Build GRCh38
rs993094657 Chr.9: 74947563-74947563 on Build GRCh38
rs968334751 Chr.9: 74947537-74947537 on Build GRCh38
rs1017826885 Chr.9: 74947487-74947487 on Build GRCh38
rs937926781 Chr.9: 74947295-74947295 on Build GRCh38
rs1048885242 Chr.9: 74947288-74947288 on Build GRCh38
rs918033784 Chr.9: 74947338-74947338 on Build GRCh38
rs1030479274 Chr.9: 74947417-74947417 on Build GRCh38
rs558952419 Chr.9: 74947526-74947526 on Build GRCh38
rs1006449568 Chr.9: 74947387-74947387 on Build GRCh38
rs927112730 Chr.9: 74947292-74947292 on Build GRCh38
rs958647991 Chr.9: 74947548-74947548 on Build GRCh38
rs1045036 Chr.9: 74947361-74947361 on Build GRCh38
rs189489080 Chr.9: 74947139-74947139 on Build GRCh38
rs900743669 Chr.9: 74947406-74947406 on Build GRCh38
rs927230245 Chr.9: 74947539-74947539 on Build GRCh38
rs184119636 Chr.9: 74947131-74947131 on Build GRCh38
rs568831669 Chr.9: 74947242-74947242 on Build GRCh38
rs878907286 Chr.9: 74947388-74947389 on Build GRCh38
rs965702737 Chr.9: 74947146-74947146 on Build GRCh38
rs556052232 Chr.9: 74947564-74947564 on Build GRCh38
rs1055069580 Chr.9: 74947323-74947323 on Build GRCh38
rs1007787212 Chr.9: 74947434-74947434 on Build GRCh38
rs974523595 Chr.9: 74947142-74947142 on Build GRCh38
rs371464445 Chr.9: 74947428-74947428 on Build GRCh38
rs149950492 Chr.9: 74947165-74947165 on Build GRCh38
rs909026014 Chr.9: 74947543-74947543 on Build GRCh38
rs192575219 Chr.9: 74947316-74947316 on Build GRCh38
rs1030526407 Chr.9: 74947164-74947164 on Build GRCh38
rs188645878 Chr.9: 74947488-74947488 on Build GRCh38
rs984973217 Chr.9: 74947531-74947531 on Build GRCh38
rs978682995 Chr.9: 74947539-74947539 on Build GRCh38
rs903269995 Chr.9: 74947388-74947388 on Build GRCh38
rs886414458 Chr.9: 74947383-74947383 on Build GRCh38
rs140836016 Chr.9: 74947491-74947491 on Build GRCh38
rs1047750913 Chr.9: 74947362-74947362 on Build GRCh38
rs773541202 Chr.9: 74947496-74947496 on Build GRCh38
rs533687645 Chr.9: 74947446-74947446 on Build GRCh38
rs909071135 Chr.9: 74947290-74947290 on Build GRCh38

View Full Product Details