Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 75117490-75117993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OSTF1
Entrez Gene ID
26578
Gene Name
osteoclast stimulating factor 1
Gene Aliases
OSF, SH3P2, bA235O14.1
UniGene
Hs.494192
Gene Chromosome Location
Chr.9: 75088480-75147201 on Build GRCh38
Transcripts
NM_012383.4 XM_006717053.3 XM_006717055.3 XM_011518524.2 XM_011518525.2 XM_017014622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1314913 c.65C>T p.T22M Chr.9: 75117534-75117534 on Build GRCh38
7258272 c.35-9C>A p.? Chr.9: 75117495-75117495 on Build GRCh38
3217265 c.66G>A p.T22T Chr.9: 75117535-75117535 on Build GRCh38
3730686 c.35-8delT p.? Chr.9: 75117496-75117496 on Build GRCh38
1110076 c.77G>T p.R26I Chr.9: 75117546-75117546 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920096413 Chr.9: 75117899-75117899 on Build GRCh38
rs948950446 Chr.9: 75117963-75117963 on Build GRCh38
rs546968482 Chr.9: 75117811-75117811 on Build GRCh38
rs531561050 Chr.9: 75117743-75117743 on Build GRCh38
rs747802004 Chr.9: 75117600-75117600 on Build GRCh38
rs1038589296 Chr.9: 75117532-75117532 on Build GRCh38
rs756350626 Chr.9: 75117545-75117545 on Build GRCh38
rs780681831 Chr.9: 75117876-75117876 on Build GRCh38
rs778009294 Chr.9: 75117546-75117546 on Build GRCh38
rs144005626 Chr.9: 75117787-75117787 on Build GRCh38
rs1003110012 Chr.9: 75117834-75117834 on Build GRCh38
rs987273942 Chr.9: 75117824-75117824 on Build GRCh38
rs749343587 Chr.9: 75117550-75117550 on Build GRCh38
rs1003020793 Chr.9: 75117629-75117629 on Build GRCh38
rs566895806 Chr.9: 75117820-75117820 on Build GRCh38
rs539882763 Chr.9: 75117535-75117535 on Build GRCh38
rs143642916 Chr.9: 75117748-75117748 on Build GRCh38
rs974210783 Chr.9: 75117586-75117586 on Build GRCh38
rs779453127 Chr.9: 75117573-75117573 on Build GRCh38
rs912486451 Chr.9: 75117673-75117673 on Build GRCh38
rs1044000856 Chr.9: 75117583-75117583 on Build GRCh38
rs559727955 Chr.9: 75117618-75117618 on Build GRCh38
rs762747586 Chr.9: 75117617-75117617 on Build GRCh38
rs750433098 Chr.9: 75117823-75117823 on Build GRCh38
rs1042061336 Chr.9: 75117694-75117694 on Build GRCh38
rs367728327 Chr.9: 75117517-75117517 on Build GRCh38
rs200664514 Chr.9: 75117589-75117589 on Build GRCh38
rs535920864 Chr.9: 75117830-75117830 on Build GRCh38
rs996111828 Chr.9: 75117798-75117798 on Build GRCh38
rs932514674 Chr.9: 75117626-75117626 on Build GRCh38
rs974178689 Chr.9: 75117886-75117886 on Build GRCh38
rs138352893 Chr.9: 75117679-75117679 on Build GRCh38
rs746258026 Chr.9: 75117579-75117579 on Build GRCh38
rs538478176 Chr.9: 75117866-75117866 on Build GRCh38
rs548683417 Chr.9: 75117664-75117664 on Build GRCh38
rs571555382 Chr.9: 75117766-75117766 on Build GRCh38
rs767694447 Chr.9: 75117534-75117534 on Build GRCh38
rs147496214 Chr.9: 75117648-75117648 on Build GRCh38
rs754768711 Chr.9: 75117525-75117525 on Build GRCh38
rs757864743 Chr.9: 75117561-75117561 on Build GRCh38
rs575731154 Chr.9: 75117862-75117862 on Build GRCh38
rs974534747 Chr.9: 75117894-75117894 on Build GRCh38
rs776205196 Chr.9: 75117597-75117597 on Build GRCh38
rs41310065 Chr.9: 75117832-75117832 on Build GRCh38
rs765384056 Chr.9: 75117742-75117742 on Build GRCh38

View Full Product Details