Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 85956258-85956733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAA35
Entrez Gene ID
60560
Gene Name
N(alpha)-acetyltransferase 35, NatC auxiliary subunit
Gene Aliases
EGAP, MAK10, MAK10P, bA379P1.1
UniGene
Hs.436098
Gene Chromosome Location
Chr.9: 85941142-86026355 on Build GRCh38
Transcripts
NM_001321881.1 NM_001321882.1 NM_024635.3 XM_005252127.3 XM_017015012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6422041 c.137G>A p.G46E Chr.9: 85956372-85956372 on Build GRCh38
4705368 c.150T>C p.H50H Chr.9: 85956385-85956385 on Build GRCh38
6837454 c.158+1G>C p.? Chr.9: 85956394-85956394 on Build GRCh38
6422040 c.137G>A p.G46E Chr.9: 85956372-85956372 on Build GRCh38
4705367 c.150T>C p.H50H Chr.9: 85956385-85956385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772001121 Chr.9: 85956311-85956311 on Build GRCh38
rs577860594 Chr.9: 85956668-85956668 on Build GRCh38
rs17051499 Chr.9: 85956708-85956708 on Build GRCh38
rs921872717 Chr.9: 85956491-85956491 on Build GRCh38
rs532973324 Chr.9: 85956306-85956306 on Build GRCh38
rs769088597 Chr.9: 85956399-85956402 on Build GRCh38
rs546445274 Chr.9: 85956318-85956318 on Build GRCh38
rs543585364 Chr.9: 85956681-85956681 on Build GRCh38
rs528219122 Chr.9: 85956347-85956347 on Build GRCh38
rs1056494165 Chr.9: 85956613-85956613 on Build GRCh38
rs376680264 Chr.9: 85956321-85956321 on Build GRCh38
rs774760167 Chr.9: 85956416-85956420 on Build GRCh38
rs781464521 Chr.9: 85956418-85956418 on Build GRCh38
rs189719853 Chr.9: 85956423-85956423 on Build GRCh38
rs921089172 Chr.9: 85956676-85956676 on Build GRCh38
rs374327350 Chr.9: 85956611-85956611 on Build GRCh38
rs761417328 Chr.9: 85956328-85956328 on Build GRCh38
rs375272389 Chr.9: 85956455-85956455 on Build GRCh38
rs908231632 Chr.9: 85956701-85956701 on Build GRCh38
rs768556162 Chr.9: 85956441-85956441 on Build GRCh38
rs747940216 Chr.9: 85956343-85956344 on Build GRCh38
rs767208702 Chr.9: 85956441-85956441 on Build GRCh38
rs758727219 Chr.9: 85956344-85956344 on Build GRCh38
rs188086581 Chr.9: 85956592-85956592 on Build GRCh38
rs778737637 Chr.9: 85956316-85956321 on Build GRCh38
rs756357488 Chr.9: 85956421-85956421 on Build GRCh38
rs767010566 Chr.9: 85956347-85956347 on Build GRCh38
rs775348825 Chr.9: 85956315-85956315 on Build GRCh38
rs572709068 Chr.9: 85956471-85956471 on Build GRCh38
rs755140893 Chr.9: 85956416-85956416 on Build GRCh38
rs200018881 Chr.9: 85956440-85956441 on Build GRCh38
rs548688543 Chr.9: 85956500-85956500 on Build GRCh38
rs771804720 Chr.9: 85956425-85956425 on Build GRCh38
rs182622237 Chr.9: 85956452-85956452 on Build GRCh38
rs999290271 Chr.9: 85956591-85956591 on Build GRCh38
rs924601334 Chr.9: 85956293-85956293 on Build GRCh38
rs778133352 Chr.9: 85956492-85956492 on Build GRCh38
rs763514770 Chr.9: 85956347-85956349 on Build GRCh38
rs775493352 Chr.9: 85956347-85956348 on Build GRCh38
rs914623205 Chr.9: 85956608-85956608 on Build GRCh38
rs990245029 Chr.9: 85956603-85956604 on Build GRCh38
rs775434728 Chr.9: 85956434-85956434 on Build GRCh38
rs748239230 Chr.9: 85956419-85956419 on Build GRCh38
rs902251957 Chr.9: 85956515-85956515 on Build GRCh38
rs781425059 Chr.9: 85956346-85956346 on Build GRCh38
rs906164979 Chr.9: 85956684-85956684 on Build GRCh38
rs1013046896 Chr.9: 85956444-85956444 on Build GRCh38
rs141394669 Chr.9: 85956665-85956665 on Build GRCh38
rs750822247 Chr.9: 85956339-85956339 on Build GRCh38
rs374157527 Chr.9: 85956346-85956347 on Build GRCh38
rs971709515 Chr.9: 85956475-85956475 on Build GRCh38
rs746984550 Chr.9: 85956436-85956436 on Build GRCh38
rs896169423 Chr.9: 85956633-85956633 on Build GRCh38
rs995407954 Chr.9: 85956685-85956685 on Build GRCh38
rs758649958 Chr.9: 85956459-85956459 on Build GRCh38
rs535401288 Chr.9: 85956456-85956456 on Build GRCh38
rs946512024 Chr.9: 85956498-85956498 on Build GRCh38
rs764931984 Chr.9: 85956329-85956329 on Build GRCh38
rs771091468 Chr.9: 85956344-85956345 on Build GRCh38
rs370514167 Chr.9: 85956573-85956573 on Build GRCh38
rs752065595 Chr.9: 85956415-85956415 on Build GRCh38
rs955999299 Chr.9: 85956597-85956597 on Build GRCh38
rs886761196 Chr.9: 85956361-85956361 on Build GRCh38
rs199805722 Chr.9: 85956346-85956346 on Build GRCh38
rs745663464 Chr.9: 85956424-85956424 on Build GRCh38
rs776085020 Chr.9: 85956325-85956325 on Build GRCh38
rs370006365 Chr.9: 85956391-85956391 on Build GRCh38
rs368172497 Chr.9: 85956440-85956440 on Build GRCh38
rs920676077 Chr.9: 85956304-85956304 on Build GRCh38

View Full Product Details