Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 85978151-85978631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAA35
Entrez Gene ID
60560
Gene Name
N(alpha)-acetyltransferase 35, NatC auxiliary subunit
Gene Aliases
EGAP, MAK10, MAK10P, bA379P1.1
UniGene
Hs.436098
Gene Chromosome Location
Chr.9: 85941142-86026355 on Build GRCh38
Transcripts
NM_001321881.1 NM_001321882.1 NM_024635.3 XM_005252127.3 XM_011518903.2 XM_011518904.2 XM_017015012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3943101 c.838C>A p.H280N Chr.9: 85978342-85978342 on Build GRCh38
6193924 c.875G>C p.G292A Chr.9: 85978379-85978379 on Build GRCh38
5810841 c.779A>G p.E260G Chr.9: 85978283-85978283 on Build GRCh38
1110742 c.770C>T p.A257V Chr.9: 85978274-85978274 on Build GRCh38
1701285 c.833C>T p.S278L Chr.9: 85978337-85978337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535074389 Chr.9: 85978385-85978385 on Build GRCh38
rs750231441 Chr.9: 85978423-85978423 on Build GRCh38
rs974439884 Chr.9: 85978557-85978557 on Build GRCh38
rs1017871203 Chr.9: 85978571-85978571 on Build GRCh38
rs769558005 Chr.9: 85978395-85978395 on Build GRCh38
rs368919696 Chr.9: 85978285-85978285 on Build GRCh38
rs542628063 Chr.9: 85978442-85978442 on Build GRCh38
rs771980543 Chr.9: 85978253-85978253 on Build GRCh38
rs776157131 Chr.9: 85978431-85978433 on Build GRCh38
rs746523288 Chr.9: 85978388-85978388 on Build GRCh38
rs751686134 Chr.9: 85978526-85978526 on Build GRCh38
rs1009183853 Chr.9: 85978212-85978212 on Build GRCh38
rs765995452 Chr.9: 85978427-85978427 on Build GRCh38
rs762114011 Chr.9: 85978281-85978281 on Build GRCh38
rs200156742 Chr.9: 85978393-85978393 on Build GRCh38
rs755031235 Chr.9: 85978311-85978311 on Build GRCh38
rs760165501 Chr.9: 85978276-85978276 on Build GRCh38
rs745851257 Chr.9: 85978234-85978234 on Build GRCh38
rs199709551 Chr.9: 85978405-85978405 on Build GRCh38
rs978515280 Chr.9: 85978342-85978342 on Build GRCh38
rs201716367 Chr.9: 85978419-85978419 on Build GRCh38
rs1046805334 Chr.9: 85978426-85978426 on Build GRCh38
rs147227765 Chr.9: 85978322-85978322 on Build GRCh38
rs1011403754 Chr.9: 85978488-85978488 on Build GRCh38
rs916375758 Chr.9: 85978399-85978399 on Build GRCh38
rs776297747 Chr.9: 85978280-85978280 on Build GRCh38
rs573994844 Chr.9: 85978437-85978437 on Build GRCh38
rs1021557004 Chr.9: 85978509-85978509 on Build GRCh38
rs758139952 Chr.9: 85978425-85978425 on Build GRCh38
rs907613463 Chr.9: 85978602-85978602 on Build GRCh38
rs202159252 Chr.9: 85978263-85978263 on Build GRCh38
rs745654883 Chr.9: 85978346-85978346 on Build GRCh38
rs202134757 Chr.9: 85978432-85978432 on Build GRCh38
rs186309627 Chr.9: 85978230-85978230 on Build GRCh38
rs966706045 Chr.9: 85978328-85978328 on Build GRCh38
rs954370247 Chr.9: 85978566-85978566 on Build GRCh38
rs760370798 Chr.9: 85978241-85978241 on Build GRCh38
rs770712466 Chr.9: 85978231-85978231 on Build GRCh38
rs939258797 Chr.9: 85978372-85978372 on Build GRCh38
rs182282750 Chr.9: 85978486-85978486 on Build GRCh38
rs150925033 Chr.9: 85978360-85978360 on Build GRCh38
rs1030377285 Chr.9: 85978561-85978561 on Build GRCh38
rs201461279 Chr.9: 85978340-85978340 on Build GRCh38
rs140476279 Chr.9: 85978403-85978403 on Build GRCh38
rs964965014 Chr.9: 85978535-85978535 on Build GRCh38
rs750796057 Chr.9: 85978295-85978295 on Build GRCh38
rs545713900 Chr.9: 85978580-85978580 on Build GRCh38
rs900635318 Chr.9: 85978572-85978572 on Build GRCh38
rs759567937 Chr.9: 85978415-85978415 on Build GRCh38
rs141267660 Chr.9: 85978558-85978558 on Build GRCh38
rs756265726 Chr.9: 85978338-85978338 on Build GRCh38
rs763613794 Chr.9: 85978297-85978297 on Build GRCh38
rs369495876 Chr.9: 85978472-85978472 on Build GRCh38
rs1051190291 Chr.9: 85978482-85978482 on Build GRCh38
rs751698413 Chr.9: 85978310-85978310 on Build GRCh38
rs779996198 Chr.9: 85978428-85978428 on Build GRCh38
rs781294752 Chr.9: 85978313-85978313 on Build GRCh38
rs201778443 Chr.9: 85978325-85978325 on Build GRCh38
rs1011092520 Chr.9: 85978577-85978577 on Build GRCh38
rs752067635 Chr.9: 85978411-85978411 on Build GRCh38
rs186244420 Chr.9: 85978606-85978606 on Build GRCh38
rs577409163 Chr.9: 85978418-85978418 on Build GRCh38
rs747005384 Chr.9: 85978431-85978431 on Build GRCh38
rs1009149269 Chr.9: 85978554-85978554 on Build GRCh38
rs778646321 Chr.9: 85978232-85978232 on Build GRCh38
rs766988271 Chr.9: 85978308-85978308 on Build GRCh38
rs548064043 Chr.9: 85978210-85978210 on Build GRCh38
rs767681620 Chr.9: 85978417-85978417 on Build GRCh38
rs967178543 Chr.9: 85978589-85978589 on Build GRCh38

View Full Product Details