Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 135815753-135816278 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMSAP1
Entrez Gene ID
157922
Gene Name
calmodulin regulated spectrin associated protein 1
Gene Aliases
-
UniGene
Hs.522493
Gene Chromosome Location
Chr.9: 135808487-135907551 on Build GRCh38
Transcripts
NM_015447.3 XM_005263396.3 XM_005263397.1 XM_011518272.1 XM_011518273.2 XM_011518274.1 XM_017014301.1 XM_017014302.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6041090 c.4337_4345delGGGAGACCG p.W1446_A1449>S Chr.9: 135815932-135815940 on Build GRCh38
6619367 c.4378G>A p.E1460K Chr.9: 135815899-135815899 on Build GRCh38
4877671 c.4380G>A p.A1460A Chr.9: 135815930-135815930 on Build GRCh38
6619392 c.4379C>T p.A1460V Chr.9: 135815931-135815931 on Build GRCh38
4416257 c.4420+9G>C p.? Chr.9: 135815881-135815881 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775761873 Chr.9: 135816005-135816005 on Build GRCh38
rs750388632 Chr.9: 135815874-135815874 on Build GRCh38
rs755989405 Chr.9: 135816025-135816026 on Build GRCh38
rs955873672 Chr.9: 135815822-135815822 on Build GRCh38
rs372983445 Chr.9: 135815885-135815885 on Build GRCh38
rs915314150 Chr.9: 135815853-135815853 on Build GRCh38
rs767471389 Chr.9: 135816038-135816038 on Build GRCh38
rs755974195 Chr.9: 135815877-135815877 on Build GRCh38
rs980898149 Chr.9: 135816177-135816177 on Build GRCh38
rs753836022 Chr.9: 135816043-135816043 on Build GRCh38
rs774549300 Chr.9: 135816021-135816021 on Build GRCh38
rs938570224 Chr.9: 135815834-135815834 on Build GRCh38
rs147269559 Chr.9: 135815927-135815927 on Build GRCh38
rs763172257 Chr.9: 135816012-135816012 on Build GRCh38
rs1031572805 Chr.9: 135816083-135816083 on Build GRCh38
rs766685503 Chr.9: 135815840-135815840 on Build GRCh38
rs913342 Chr.9: 135815881-135815881 on Build GRCh38
rs746577186 Chr.9: 135815980-135815980 on Build GRCh38
rs977283555 Chr.9: 135816103-135816103 on Build GRCh38
rs770345894 Chr.9: 135815982-135815982 on Build GRCh38
rs940301587 Chr.9: 135816222-135816225 on Build GRCh38
rs754724416 Chr.9: 135815882-135815882 on Build GRCh38
rs115517832 Chr.9: 135816130-135816130 on Build GRCh38
rs992706148 Chr.9: 135816142-135816142 on Build GRCh38
rs113539678 Chr.9: 135816044-135816044 on Build GRCh38
rs371914527 Chr.9: 135815884-135815884 on Build GRCh38
rs924410084 Chr.9: 135815780-135815780 on Build GRCh38
rs140850961 Chr.9: 135815933-135815933 on Build GRCh38
rs758518511 Chr.9: 135815862-135815862 on Build GRCh38
rs538629814 Chr.9: 135816082-135816082 on Build GRCh38
rs147881781 Chr.9: 135815996-135815996 on Build GRCh38
rs764159651 Chr.9: 135815932-135815932 on Build GRCh38
rs753684895 Chr.9: 135815948-135815948 on Build GRCh38
rs887388663 Chr.9: 135815989-135815989 on Build GRCh38
rs908786238 Chr.9: 135816214-135816214 on Build GRCh38
rs766979420 Chr.9: 135815843-135815844 on Build GRCh38
rs760014604 Chr.9: 135815925-135815925 on Build GRCh38
rs568965230 Chr.9: 135815835-135815835 on Build GRCh38
rs917933156 Chr.9: 135816162-135816162 on Build GRCh38
rs199584430 Chr.9: 135816002-135816002 on Build GRCh38
rs761991213 Chr.9: 135816034-135816034 on Build GRCh38
rs1021762535 Chr.9: 135816066-135816066 on Build GRCh38
rs778522447 Chr.9: 135815957-135815957 on Build GRCh38
rs765557548 Chr.9: 135815858-135815858 on Build GRCh38
rs975912510 Chr.9: 135816151-135816151 on Build GRCh38
rs771728810 Chr.9: 135815893-135815893 on Build GRCh38
rs921237375 Chr.9: 135816165-135816165 on Build GRCh38
rs1038497569 Chr.9: 135816234-135816234 on Build GRCh38
rs1038972131 Chr.9: 135815997-135815997 on Build GRCh38
rs948468588 Chr.9: 135816255-135816255 on Build GRCh38
rs776989736 Chr.9: 135815842-135815842 on Build GRCh38
rs746343516 Chr.9: 135816000-135816000 on Build GRCh38
rs747525428 Chr.9: 135815906-135815906 on Build GRCh38
rs772810211 Chr.9: 135815900-135815900 on Build GRCh38
rs34619268 Chr.9: 135816109-135816110 on Build GRCh38
rs771413183 Chr.9: 135815921-135815921 on Build GRCh38
rs754850459 Chr.9: 135815956-135815956 on Build GRCh38
rs764141075 Chr.9: 135815865-135815865 on Build GRCh38
rs752915410 Chr.9: 135815859-135815859 on Build GRCh38
rs990410902 Chr.9: 135815824-135815825 on Build GRCh38
rs777441006 Chr.9: 135815976-135815976 on Build GRCh38
rs766261153 Chr.9: 135816032-135816032 on Build GRCh38
rs144593848 Chr.9: 135815946-135815946 on Build GRCh38
rs747835528 Chr.9: 135815970-135815970 on Build GRCh38
rs151126173 Chr.9: 135816176-135816176 on Build GRCh38
rs367938620 Chr.9: 135815942-135815942 on Build GRCh38
rs776993104 Chr.9: 135815924-135815924 on Build GRCh38
rs758058618 Chr.9: 135815971-135815971 on Build GRCh38
rs372082294 Chr.9: 135816015-135816015 on Build GRCh38
rs763117472 Chr.9: 135815931-135815931 on Build GRCh38
rs867061500 Chr.9: 135816251-135816251 on Build GRCh38
rs543069501 Chr.9: 135816252-135816252 on Build GRCh38
rs775741548 Chr.9: 135815930-135815930 on Build GRCh38
rs549742967 Chr.9: 135816125-135816125 on Build GRCh38
rs898676954 Chr.9: 135815928-135815928 on Build GRCh38
rs779723893 Chr.9: 135815878-135815878 on Build GRCh38
rs750420561 Chr.9: 135815880-135815882 on Build GRCh38
rs759692542 Chr.9: 135815843-135815843 on Build GRCh38

View Full Product Details