Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137088771-137089172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAN1B1
Entrez Gene ID
11253
Gene Name
mannosidase alpha class 1B member 1
Gene Aliases
ERMAN1, ERManI, MANA-ER, MRT15
UniGene
Hs.279881
Gene Chromosome Location
Chr.9: 137086862-137109187 on Build GRCh38
Transcripts
NM_016219.4 NR_045720.1 NR_045721.1 XM_006716945.3 XM_017014239.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3848104 c.376G>A p.A126T Chr.9: 137088916-137088916 on Build GRCh38
2156333 c.465G>T p.Q155H Chr.9: 137089005-137089005 on Build GRCh38
5998613 c.465+3A>G p.? Chr.9: 137089008-137089008 on Build GRCh38
1319630 c.461C>T p.S154L Chr.9: 137089001-137089001 on Build GRCh38
2156334 c.463C>A p.Q155K Chr.9: 137089003-137089003 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs114461317 Chr.9: 137089067-137089067 on Build GRCh38
rs771177123 Chr.9: 137088949-137088949 on Build GRCh38
rs199738190 Chr.9: 137088966-137088966 on Build GRCh38
rs751295123 Chr.9: 137088908-137088908 on Build GRCh38
rs775457888 Chr.9: 137088845-137088845 on Build GRCh38
rs376808255 Chr.9: 137089046-137089046 on Build GRCh38
rs763520482 Chr.9: 137088970-137088970 on Build GRCh38
rs745680058 Chr.9: 137088920-137088920 on Build GRCh38
rs763358639 Chr.9: 137089024-137089024 on Build GRCh38
rs752411836 Chr.9: 137088979-137088979 on Build GRCh38
rs765617693 Chr.9: 137088965-137088965 on Build GRCh38
rs776849013 Chr.9: 137088958-137088958 on Build GRCh38
rs376313503 Chr.9: 137088989-137088989 on Build GRCh38
rs373259904 Chr.9: 137089015-137089015 on Build GRCh38
rs761422081 Chr.9: 137089032-137089032 on Build GRCh38
rs986628305 Chr.9: 137088831-137088831 on Build GRCh38
rs142586918 Chr.9: 137088942-137088942 on Build GRCh38
rs754010020 Chr.9: 137088843-137088843 on Build GRCh38
rs770914912 Chr.9: 137088823-137088823 on Build GRCh38
rs750977794 Chr.9: 137088868-137088868 on Build GRCh38
rs745613133 Chr.9: 137089034-137089035 on Build GRCh38
rs191443938 Chr.9: 137088993-137088993 on Build GRCh38
rs777414894 Chr.9: 137088903-137088903 on Build GRCh38
rs747045528 Chr.9: 137088946-137088946 on Build GRCh38
rs75442795 Chr.9: 137088943-137088943 on Build GRCh38
rs778838753 Chr.9: 137089055-137089055 on Build GRCh38
rs147529965 Chr.9: 137088877-137088877 on Build GRCh38
rs760858034 Chr.9: 137088875-137088875 on Build GRCh38
rs111492711 Chr.9: 137089029-137089029 on Build GRCh38
rs373546330 Chr.9: 137089054-137089054 on Build GRCh38
rs919921405 Chr.9: 137088904-137088904 on Build GRCh38
rs754933555 Chr.9: 137088998-137088998 on Build GRCh38
rs140105878 Chr.9: 137088999-137088999 on Build GRCh38
rs747100771 Chr.9: 137089004-137089004 on Build GRCh38
rs74967304 Chr.9: 137089028-137089028 on Build GRCh38
rs200572517 Chr.9: 137088878-137088878 on Build GRCh38
rs753185173 Chr.9: 137088888-137088888 on Build GRCh38
rs759835623 Chr.9: 137088959-137088959 on Build GRCh38
rs372332688 Chr.9: 137088972-137088972 on Build GRCh38
rs531368833 Chr.9: 137089099-137089099 on Build GRCh38
rs758036422 Chr.9: 137088890-137088890 on Build GRCh38
rs886063727 Chr.9: 137088981-137088981 on Build GRCh38
rs749774892 Chr.9: 137088807-137088807 on Build GRCh38
rs992384640 Chr.9: 137088950-137088950 on Build GRCh38
rs770944364 Chr.9: 137089010-137089010 on Build GRCh38
rs750083808 Chr.9: 137089035-137089035 on Build GRCh38
rs777562916 Chr.9: 137089111-137089111 on Build GRCh38
rs776771824 Chr.9: 137089012-137089012 on Build GRCh38
rs139630016 Chr.9: 137088973-137088973 on Build GRCh38
rs768831171 Chr.9: 137089037-137089038 on Build GRCh38
rs369827554 Chr.9: 137088915-137088915 on Build GRCh38
rs745866265 Chr.9: 137088828-137088828 on Build GRCh38
rs755870919 Chr.9: 137088940-137088940 on Build GRCh38
rs889682493 Chr.9: 137089098-137089098 on Build GRCh38
rs115335100 Chr.9: 137088887-137088887 on Build GRCh38
rs750077096 Chr.9: 137088988-137088988 on Build GRCh38
rs183157857 Chr.9: 137089053-137089053 on Build GRCh38
rs112610040 Chr.9: 137088820-137088820 on Build GRCh38
rs548483428 Chr.9: 137089123-137089123 on Build GRCh38
rs371270807 Chr.9: 137088871-137088871 on Build GRCh38
rs374795596 Chr.9: 137088879-137088879 on Build GRCh38
rs756945518 Chr.9: 137088914-137088914 on Build GRCh38
rs772127475 Chr.9: 137088859-137088859 on Build GRCh38
rs781101074 Chr.9: 137088827-137088827 on Build GRCh38
rs761813458 Chr.9: 137088851-137088851 on Build GRCh38
rs116668948 Chr.9: 137089066-137089066 on Build GRCh38
rs945341759 Chr.9: 137089146-137089146 on Build GRCh38
rs763744643 Chr.9: 137088971-137088971 on Build GRCh38
rs201813871 Chr.9: 137089040-137089040 on Build GRCh38
rs775899373 Chr.9: 137089021-137089021 on Build GRCh38
rs145520113 Chr.9: 137089033-137089033 on Build GRCh38
rs770092466 Chr.9: 137089016-137089016 on Build GRCh38

View Full Product Details