Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 126881210-126881715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB34
Entrez Gene ID
403341
Gene Name
zinc finger and BTB domain containing 34
Gene Aliases
ZNF918
UniGene
Hs.177633
Gene Chromosome Location
Chr.9: 126860665-126885878 on Build GRCh38
Transcripts
NM_001099270.1 XM_005251989.4 XM_011518699.2

SNP Detail

SNP ID Chromosome Location
rs964967481 Chr.9: 126881312-126881312 on Build GRCh38
rs1048714907 Chr.9: 126881335-126881335 on Build GRCh38
rs149772438 Chr.9: 126881458-126881458 on Build GRCh38
rs755399500 Chr.9: 126881363-126881364 on Build GRCh38
rs974368572 Chr.9: 126881320-126881320 on Build GRCh38
rs983194869 Chr.9: 126881346-126881346 on Build GRCh38
rs888327018 Chr.9: 126881340-126881340 on Build GRCh38
rs956333320 Chr.9: 126881387-126881391 on Build GRCh38
rs779588311 Chr.9: 126881282-126881282 on Build GRCh38
rs1031717231 Chr.9: 126881495-126881495 on Build GRCh38
rs988959576 Chr.9: 126881421-126881421 on Build GRCh38
rs907617618 Chr.9: 126881348-126881348 on Build GRCh38
rs926930630 Chr.9: 126881461-126881461 on Build GRCh38
rs566181593 Chr.9: 126881679-126881679 on Build GRCh38
rs149699043 Chr.9: 126881384-126881386 on Build GRCh38
rs561265032 Chr.9: 126881455-126881455 on Build GRCh38
rs546682192 Chr.9: 126881641-126881641 on Build GRCh38
rs1011694498 Chr.9: 126881529-126881529 on Build GRCh38
rs888315840 Chr.9: 126881636-126881636 on Build GRCh38
rs1017553261 Chr.9: 126881351-126881351 on Build GRCh38
rs546350393 Chr.9: 126881464-126881464 on Build GRCh38
rs1030556435 Chr.9: 126881670-126881670 on Build GRCh38
rs570407847 Chr.9: 126881584-126881584 on Build GRCh38
rs980383864 Chr.9: 126881459-126881459 on Build GRCh38
rs938403611 Chr.9: 126881297-126881297 on Build GRCh38
rs891915087 Chr.9: 126881503-126881503 on Build GRCh38
rs993989901 Chr.9: 126881328-126881328 on Build GRCh38
rs1046365053 Chr.9: 126881423-126881423 on Build GRCh38
rs550340954 Chr.9: 126881472-126881472 on Build GRCh38
rs922966184 Chr.9: 126881338-126881338 on Build GRCh38
rs933002493 Chr.9: 126881339-126881339 on Build GRCh38
rs1003293453 Chr.9: 126881483-126881483 on Build GRCh38
rs1006802819 Chr.9: 126881341-126881341 on Build GRCh38
rs544745109 Chr.9: 126881308-126881308 on Build GRCh38
rs371169040 Chr.9: 126881278-126881278 on Build GRCh38
rs895049227 Chr.9: 126881372-126881372 on Build GRCh38
rs1056909834 Chr.9: 126881304-126881304 on Build GRCh38
rs989404485 Chr.9: 126881265-126881265 on Build GRCh38
rs941868751 Chr.9: 126881360-126881360 on Build GRCh38
rs532923054 Chr.9: 126881596-126881596 on Build GRCh38
rs1021777919 Chr.9: 126881553-126881553 on Build GRCh38
rs914727215 Chr.9: 126881435-126881435 on Build GRCh38
rs964711042 Chr.9: 126881362-126881363 on Build GRCh38
rs901116897 Chr.9: 126881652-126881652 on Build GRCh38
rs1044045506 Chr.9: 126881288-126881288 on Build GRCh38
rs181073919 Chr.9: 126881284-126881284 on Build GRCh38
rs990726653 Chr.9: 126881296-126881296 on Build GRCh38
rs983559613 Chr.9: 126881680-126881680 on Build GRCh38
rs531011623 Chr.9: 126881302-126881302 on Build GRCh38
rs1037571049 Chr.9: 126881364-126881364 on Build GRCh38
rs1048223942 Chr.9: 126881635-126881635 on Build GRCh38
rs947974225 Chr.9: 126881395-126881395 on Build GRCh38
rs756664502 Chr.9: 126881558-126881558 on Build GRCh38
rs79612201 Chr.9: 126881256-126881256 on Build GRCh38
rs1057217656 Chr.9: 126881574-126881574 on Build GRCh38
rs758147548 Chr.9: 126881277-126881277 on Build GRCh38

View Full Product Details