Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128340437-128340976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC27A4
Entrez Gene ID
10999
Gene Name
solute carrier family 27 member 4
Gene Aliases
ACSVL4, FATP4, IPS
UniGene
Hs.656699
Gene Chromosome Location
Chr.9: 128340517-128361470 on Build GRCh38
Transcripts
NM_005094.3 XM_017014222.1

SNP Detail

SNP ID Chromosome Location
rs912920946 Chr.9: 128340640-128340640 on Build GRCh38
rs112132745 Chr.9: 128340656-128340656 on Build GRCh38
rs895232444 Chr.9: 128340506-128340506 on Build GRCh38
rs974570243 Chr.9: 128340609-128340609 on Build GRCh38
rs549113231 Chr.9: 128340667-128340667 on Build GRCh38
rs766711068 Chr.9: 128340782-128340782 on Build GRCh38
rs912860842 Chr.9: 128340471-128340471 on Build GRCh38
rs1019826867 Chr.9: 128340928-128340928 on Build GRCh38
rs562132263 Chr.9: 128340885-128340885 on Build GRCh38
rs1055177986 Chr.9: 128340500-128340500 on Build GRCh38
rs867660166 Chr.9: 128340814-128340814 on Build GRCh38
rs12344475 Chr.9: 128340873-128340873 on Build GRCh38
rs559736012 Chr.9: 128340817-128340817 on Build GRCh38
rs1055226122 Chr.9: 128340704-128340704 on Build GRCh38
rs760777437 Chr.9: 128340781-128340781 on Build GRCh38
rs1020447611 Chr.9: 128340858-128340858 on Build GRCh38
rs996065722 Chr.9: 128340841-128340841 on Build GRCh38
rs113785080 Chr.9: 128340767-128340767 on Build GRCh38
rs978402204 Chr.9: 128340878-128340878 on Build GRCh38
rs949513176 Chr.9: 128340758-128340758 on Build GRCh38
rs572741027 Chr.9: 128340600-128340600 on Build GRCh38
rs577542298 Chr.9: 128340798-128340798 on Build GRCh38
rs7034056 Chr.9: 128340498-128340498 on Build GRCh38
rs965575998 Chr.9: 128340940-128340940 on Build GRCh38
rs1041916490 Chr.9: 128340857-128340857 on Build GRCh38
rs984168656 Chr.9: 128340475-128340475 on Build GRCh38
rs1014051101 Chr.9: 128340520-128340520 on Build GRCh38
rs904061442 Chr.9: 128340495-128340495 on Build GRCh38
rs1016175415 Chr.9: 128340593-128340593 on Build GRCh38
rs772270222 Chr.9: 128340728-128340738 on Build GRCh38
rs886413835 Chr.9: 128340570-128340574 on Build GRCh38
rs754070943 Chr.9: 128340827-128340827 on Build GRCh38
rs543966789 Chr.9: 128340736-128340736 on Build GRCh38
rs759496502 Chr.9: 128340611-128340611 on Build GRCh38
rs189718198 Chr.9: 128340890-128340890 on Build GRCh38
rs761778601 Chr.9: 128340734-128340734 on Build GRCh38
rs925538675 Chr.9: 128340662-128340662 on Build GRCh38
rs551314110 Chr.9: 128340465-128340465 on Build GRCh38
rs991006757 Chr.9: 128340919-128340919 on Build GRCh38
rs963240520 Chr.9: 128340606-128340606 on Build GRCh38
rs1028850831 Chr.9: 128340845-128340845 on Build GRCh38
rs917139713 Chr.9: 128340839-128340839 on Build GRCh38
rs575118505 Chr.9: 128340725-128340725 on Build GRCh38
rs368302025 Chr.9: 128340539-128340539 on Build GRCh38
rs1057066212 Chr.9: 128340808-128340808 on Build GRCh38
rs895274146 Chr.9: 128340708-128340708 on Build GRCh38
rs1003917444 Chr.9: 128340664-128340664 on Build GRCh38
rs987603691 Chr.9: 128340639-128340639 on Build GRCh38
rs528433086 Chr.9: 128340852-128340852 on Build GRCh38
rs1042603474 Chr.9: 128340490-128340490 on Build GRCh38
rs904283120 Chr.9: 128340860-128340860 on Build GRCh38
rs535547240 Chr.9: 128340617-128340617 on Build GRCh38
rs540151462 Chr.9: 128340799-128340799 on Build GRCh38
rs978349540 Chr.9: 128340661-128340661 on Build GRCh38
rs936887507 Chr.9: 128340696-128340696 on Build GRCh38
rs1016228175 Chr.9: 128340815-128340815 on Build GRCh38
rs766898049 Chr.9: 128340952-128340952 on Build GRCh38
rs886470179 Chr.9: 128340774-128340775 on Build GRCh38
rs1004825174 Chr.9: 128340570-128340570 on Build GRCh38
rs945667124 Chr.9: 128340477-128340477 on Build GRCh38
rs937283121 Chr.9: 128340783-128340783 on Build GRCh38
rs552981503 Chr.9: 128340542-128340542 on Build GRCh38
rs1007131332 Chr.9: 128340923-128340923 on Build GRCh38
rs945692628 Chr.9: 128340642-128340642 on Build GRCh38
rs994307836 Chr.9: 128340951-128340951 on Build GRCh38
rs540116397 Chr.9: 128340516-128340516 on Build GRCh38

View Full Product Details