Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128683577-128684077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SET
Entrez Gene ID
6418
Gene Name
SET nuclear proto-oncogene
Gene Aliases
2PP2A, I2PP2A, IGAAD, IPP2A2, PHAPII, TAF-I, TAF-IBETA
UniGene
Hs.436687
Gene Chromosome Location
Chr.9: 128683432-128696396 on Build GRCh38
Transcripts
NM_001122821.1 XM_017015013.1 XM_017015014.1 XM_017015015.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
397908 c.32C>T p.P11L Chr.9: 128683927-128683927 on Build GRCh38
1105691 c.92C>T p.A31V Chr.9: 128683987-128683987 on Build GRCh38
6455965 c.108G>A p.K36K Chr.9: 128684003-128684003 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs979757073 Chr.9: 128683728-128683728 on Build GRCh38
rs1004908283 Chr.9: 128683830-128683830 on Build GRCh38
rs959100010 Chr.9: 128683732-128683732 on Build GRCh38
rs1017083097 Chr.9: 128683726-128683726 on Build GRCh38
rs551070582 Chr.9: 128683808-128683808 on Build GRCh38
rs992067058 Chr.9: 128683924-128683924 on Build GRCh38
rs942588019 Chr.9: 128684008-128684008 on Build GRCh38
rs567108916 Chr.9: 128683876-128683876 on Build GRCh38
rs539856262 Chr.9: 128683662-128683662 on Build GRCh38
rs887796117 Chr.9: 128683967-128683967 on Build GRCh38
rs766762056 Chr.9: 128683932-128683934 on Build GRCh38
rs914936171 Chr.9: 128683966-128683966 on Build GRCh38
rs896540098 Chr.9: 128684043-128684043 on Build GRCh38
rs916815536 Chr.9: 128683712-128683712 on Build GRCh38
rs376347261 Chr.9: 128683775-128683775 on Build GRCh38
rs935625468 Chr.9: 128683888-128683888 on Build GRCh38
rs944533515 Chr.9: 128683923-128683923 on Build GRCh38
rs988578769 Chr.9: 128683909-128683909 on Build GRCh38
rs755125117 Chr.9: 128684023-128684024 on Build GRCh38
rs531568851 Chr.9: 128683787-128683787 on Build GRCh38
rs768688269 Chr.9: 128683658-128683658 on Build GRCh38
rs1039933239 Chr.9: 128684012-128684012 on Build GRCh38
rs901182671 Chr.9: 128683615-128683615 on Build GRCh38
rs991764624 Chr.9: 128683736-128683736 on Build GRCh38
rs971609222 Chr.9: 128683930-128683930 on Build GRCh38
rs762111997 Chr.9: 128683669-128683669 on Build GRCh38
rs1040319888 Chr.9: 128683766-128683766 on Build GRCh38
rs761107553 Chr.9: 128683985-128683985 on Build GRCh38
rs1053923658 Chr.9: 128683819-128683819 on Build GRCh38
rs762143442 Chr.9: 128683743-128683744 on Build GRCh38
rs546689765 Chr.9: 128683855-128683855 on Build GRCh38
rs1052969839 Chr.9: 128683744-128683745 on Build GRCh38
rs746550100 Chr.9: 128683851-128683851 on Build GRCh38
rs373867950 Chr.9: 128684031-128684031 on Build GRCh38
rs900458307 Chr.9: 128683951-128683951 on Build GRCh38
rs777332031 Chr.9: 128684019-128684019 on Build GRCh38
rs756407197 Chr.9: 128683982-128683982 on Build GRCh38
rs577760252 Chr.9: 128683754-128683754 on Build GRCh38
rs764797333 Chr.9: 128684051-128684051 on Build GRCh38
rs996616384 Chr.9: 128683784-128683784 on Build GRCh38
rs191311210 Chr.9: 128683961-128683961 on Build GRCh38
rs1027655297 Chr.9: 128683796-128683796 on Build GRCh38
rs917452550 Chr.9: 128683738-128683738 on Build GRCh38
rs752300146 Chr.9: 128683988-128683988 on Build GRCh38
rs958906816 Chr.9: 128683901-128683901 on Build GRCh38
rs751632213 Chr.9: 128683949-128683949 on Build GRCh38
rs952156563 Chr.9: 128683675-128683675 on Build GRCh38
rs983622496 Chr.9: 128683689-128683689 on Build GRCh38
rs923629041 Chr.9: 128683940-128683940 on Build GRCh38
rs923720878 Chr.9: 128683777-128683777 on Build GRCh38
rs949096582 Chr.9: 128683741-128683741 on Build GRCh38
rs145061646 Chr.9: 128683841-128683841 on Build GRCh38
rs533332313 Chr.9: 128683852-128683852 on Build GRCh38
rs750909946 Chr.9: 128683979-128683979 on Build GRCh38
rs754087314 Chr.9: 128684007-128684007 on Build GRCh38
rs981735724 Chr.9: 128683762-128683762 on Build GRCh38
rs887881022 Chr.9: 128683797-128683797 on Build GRCh38
rs746524540 Chr.9: 128684030-128684030 on Build GRCh38
rs981767052 Chr.9: 128683939-128683939 on Build GRCh38
rs1040201938 Chr.9: 128683926-128683926 on Build GRCh38
rs573855930 Chr.9: 128683699-128683699 on Build GRCh38
rs769722562 Chr.9: 128683866-128683866 on Build GRCh38
rs942493113 Chr.9: 128683831-128683831 on Build GRCh38
rs369353971 Chr.9: 128683925-128683925 on Build GRCh38
rs948392879 Chr.9: 128683721-128683721 on Build GRCh38
rs749359159 Chr.9: 128683653-128683653 on Build GRCh38
rs542952710 Chr.9: 128683757-128683757 on Build GRCh38
rs758023705 Chr.9: 128683999-128683999 on Build GRCh38
rs1014964919 Chr.9: 128684041-128684041 on Build GRCh38
rs933876922 Chr.9: 128684050-128684050 on Build GRCh38
rs912947587 Chr.9: 128683919-128683919 on Build GRCh38
rs538070221 Chr.9: 128684020-128684020 on Build GRCh38
rs138894709 Chr.9: 128683960-128683960 on Build GRCh38
rs935064827 Chr.9: 128683795-128683795 on Build GRCh38
rs1017402174 Chr.9: 128683899-128683899 on Build GRCh38
rs1005650641 Chr.9: 128683873-128683873 on Build GRCh38
rs893652810 Chr.9: 128683822-128683822 on Build GRCh38
rs776499093 Chr.9: 128683856-128683856 on Build GRCh38
rs564498550 Chr.9: 128683694-128683694 on Build GRCh38
rs759360846 Chr.9: 128683865-128683865 on Build GRCh38
rs775785887 Chr.9: 128683869-128683869 on Build GRCh38
rs992380539 Chr.9: 128683701-128683701 on Build GRCh38

View Full Product Details