Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129803046-129803535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOR1B
Entrez Gene ID
27348
Gene Name
torsin family 1 member B
Gene Aliases
DQ1
UniGene
Hs.252682
Gene Chromosome Location
Chr.9: 129803143-129811424 on Build GRCh38
Transcripts
NM_001317893.1 NM_001317894.1 NM_014506.2 NR_133948.1 NR_134027.1 NR_134113.1 XM_005251927.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6806316 c.124G>T p.G42C Chr.9: 129803336-129803336 on Build GRCh38
1624616 c.133T>C p.S45P Chr.9: 129803345-129803345 on Build GRCh38
6806313 c.191A>G p.N64S Chr.9: 129803403-129803403 on Build GRCh38
5963710 c.108C>T p.A36A Chr.9: 129803320-129803320 on Build GRCh38
3779727 c.163G>A p.E55K Chr.9: 129803375-129803375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763298816 Chr.9: 129803173-129803173 on Build GRCh38
rs920866091 Chr.9: 129803106-129803106 on Build GRCh38
rs759964640 Chr.9: 129803274-129803274 on Build GRCh38
rs930622472 Chr.9: 129803480-129803480 on Build GRCh38
rs956267482 Chr.9: 129803418-129803418 on Build GRCh38
rs749033850 Chr.9: 129803236-129803236 on Build GRCh38
rs771230892 Chr.9: 129803170-129803170 on Build GRCh38
rs372151069 Chr.9: 129803409-129803409 on Build GRCh38
rs1022595881 Chr.9: 129803139-129803139 on Build GRCh38
rs1052559349 Chr.9: 129803160-129803160 on Build GRCh38
rs890317313 Chr.9: 129803204-129803204 on Build GRCh38
rs777926994 Chr.9: 129803407-129803407 on Build GRCh38
rs939878193 Chr.9: 129803511-129803511 on Build GRCh38
rs758064306 Chr.9: 129803201-129803201 on Build GRCh38
rs948268039 Chr.9: 129803121-129803121 on Build GRCh38
rs551922485 Chr.9: 129803212-129803212 on Build GRCh38
rs540150293 Chr.9: 129803104-129803104 on Build GRCh38
rs774585480 Chr.9: 129803175-129803175 on Build GRCh38
rs576450886 Chr.9: 129803200-129803200 on Build GRCh38
rs372426151 Chr.9: 129803163-129803163 on Build GRCh38
rs751211813 Chr.9: 129803205-129803205 on Build GRCh38
rs959726673 Chr.9: 129803113-129803113 on Build GRCh38
rs756091262 Chr.9: 129803293-129803293 on Build GRCh38
rs748433813 Chr.9: 129803311-129803311 on Build GRCh38
rs766798336 Chr.9: 129803197-129803197 on Build GRCh38
rs974718185 Chr.9: 129803188-129803188 on Build GRCh38
rs928189827 Chr.9: 129803260-129803260 on Build GRCh38
rs765610206 Chr.9: 129803366-129803366 on Build GRCh38
rs763158737 Chr.9: 129803448-129803463 on Build GRCh38
rs969019204 Chr.9: 129803231-129803231 on Build GRCh38
rs3816260 Chr.9: 129803082-129803082 on Build GRCh38
rs770193541 Chr.9: 129803447-129803454 on Build GRCh38
rs867971690 Chr.9: 129803457-129803457 on Build GRCh38
rs76154399 Chr.9: 129803079-129803079 on Build GRCh38
rs866427260 Chr.9: 129803223-129803223 on Build GRCh38
rs772120016 Chr.9: 129803314-129803314 on Build GRCh38
rs537599585 Chr.9: 129803427-129803427 on Build GRCh38
rs1009135784 Chr.9: 129803209-129803209 on Build GRCh38
rs754928353 Chr.9: 129803401-129803401 on Build GRCh38
rs772779058 Chr.9: 129803438-129803438 on Build GRCh38
rs780543185 Chr.9: 129803211-129803211 on Build GRCh38
rs770997150 Chr.9: 129803349-129803349 on Build GRCh38
rs974772223 Chr.9: 129803475-129803475 on Build GRCh38
rs767082510 Chr.9: 129803285-129803285 on Build GRCh38
rs766817980 Chr.9: 129803258-129803258 on Build GRCh38
rs535939331 Chr.9: 129803281-129803281 on Build GRCh38
rs777166035 Chr.9: 129803352-129803352 on Build GRCh38
rs769675081 Chr.9: 129803167-129803167 on Build GRCh38
rs984096457 Chr.9: 129803483-129803483 on Build GRCh38
rs771891117 Chr.9: 129803203-129803205 on Build GRCh38
rs775673014 Chr.9: 129803367-129803367 on Build GRCh38
rs375088316 Chr.9: 129803448-129803448 on Build GRCh38
rs939513533 Chr.9: 129803156-129803156 on Build GRCh38
rs577481907 Chr.9: 129803459-129803459 on Build GRCh38
rs747120330 Chr.9: 129803335-129803335 on Build GRCh38
rs761039008 Chr.9: 129803196-129803196 on Build GRCh38
rs772218416 Chr.9: 129803239-129803239 on Build GRCh38
rs987275098 Chr.9: 129803067-129803067 on Build GRCh38
rs754870373 Chr.9: 129803304-129803304 on Build GRCh38
rs747841289 Chr.9: 129803404-129803404 on Build GRCh38
rs745759349 Chr.9: 129803166-129803166 on Build GRCh38
rs964784018 Chr.9: 129803473-129803473 on Build GRCh38
rs932618689 Chr.9: 129803317-129803317 on Build GRCh38
rs753851057 Chr.9: 129803399-129803399 on Build GRCh38
rs546986970 Chr.9: 129803105-129803105 on Build GRCh38
rs540159997 Chr.9: 129803508-129803508 on Build GRCh38
rs980913855 Chr.9: 129803123-129803123 on Build GRCh38
rs756830163 Chr.9: 129803210-129803210 on Build GRCh38
rs761527185 Chr.9: 129803373-129803373 on Build GRCh38
rs750076924 Chr.9: 129803390-129803390 on Build GRCh38
rs375663995 Chr.9: 129803215-129803215 on Build GRCh38
rs904446167 Chr.9: 129803168-129803168 on Build GRCh38
rs766020134 Chr.9: 129803451-129803451 on Build GRCh38
rs141359830 Chr.9: 129803305-129803305 on Build GRCh38
rs746773468 Chr.9: 129803416-129803417 on Build GRCh38
rs10988518 Chr.9: 129803372-129803372 on Build GRCh38
rs770216432 Chr.9: 129803424-129803424 on Build GRCh38
rs557942444 Chr.9: 129803447-129803447 on Build GRCh38
rs920636711 Chr.9: 129803479-129803479 on Build GRCh38
rs756806983 Chr.9: 129803283-129803283 on Build GRCh38
rs754333031 Chr.9: 129803263-129803263 on Build GRCh38
rs960916960 Chr.9: 129803261-129803261 on Build GRCh38
rs776640472 Chr.9: 129803416-129803416 on Build GRCh38
rs750988877 Chr.9: 129803195-129803195 on Build GRCh38
rs773358359 Chr.9: 129803326-129803326 on Build GRCh38
rs962078935 Chr.9: 129803214-129803214 on Build GRCh38
rs775371252 Chr.9: 129803172-129803172 on Build GRCh38
rs201987743 Chr.9: 129803415-129803415 on Build GRCh38
rs894422 Chr.9: 129803333-129803333 on Build GRCh38
rs575395775 Chr.9: 129803371-129803371 on Build GRCh38
rs761919974 Chr.9: 129803238-129803238 on Build GRCh38
rs879398593 Chr.9: 129803460-129803460 on Build GRCh38
rs780196760 Chr.9: 129803297-129803297 on Build GRCh38
rs753332525 Chr.9: 129803455-129803455 on Build GRCh38
rs764512460 Chr.9: 129803430-129803430 on Build GRCh38
rs769573849 Chr.9: 129803216-129803216 on Build GRCh38
rs760011644 Chr.9: 129803361-129803361 on Build GRCh38
rs544515552 Chr.9: 129803466-129803473 on Build GRCh38
rs777463909 Chr.9: 129803202-129803202 on Build GRCh38
rs773126552 Chr.9: 129803359-129803360 on Build GRCh38
rs149815190 Chr.9: 129803397-129803397 on Build GRCh38
rs939826062 Chr.9: 129803233-129803233 on Build GRCh38
rs768931905 Chr.9: 129803174-129803174 on Build GRCh38
rs758202947 Chr.9: 129803406-129803406 on Build GRCh38
rs761249203 Chr.9: 129803257-129803257 on Build GRCh38
rs767019511 Chr.9: 129803385-129803385 on Build GRCh38
rs151251684 Chr.9: 129803448-129803455 on Build GRCh38
rs749835044 Chr.9: 129803288-129803288 on Build GRCh38
rs773126083 Chr.9: 129803248-129803248 on Build GRCh38
rs943423239 Chr.9: 129803183-129803183 on Build GRCh38
rs368352137 Chr.9: 129803280-129803280 on Build GRCh38
rs778856394 Chr.9: 129803403-129803403 on Build GRCh38
rs774779742 Chr.9: 129803237-129803237 on Build GRCh38
rs749232674 Chr.9: 129803300-129803300 on Build GRCh38
rs1027243266 Chr.9: 129803222-129803222 on Build GRCh38
rs779998878 Chr.9: 129803235-129803235 on Build GRCh38
rs928189487 Chr.9: 129803151-129803151 on Build GRCh38
rs1008752197 Chr.9: 129803387-129803387 on Build GRCh38
rs913945957 Chr.9: 129803162-129803162 on Build GRCh38
rs886971453 Chr.9: 129803228-129803228 on Build GRCh38
rs921303147 Chr.9: 129803312-129803312 on Build GRCh38
rs767465452 Chr.9: 129803176-129803176 on Build GRCh38
rs765955593 Chr.9: 129803398-129803398 on Build GRCh38

View Full Product Details