Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 53536063-53536565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HUWE1
Entrez Gene ID
10075
Gene Name
HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase
Gene Aliases
ARF-BP1, HECTH9, HSPC272, Ib772, LASU1, MULE, URE-B1, UREB1
UniGene
Hs.136905
Gene Chromosome Location
Chr.X: 53532096-53686729 on Build GRCh38
Transcripts
NM_031407.6 XM_005261965.3 XM_011530747.1 XM_011530751.2 XM_017029191.1 XM_017029192.1 XM_017029193.1 XM_017029194.1 XM_017029195.1 XM_017029196.1 XM_017029197.1 XM_017029198.1 XM_017029199.1 XM_017029200.1 XM_017029201.1 XM_017029202.1 XM_017029203.1 XM_017029204.1 XM_017029205.1 XM_017029206.1

SNP Detail

SNP ID Chromosome Location
rs782702314 Chr.X: 53536343-53536343 on Build GRCh38
rs782204774 Chr.X: 53536100-53536100 on Build GRCh38
rs182978092 Chr.X: 53536323-53536323 on Build GRCh38
rs782687065 Chr.X: 53536292-53536292 on Build GRCh38
rs371060814 Chr.X: 53536451-53536451 on Build GRCh38
rs782799129 Chr.X: 53536153-53536153 on Build GRCh38
rs782148553 Chr.X: 53536526-53536526 on Build GRCh38
rs921263797 Chr.X: 53536361-53536361 on Build GRCh38
rs369848701 Chr.X: 53536104-53536104 on Build GRCh38
rs782649014 Chr.X: 53536106-53536106 on Build GRCh38
rs782092697 Chr.X: 53536297-53536297 on Build GRCh38
rs375367345 Chr.X: 53536475-53536475 on Build GRCh38
rs782019798 Chr.X: 53536159-53536159 on Build GRCh38
rs782576498 Chr.X: 53536418-53536418 on Build GRCh38
rs782333060 Chr.X: 53536528-53536528 on Build GRCh38
rs782133761 Chr.X: 53536341-53536341 on Build GRCh38
rs782473788 Chr.X: 53536360-53536360 on Build GRCh38
rs782259876 Chr.X: 53536471-53536471 on Build GRCh38
rs781909094 Chr.X: 53536466-53536466 on Build GRCh38
rs150862364 Chr.X: 53536463-53536463 on Build GRCh38
rs372891869 Chr.X: 53536274-53536274 on Build GRCh38
rs782411460 Chr.X: 53536111-53536111 on Build GRCh38
rs376506320 Chr.X: 53536350-53536350 on Build GRCh38
rs781956427 Chr.X: 53536340-53536340 on Build GRCh38
rs781814063 Chr.X: 53536276-53536276 on Build GRCh38
rs782042293 Chr.X: 53536496-53536496 on Build GRCh38
rs932921780 Chr.X: 53536424-53536424 on Build GRCh38
rs782693101 Chr.X: 53536176-53536176 on Build GRCh38
rs782387942 Chr.X: 53536259-53536259 on Build GRCh38
rs2234477 Chr.X: 53536301-53536301 on Build GRCh38
rs1006001849 Chr.X: 53536089-53536089 on Build GRCh38
rs143059303 Chr.X: 53536246-53536246 on Build GRCh38
rs781930538 Chr.X: 53536296-53536296 on Build GRCh38
rs372560147 Chr.X: 53536108-53536108 on Build GRCh38
rs781918213 Chr.X: 53536291-53536292 on Build GRCh38
rs782405497 Chr.X: 53536336-53536336 on Build GRCh38
rs782035613 Chr.X: 53536342-53536343 on Build GRCh38
rs782362460 Chr.X: 53536129-53536129 on Build GRCh38
rs782606084 Chr.X: 53536097-53536097 on Build GRCh38
rs1015068822 Chr.X: 53536090-53536090 on Build GRCh38
rs782766541 Chr.X: 53536371-53536371 on Build GRCh38
rs782806655 Chr.X: 53536337-53536337 on Build GRCh38
rs782435818 Chr.X: 53536287-53536287 on Build GRCh38
rs369621691 Chr.X: 53536249-53536249 on Build GRCh38
rs782366300 Chr.X: 53536290-53536290 on Build GRCh38
rs980649233 Chr.X: 53536281-53536281 on Build GRCh38
rs781831162 Chr.X: 53536195-53536195 on Build GRCh38
rs781935670 Chr.X: 53536112-53536112 on Build GRCh38
rs782459306 Chr.X: 53536228-53536228 on Build GRCh38
rs199784760 Chr.X: 53536294-53536294 on Build GRCh38
rs782119171 Chr.X: 53536331-53536331 on Build GRCh38
rs782111122 Chr.X: 53536139-53536139 on Build GRCh38
rs3761631 Chr.X: 53536535-53536535 on Build GRCh38
rs782542384 Chr.X: 53536373-53536373 on Build GRCh38
rs781954633 Chr.X: 53536132-53536132 on Build GRCh38
rs782340208 Chr.X: 53536298-53536298 on Build GRCh38
rs782208503 Chr.X: 53536174-53536174 on Build GRCh38
rs782654746 Chr.X: 53536363-53536363 on Build GRCh38
rs781920310 Chr.X: 53536295-53536295 on Build GRCh38
rs587780358 Chr.X: 53536530-53536530 on Build GRCh38
rs782197749 Chr.X: 53536292-53536292 on Build GRCh38

View Full Product Details