Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 54951823-54952321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PFKFB1
Entrez Gene ID
5207
Gene Name
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 1
Gene Aliases
F6PK, HL2K, PFRX
UniGene
Hs.444304
Gene Chromosome Location
Chr.X: 54933134-54998534 on Build GRCh38
Transcripts
NM_001271804.1 NM_001271805.1 NM_002625.3 NR_073450.1 XM_017029576.1 XM_017029577.1 XM_017029578.1

SNP Detail

SNP ID Chromosome Location
rs754699760 Chr.X: 54951867-54951867 on Build GRCh38
rs770176292 Chr.X: 54952287-54952287 on Build GRCh38
rs749925058 Chr.X: 54951914-54951914 on Build GRCh38
rs146369182 Chr.X: 54951990-54951990 on Build GRCh38
rs769684822 Chr.X: 54952075-54952075 on Build GRCh38
rs189321628 Chr.X: 54952174-54952174 on Build GRCh38
rs763051175 Chr.X: 54952084-54952084 on Build GRCh38
rs767193275 Chr.X: 54951978-54951978 on Build GRCh38
rs370162232 Chr.X: 54952150-54952150 on Build GRCh38
rs372559818 Chr.X: 54952124-54952124 on Build GRCh38
rs967323920 Chr.X: 54952143-54952143 on Build GRCh38
rs34971789 Chr.X: 54951895-54951895 on Build GRCh38
rs777399343 Chr.X: 54951997-54951997 on Build GRCh38
rs750912548 Chr.X: 54952115-54952115 on Build GRCh38
rs780853261 Chr.X: 54951884-54951884 on Build GRCh38
rs148350116 Chr.X: 54951941-54951941 on Build GRCh38
rs773039614 Chr.X: 54952117-54952117 on Build GRCh38
rs201795827 Chr.X: 54951916-54951916 on Build GRCh38
rs771400427 Chr.X: 54952032-54952033 on Build GRCh38
rs748456222 Chr.X: 54952068-54952068 on Build GRCh38
rs766304510 Chr.X: 54951962-54951962 on Build GRCh38
rs765862449 Chr.X: 54952175-54952175 on Build GRCh38
rs372621668 Chr.X: 54952241-54952241 on Build GRCh38
rs149578224 Chr.X: 54951913-54951913 on Build GRCh38
rs143062179 Chr.X: 54952016-54952016 on Build GRCh38
rs199798468 Chr.X: 54952158-54952158 on Build GRCh38
rs989776155 Chr.X: 54952100-54952100 on Build GRCh38
rs755422795 Chr.X: 54952157-54952157 on Build GRCh38
rs780740994 Chr.X: 54952035-54952035 on Build GRCh38
rs763659560 Chr.X: 54952127-54952127 on Build GRCh38
rs199630999 Chr.X: 54952057-54952057 on Build GRCh38
rs143311323 Chr.X: 54951940-54951940 on Build GRCh38
rs56031427 Chr.X: 54952006-54952006 on Build GRCh38
rs940967286 Chr.X: 54952283-54952283 on Build GRCh38
rs769919634 Chr.X: 54952074-54952074 on Build GRCh38
rs916627232 Chr.X: 54952183-54952183 on Build GRCh38
rs376959812 Chr.X: 54951945-54951945 on Build GRCh38
rs767413817 Chr.X: 54952114-54952114 on Build GRCh38
rs748463335 Chr.X: 54952163-54952163 on Build GRCh38
rs769394640 Chr.X: 54951898-54951898 on Build GRCh38
rs141537723 Chr.X: 54951971-54951971 on Build GRCh38
rs138301588 Chr.X: 54951911-54951911 on Build GRCh38
rs761648445 Chr.X: 54951996-54951996 on Build GRCh38
rs943846283 Chr.X: 54952251-54952251 on Build GRCh38
rs774321022 Chr.X: 54952087-54952087 on Build GRCh38
rs770987452 Chr.X: 54951864-54951864 on Build GRCh38
rs887770484 Chr.X: 54952260-54952260 on Build GRCh38
rs751021512 Chr.X: 54952261-54952261 on Build GRCh38
rs139743666 Chr.X: 54951992-54951992 on Build GRCh38
rs56761102 Chr.X: 54952177-54952177 on Build GRCh38
rs762959184 Chr.X: 54951946-54951946 on Build GRCh38
rs948057563 Chr.X: 54952213-54952213 on Build GRCh38
rs200363461 Chr.X: 54952155-54952155 on Build GRCh38
rs766396112 Chr.X: 54952085-54952085 on Build GRCh38
rs899542183 Chr.X: 54952263-54952263 on Build GRCh38
rs369550583 Chr.X: 54952060-54952060 on Build GRCh38
rs183772570 Chr.X: 54952132-54952132 on Build GRCh38
rs769195741 Chr.X: 54952036-54952036 on Build GRCh38
rs747386480 Chr.X: 54951915-54951915 on Build GRCh38
rs753504330 Chr.X: 54952128-54952128 on Build GRCh38
rs997697891 Chr.X: 54952050-54952050 on Build GRCh38
rs751457409 Chr.X: 54951970-54951970 on Build GRCh38

View Full Product Details