Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 56993619-56994116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPIN3
Entrez Gene ID
169981
Gene Name
spindlin family member 3
Gene Aliases
SPIN-3, TDRD27, bA445O16.1
UniGene
Hs.522672
Gene Chromosome Location
Chr.X: 56976370-56995801 on Build GRCh38
Transcripts
NM_001010862.2 NR_027139.1 XM_006724577.3 XM_006724578.3 XM_017029315.1

SNP Detail

SNP ID Chromosome Location
rs533572753 Chr.X: 56993912-56993912 on Build GRCh38
rs1013457355 Chr.X: 56994092-56994092 on Build GRCh38
rs1016688561 Chr.X: 56993804-56993804 on Build GRCh38
rs1052205368 Chr.X: 56994036-56994036 on Build GRCh38
rs969699878 Chr.X: 56993732-56993732 on Build GRCh38
rs772724964 Chr.X: 56993870-56993870 on Build GRCh38
rs933520494 Chr.X: 56993921-56993921 on Build GRCh38
rs892205235 Chr.X: 56994085-56994085 on Build GRCh38
rs987592630 Chr.X: 56993944-56993944 on Build GRCh38
rs186414056 Chr.X: 56994024-56994024 on Build GRCh38
rs759597535 Chr.X: 56993997-56993997 on Build GRCh38
rs988788969 Chr.X: 56993933-56993933 on Build GRCh38
rs978823766 Chr.X: 56993909-56993909 on Build GRCh38
rs1023110711 Chr.X: 56993800-56993800 on Build GRCh38
rs910844231 Chr.X: 56993958-56993958 on Build GRCh38
rs766393513 Chr.X: 56994029-56994030 on Build GRCh38
rs771781014 Chr.X: 56993970-56993970 on Build GRCh38
rs963384389 Chr.X: 56993851-56993851 on Build GRCh38
rs944595384 Chr.X: 56994031-56994031 on Build GRCh38
rs993015494 Chr.X: 56993753-56993753 on Build GRCh38
rs762373964 Chr.X: 56993900-56993900 on Build GRCh38
rs911816493 Chr.X: 56993981-56993981 on Build GRCh38
rs968000141 Chr.X: 56993785-56993785 on Build GRCh38
rs926009883 Chr.X: 56993916-56993916 on Build GRCh38
rs183567548 Chr.X: 56993924-56993924 on Build GRCh38
rs903528985 Chr.X: 56994029-56994029 on Build GRCh38
rs943620311 Chr.X: 56993979-56993979 on Build GRCh38
rs970160301 Chr.X: 56993855-56993855 on Build GRCh38
rs1325856 Chr.X: 56993682-56993682 on Build GRCh38

View Full Product Details