Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 65529122-65529619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LAS1L
Entrez Gene ID
81887
Gene Name
LAS1 like, ribosome biogenesis factor
Gene Aliases
Las1-like, dJ475B7.2
UniGene
Hs.522675
Gene Chromosome Location
Chr.X: 65512582-65534806 on Build GRCh38
Transcripts
NM_001170649.1 NM_001170650.1 NM_031206.4 XM_005262301.1 XM_005262304.1 XM_005262305.3 XM_005262306.3 XM_011531045.1 XM_011531046.1 XM_017029877.1 XM_017029878.1 XM_017029879.1 XM_017029880.1

SNP Detail

SNP ID Chromosome Location
rs762778439 Chr.X: 65529283-65529283 on Build GRCh38
rs756257847 Chr.X: 65529568-65529568 on Build GRCh38
rs775979610 Chr.X: 65529221-65529221 on Build GRCh38
rs759123464 Chr.X: 65529233-65529233 on Build GRCh38
rs188092641 Chr.X: 65529411-65529411 on Build GRCh38
rs777771709 Chr.X: 65529346-65529346 on Build GRCh38
rs775705564 Chr.X: 65529440-65529440 on Build GRCh38
rs764975723 Chr.X: 65529234-65529234 on Build GRCh38
rs886261823 Chr.X: 65529443-65529443 on Build GRCh38
rs938348974 Chr.X: 65529303-65529303 on Build GRCh38
rs1008197828 Chr.X: 65529511-65529511 on Build GRCh38
rs769071260 Chr.X: 65529187-65529187 on Build GRCh38
rs753519650 Chr.X: 65529465-65529465 on Build GRCh38
rs781467975 Chr.X: 65529327-65529328 on Build GRCh38
rs927301396 Chr.X: 65529146-65529147 on Build GRCh38
rs1057160106 Chr.X: 65529295-65529295 on Build GRCh38
rs750441529 Chr.X: 65529565-65529565 on Build GRCh38
rs749511994 Chr.X: 65529578-65529578 on Build GRCh38
rs780297234 Chr.X: 65529173-65529173 on Build GRCh38
rs192588893 Chr.X: 65529500-65529500 on Build GRCh38
rs892695459 Chr.X: 65529325-65529325 on Build GRCh38
rs183266168 Chr.X: 65529389-65529389 on Build GRCh38
rs1028387702 Chr.X: 65529471-65529471 on Build GRCh38
rs373716165 Chr.X: 65529555-65529555 on Build GRCh38
rs201435926 Chr.X: 65529305-65529305 on Build GRCh38
rs763986446 Chr.X: 65529293-65529293 on Build GRCh38
rs779164715 Chr.X: 65529584-65529584 on Build GRCh38
rs1009779793 Chr.X: 65529326-65529326 on Build GRCh38
rs755293066 Chr.X: 65529579-65529579 on Build GRCh38
rs749652693 Chr.X: 65529186-65529186 on Build GRCh38
rs752454328 Chr.X: 65529261-65529261 on Build GRCh38
rs372246177 Chr.X: 65529580-65529580 on Build GRCh38
rs770423824 Chr.X: 65529213-65529213 on Build GRCh38
rs56399692 Chr.X: 65529189-65529189 on Build GRCh38
rs770520405 Chr.X: 65529327-65529327 on Build GRCh38
rs1047584164 Chr.X: 65529385-65529385 on Build GRCh38
rs770440934 Chr.X: 65529433-65529433 on Build GRCh38
rs900177161 Chr.X: 65529525-65529525 on Build GRCh38
rs763441412 Chr.X: 65529475-65529475 on Build GRCh38
rs895874623 Chr.X: 65529317-65529317 on Build GRCh38
rs780245971 Chr.X: 65529569-65529569 on Build GRCh38
rs202161094 Chr.X: 65529156-65529156 on Build GRCh38
rs781241539 Chr.X: 65529559-65529559 on Build GRCh38
rs954515049 Chr.X: 65529595-65529595 on Build GRCh38
rs774684819 Chr.X: 65529543-65529543 on Build GRCh38

View Full Product Details