Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 149933810-149934307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf40B
Entrez Gene ID
541578
Gene Name
chromosome X open reading frame 40B
Gene Aliases
-
UniGene
Hs.545950
Gene Chromosome Location
Chr.X: 149924152-149938850 on Build GRCh38
Transcripts
NM_001013845.1 XM_005274698.1 XM_005274699.1 XM_005274700.1 XM_005274701.2 XM_005274702.3 XM_006724826.3 XM_011531180.2 XM_011531181.2 XM_017029585.1 XM_017029586.1 XM_017029587.1 XM_017029588.1 XM_017029589.1 XM_017029590.1 XM_017029591.1 XM_017029592.1 XM_017029593.1

SNP Detail

SNP ID Chromosome Location
rs35673661 Chr.X: 149933940-149933940 on Build GRCh38
rs371358984 Chr.X: 149933884-149933884 on Build GRCh38
rs200141791 Chr.X: 149933878-149933878 on Build GRCh38
rs184534288 Chr.X: 149934220-149934220 on Build GRCh38
rs781787334 Chr.X: 149933850-149933850 on Build GRCh38
rs782645159 Chr.X: 149933879-149933879 on Build GRCh38
rs782198624 Chr.X: 149933898-149933898 on Build GRCh38
rs782781489 Chr.X: 149934066-149934066 on Build GRCh38
rs567601 Chr.X: 149934162-149934162 on Build GRCh38
rs782398204 Chr.X: 149934215-149934216 on Build GRCh38
rs185155648 Chr.X: 149934042-149934042 on Build GRCh38
rs782543576 Chr.X: 149934078-149934078 on Build GRCh38
rs781978390 Chr.X: 149933899-149933899 on Build GRCh38
rs201330062 Chr.X: 149933860-149933860 on Build GRCh38
rs782725214 Chr.X: 149933841-149933841 on Build GRCh38
rs782813556 Chr.X: 149933915-149933915 on Build GRCh38
rs376109506 Chr.X: 149933917-149933917 on Build GRCh38
rs544843 Chr.X: 149934041-149934041 on Build GRCh38
rs781960033 Chr.X: 149933922-149933922 on Build GRCh38
rs375672264 Chr.X: 149934214-149934214 on Build GRCh38
rs782184964 Chr.X: 149934147-149934147 on Build GRCh38
rs141401249 Chr.X: 149934040-149934040 on Build GRCh38
rs782674667 Chr.X: 149933854-149933854 on Build GRCh38
rs782016278 Chr.X: 149934243-149934243 on Build GRCh38
rs544768 Chr.X: 149934016-149934016 on Build GRCh38
rs782583014 Chr.X: 149933850-149933850 on Build GRCh38
rs181122264 Chr.X: 149934216-149934216 on Build GRCh38
rs34347368 Chr.X: 149934072-149934072 on Build GRCh38
rs782043041 Chr.X: 149933967-149933968 on Build GRCh38
rs782133219 Chr.X: 149933900-149933900 on Build GRCh38
rs782565719 Chr.X: 149933885-149933885 on Build GRCh38
rs782684359 Chr.X: 149934129-149934129 on Build GRCh38
rs782394091 Chr.X: 149933921-149933921 on Build GRCh38
rs782444250 Chr.X: 149933852-149933852 on Build GRCh38
rs189855652 Chr.X: 149934044-149934044 on Build GRCh38
rs140366312 Chr.X: 149933937-149933937 on Build GRCh38
rs782210277 Chr.X: 149933880-149933880 on Build GRCh38
rs782381333 Chr.X: 149934223-149934223 on Build GRCh38

View Full Product Details