Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 152767328-152766824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGEA6
Entrez Gene ID
4105
Gene Name
MAGE family member A6
Gene Aliases
CT1.6, MAGE-3b, MAGE3B, MAGE6
UniGene
Hs.441113
Gene Chromosome Location
Chr.X: 152766136-152769729 on Build GRCh38
Transcripts
NM_005363.3 NM_175868.2

View Full Product Details