Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 230703201-230703698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGT
Entrez Gene ID
183
Gene Name
angiotensinogen
Gene Aliases
ANHU, SERPINA8
UniGene
Hs.19383
Gene Chromosome Location
Chr.1: 230702523-230714590 on Build GRCh38
Transcripts
NM_000029.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
905371 c.1298A>C p.E433A Chr.1: 230703301-230703301 on Build GRCh38
7154712 c.1297G>A p.E433K Chr.1: 230703302-230703302 on Build GRCh38
905372 c.1277A>G p.N426S Chr.1: 230703322-230703322 on Build GRCh38
5380030 c.1329C>T p.T443T Chr.1: 230703270-230703270 on Build GRCh38
4611891 c.1290_1291insT p.E431fs*1 Chr.1: 230703308-230703309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745975450 Chr.1: 230703308-230703309 on Build GRCh38
rs150388186 Chr.1: 230703345-230703345 on Build GRCh38
rs895513639 Chr.1: 230703490-230703490 on Build GRCh38
rs142379057 Chr.1: 230703402-230703402 on Build GRCh38
rs564906092 Chr.1: 230703665-230703665 on Build GRCh38
rs770652568 Chr.1: 230703226-230703226 on Build GRCh38
rs1002333564 Chr.1: 230703530-230703530 on Build GRCh38
rs780763836 Chr.1: 230703308-230703308 on Build GRCh38
rs1000819860 Chr.1: 230703257-230703257 on Build GRCh38
rs550007631 Chr.1: 230703438-230703438 on Build GRCh38
rs61751078 Chr.1: 230703254-230703254 on Build GRCh38
rs1053227909 Chr.1: 230703264-230703264 on Build GRCh38
rs986079735 Chr.1: 230703348-230703348 on Build GRCh38
rs1015514373 Chr.1: 230703558-230703558 on Build GRCh38
rs950212775 Chr.1: 230703476-230703476 on Build GRCh38
rs775451108 Chr.1: 230703376-230703376 on Build GRCh38
rs1013998268 Chr.1: 230703525-230703525 on Build GRCh38
rs759472175 Chr.1: 230703250-230703250 on Build GRCh38
rs571188083 Chr.1: 230703520-230703520 on Build GRCh38
rs888093710 Chr.1: 230703551-230703551 on Build GRCh38
rs1029464971 Chr.1: 230703608-230703608 on Build GRCh38
rs61751071 Chr.1: 230703647-230703647 on Build GRCh38
rs954166403 Chr.1: 230703631-230703631 on Build GRCh38
rs191959532 Chr.1: 230703403-230703403 on Build GRCh38
rs553581380 Chr.1: 230703598-230703598 on Build GRCh38
rs745906986 Chr.1: 230703325-230703325 on Build GRCh38
rs556000622 Chr.1: 230703578-230703578 on Build GRCh38
rs778583535 Chr.1: 230703352-230703352 on Build GRCh38
rs555751882 Chr.1: 230703431-230703431 on Build GRCh38
rs61751077 Chr.1: 230703274-230703274 on Build GRCh38
rs904454375 Chr.1: 230703439-230703439 on Build GRCh38
rs137880480 Chr.1: 230703351-230703351 on Build GRCh38
rs1046169858 Chr.1: 230703502-230703502 on Build GRCh38
rs911888987 Chr.1: 230703672-230703672 on Build GRCh38
rs375261929 Chr.1: 230703227-230703227 on Build GRCh38
rs986385581 Chr.1: 230703654-230703654 on Build GRCh38
rs1052982705 Chr.1: 230703403-230703403 on Build GRCh38
rs906290203 Chr.1: 230703529-230703529 on Build GRCh38
rs387906578 Chr.1: 230703309-230703309 on Build GRCh38
rs200657291 Chr.1: 230703276-230703276 on Build GRCh38
rs772226813 Chr.1: 230703345-230703347 on Build GRCh38
rs925998622 Chr.1: 230703652-230703652 on Build GRCh38
rs61751072 Chr.1: 230703544-230703544 on Build GRCh38
rs1005502733 Chr.1: 230703553-230703553 on Build GRCh38
rs187483077 Chr.1: 230703563-230703563 on Build GRCh38
rs777286397 Chr.1: 230703329-230703329 on Build GRCh38
rs771845691 Chr.1: 230703366-230703366 on Build GRCh38
rs935985238 Chr.1: 230703659-230703659 on Build GRCh38
rs755733687 Chr.1: 230703324-230703324 on Build GRCh38
rs149041409 Chr.1: 230703443-230703443 on Build GRCh38
rs749695432 Chr.1: 230703597-230703597 on Build GRCh38
rs748948502 Chr.1: 230703225-230703225 on Build GRCh38
rs989055893 Chr.1: 230703667-230703667 on Build GRCh38
rs61751076 Chr.1: 230703298-230703298 on Build GRCh38
rs113452955 Chr.1: 230703332-230703332 on Build GRCh38
rs756687983 Chr.1: 230703228-230703229 on Build GRCh38
rs61751075 Chr.1: 230703405-230703405 on Build GRCh38
rs573547402 Chr.1: 230703562-230703562 on Build GRCh38
rs757454311 Chr.1: 230703297-230703297 on Build GRCh38
rs1001879661 Chr.1: 230703321-230703321 on Build GRCh38
rs545648872 Chr.1: 230703398-230703398 on Build GRCh38
rs752080391 Chr.1: 230703311-230703311 on Build GRCh38
rs144932571 Chr.1: 230703364-230703364 on Build GRCh38
rs760807040 Chr.1: 230703258-230703258 on Build GRCh38
rs575963635 Chr.1: 230703594-230703594 on Build GRCh38
rs374998292 Chr.1: 230703300-230703300 on Build GRCh38
rs574211807 Chr.1: 230703470-230703470 on Build GRCh38
rs976870144 Chr.1: 230703378-230703378 on Build GRCh38
rs184180420 Chr.1: 230703527-230703527 on Build GRCh38
rs61751073 Chr.1: 230703542-230703542 on Build GRCh38
rs61751074 Chr.1: 230703447-230703447 on Build GRCh38
rs764132874 Chr.1: 230703259-230703259 on Build GRCh38
rs149063395 Chr.1: 230703365-230703365 on Build GRCh38
rs567898140 Chr.1: 230703407-230703407 on Build GRCh38

View Full Product Details