Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 233671171-233671712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK1
Entrez Gene ID
3775
Gene Name
potassium two pore domain channel subfamily K member 1
Gene Aliases
DPK, HOHO, K2P1, K2p1.1, KCNO1, TWIK-1, TWIK1
UniGene
Hs.208544
Gene Chromosome Location
Chr.1: 233614004-233672512 on Build GRCh38
Transcripts
NM_002245.3 XM_011544184.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3485439 c.480C>T p.F160F Chr.1: 233671347-233671347 on Build GRCh38
3864836 c.683G>A p.G228D Chr.1: 233671448-233671448 on Build GRCh38
4896514 c.516G>A p.E172E Chr.1: 233671383-233671383 on Build GRCh38
5131254 c.614A>G p.D205G Chr.1: 233671379-233671379 on Build GRCh38
6380395 c.565A>G p.T189A Chr.1: 233671432-233671432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888787148 Chr.1: 233671438-233671438 on Build GRCh38
rs549205062 Chr.1: 233671472-233671472 on Build GRCh38
rs753100891 Chr.1: 233671367-233671367 on Build GRCh38
rs913810427 Chr.1: 233671649-233671649 on Build GRCh38
rs149408317 Chr.1: 233671427-233671427 on Build GRCh38
rs866643709 Chr.1: 233671359-233671359 on Build GRCh38
rs1003588609 Chr.1: 233671616-233671616 on Build GRCh38
rs772618831 Chr.1: 233671433-233671433 on Build GRCh38
rs147294916 Chr.1: 233671381-233671381 on Build GRCh38
rs115321408 Chr.1: 233671331-233671331 on Build GRCh38
rs371437028 Chr.1: 233671287-233671287 on Build GRCh38
rs747494516 Chr.1: 233671531-233671531 on Build GRCh38
rs767922740 Chr.1: 233671361-233671361 on Build GRCh38
rs781062486 Chr.1: 233671399-233671399 on Build GRCh38
rs760129381 Chr.1: 233671356-233671356 on Build GRCh38
rs778495840 Chr.1: 233671254-233671254 on Build GRCh38
rs776986042 Chr.1: 233671533-233671533 on Build GRCh38
rs761068973 Chr.1: 233671450-233671450 on Build GRCh38
rs936352534 Chr.1: 233671593-233671593 on Build GRCh38
rs775442826 Chr.1: 233671650-233671650 on Build GRCh38
rs764919322 Chr.1: 233671371-233671371 on Build GRCh38
rs779937030 Chr.1: 233671275-233671275 on Build GRCh38
rs755030248 Chr.1: 233671469-233671470 on Build GRCh38
rs1050709 Chr.1: 233671690-233671690 on Build GRCh38
rs543539468 Chr.1: 233671574-233671574 on Build GRCh38
rs571173893 Chr.1: 233671678-233671678 on Build GRCh38
rs771162548 Chr.1: 233671352-233671352 on Build GRCh38
rs1026997545 Chr.1: 233671503-233671503 on Build GRCh38
rs749450155 Chr.1: 233671498-233671498 on Build GRCh38
rs749934083 Chr.1: 233671264-233671264 on Build GRCh38
rs761002286 Chr.1: 233671368-233671368 on Build GRCh38
rs754831522 Chr.1: 233671506-233671506 on Build GRCh38
rs949127217 Chr.1: 233671311-233671311 on Build GRCh38
rs751593176 Chr.1: 233671419-233671421 on Build GRCh38
rs1033766436 Chr.1: 233671638-233671638 on Build GRCh38
rs1009466525 Chr.1: 233671509-233671509 on Build GRCh38
rs759018571 Chr.1: 233671552-233671552 on Build GRCh38
rs754918285 Chr.1: 233671396-233671396 on Build GRCh38
rs544929189 Chr.1: 233671299-233671299 on Build GRCh38
rs746982221 Chr.1: 233671282-233671282 on Build GRCh38
rs769008300 Chr.1: 233671532-233671532 on Build GRCh38
rs751441096 Chr.1: 233671390-233671390 on Build GRCh38
rs200336822 Chr.1: 233671467-233671467 on Build GRCh38
rs984115041 Chr.1: 233671663-233671663 on Build GRCh38
rs199890728 Chr.1: 233671268-233671268 on Build GRCh38
rs1053422085 Chr.1: 233671620-233671620 on Build GRCh38
rs756005662 Chr.1: 233671566-233671566 on Build GRCh38
rs369234438 Chr.1: 233671496-233671496 on Build GRCh38
rs17851602 Chr.1: 233671419-233671419 on Build GRCh38
rs147325030 Chr.1: 233671428-233671428 on Build GRCh38
rs766007358 Chr.1: 233671537-233671537 on Build GRCh38
rs375598218 Chr.1: 233671547-233671547 on Build GRCh38
rs34957762 Chr.1: 233671322-233671322 on Build GRCh38
rs773204035 Chr.1: 233671400-233671400 on Build GRCh38
rs375764835 Chr.1: 233671213-233671213 on Build GRCh38
rs906334169 Chr.1: 233671578-233671578 on Build GRCh38
rs775908211 Chr.1: 233671437-233671437 on Build GRCh38
rs568968629 Chr.1: 233671502-233671502 on Build GRCh38
rs752620569 Chr.1: 233671561-233671561 on Build GRCh38
rs1032220898 Chr.1: 233671250-233671250 on Build GRCh38
rs777440124 Chr.1: 233671497-233671497 on Build GRCh38
rs763068423 Chr.1: 233671347-233671347 on Build GRCh38
rs765810464 Chr.1: 233671477-233671477 on Build GRCh38
rs890279828 Chr.1: 233671666-233671666 on Build GRCh38
rs750085516 Chr.1: 233671376-233671376 on Build GRCh38
rs992984063 Chr.1: 233671294-233671294 on Build GRCh38
rs980477151 Chr.1: 233671559-233671559 on Build GRCh38
rs757866197 Chr.1: 233671377-233671377 on Build GRCh38
rs765391065 Chr.1: 233671553-233671553 on Build GRCh38
rs917668918 Chr.1: 233671295-233671295 on Build GRCh38
rs764398476 Chr.1: 233671457-233671457 on Build GRCh38
rs778013725 Chr.1: 233671408-233671408 on Build GRCh38
rs542439358 Chr.1: 233671204-233671204 on Build GRCh38
rs890573305 Chr.1: 233671449-233671449 on Build GRCh38
rs1016918474 Chr.1: 233671675-233671675 on Build GRCh38
rs371852106 Chr.1: 233671507-233671507 on Build GRCh38
rs747965353 Chr.1: 233671404-233671404 on Build GRCh38
rs749541795 Chr.1: 233671409-233671409 on Build GRCh38
rs201127875 Chr.1: 233671431-233671431 on Build GRCh38
rs770054172 Chr.1: 233671330-233671330 on Build GRCh38
rs973637160 Chr.1: 233671193-233671193 on Build GRCh38
rs752620177 Chr.1: 233671490-233671490 on Build GRCh38
rs146074537 Chr.1: 233671481-233671481 on Build GRCh38
rs551285036 Chr.1: 233671608-233671608 on Build GRCh38
rs375357768 Chr.1: 233671479-233671479 on Build GRCh38
rs776024027 Chr.1: 233671511-233671511 on Build GRCh38
rs183890049 Chr.1: 233671510-233671510 on Build GRCh38
rs774947416 Chr.1: 233671353-233671353 on Build GRCh38
rs41272731 Chr.1: 233671405-233671405 on Build GRCh38
rs888754496 Chr.1: 233671212-233671212 on Build GRCh38

View Full Product Details