Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 104641003-104641510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SORCS3
Entrez Gene ID
22986
Gene Name
sortilin related VPS10 domain containing receptor 3
Gene Aliases
SORCS
UniGene
Hs.671950
Gene Chromosome Location
Chr.10: 104641101-105265242 on Build GRCh38
Transcripts
NM_014978.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7329957 c.183G>A p.P61P Chr.10: 104641510-104641510 on Build GRCh38
6053779 c.154C>T p.R52W Chr.10: 104641481-104641481 on Build GRCh38
4798264 c.55C>T p.R19W Chr.10: 104641382-104641382 on Build GRCh38
6419772 c.92C>T p.A31V Chr.10: 104641419-104641419 on Build GRCh38
6295473 c.18G>T p.T6T Chr.10: 104641345-104641345 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027963729 Chr.10: 104641470-104641470 on Build GRCh38
rs898209268 Chr.10: 104641451-104641451 on Build GRCh38
rs112044346 Chr.10: 104641450-104641450 on Build GRCh38
rs1028412752 Chr.10: 104641226-104641226 on Build GRCh38
rs559714338 Chr.10: 104641214-104641214 on Build GRCh38
rs974141784 Chr.10: 104641473-104641473 on Build GRCh38
rs764984191 Chr.10: 104641387-104641387 on Build GRCh38
rs910479980 Chr.10: 104641461-104641461 on Build GRCh38
rs919915291 Chr.10: 104641488-104641488 on Build GRCh38
rs758640408 Chr.10: 104641404-104641404 on Build GRCh38
rs962815435 Chr.10: 104641204-104641204 on Build GRCh38
rs574180991 Chr.10: 104641138-104641138 on Build GRCh38
rs1043849286 Chr.10: 104641049-104641049 on Build GRCh38
rs374520030 Chr.10: 104641184-104641184 on Build GRCh38
rs886628417 Chr.10: 104641175-104641175 on Build GRCh38
rs987880126 Chr.10: 104641130-104641130 on Build GRCh38
rs536070569 Chr.10: 104641033-104641033 on Build GRCh38
rs759388666 Chr.10: 104641379-104641379 on Build GRCh38
rs531369089 Chr.10: 104641408-104641408 on Build GRCh38
rs143668607 Chr.10: 104641421-104641421 on Build GRCh38
rs1032383267 Chr.10: 104641083-104641083 on Build GRCh38
rs757347409 Chr.10: 104641464-104641464 on Build GRCh38
rs1006374248 Chr.10: 104641176-104641176 on Build GRCh38
rs905414431 Chr.10: 104641028-104641028 on Build GRCh38
rs576782813 Chr.10: 104641236-104641236 on Build GRCh38
rs955276176 Chr.10: 104641228-104641231 on Build GRCh38
rs896727700 Chr.10: 104641160-104641160 on Build GRCh38
rs975096703 Chr.10: 104641219-104641219 on Build GRCh38
rs985935100 Chr.10: 104641168-104641168 on Build GRCh38
rs7923445 Chr.10: 104641244-104641244 on Build GRCh38
rs945181687 Chr.10: 104641247-104641247 on Build GRCh38
rs773411676 Chr.10: 104641370-104641370 on Build GRCh38
rs1016420269 Chr.10: 104641444-104641444 on Build GRCh38
rs954507458 Chr.10: 104641156-104641156 on Build GRCh38
rs956519886 Chr.10: 104641124-104641124 on Build GRCh38
rs938009990 Chr.10: 104641136-104641136 on Build GRCh38
rs562822100 Chr.10: 104641362-104641362 on Build GRCh38
rs903969861 Chr.10: 104641133-104641134 on Build GRCh38
rs896481097 Chr.10: 104641292-104641292 on Build GRCh38
rs1026401945 Chr.10: 104641162-104641162 on Build GRCh38
rs752962718 Chr.10: 104641401-104641401 on Build GRCh38
rs979285333 Chr.10: 104641253-104641253 on Build GRCh38
rs1000543452 Chr.10: 104641055-104641055 on Build GRCh38
rs925357381 Chr.10: 104641256-104641256 on Build GRCh38
rs554210791 Chr.10: 104641127-104641127 on Build GRCh38
rs1055100919 Chr.10: 104641280-104641280 on Build GRCh38
rs938129344 Chr.10: 104641257-104641257 on Build GRCh38

View Full Product Details