Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 8413414-8413920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STK33
Entrez Gene ID
65975
Gene Name
serine/threonine kinase 33
Gene Aliases
-
UniGene
Hs.501833
Gene Chromosome Location
Chr.11: 8335029-8594289 on Build GRCh38
Transcripts
NM_001289058.1 NM_001289059.1 NM_001289061.1 NM_030906.3 XM_011520288.2 XM_011520290.2 XM_011520292.2 XM_011520293.2 XM_011520294.1 XM_011520295.2 XM_011520297.2 XM_017018143.1 XM_017018144.1 XM_017018145.1 XM_017018146.1 XM_017018147.1 XM_017018148.1 XM_017018149.1 XM_017018150.1 XM_017018151.1 XM_017018152.1 XM_017018153.1 XM_017018154.1 XM_017018155.1 XM_017018156.1 XM_017018157.1 XM_017018158.1 XM_017018159.1 XM_017018160.1 XM_017018165.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6302033 c.1336G>T p.E446* Chr.11: 8413503-8413503 on Build GRCh38
1357467 c.1164G>A p.S388S Chr.11: 8413675-8413675 on Build GRCh38
6134833 c.1300G>A p.V434I Chr.11: 8413539-8413539 on Build GRCh38
3870338 c.1213C>T p.P405S Chr.11: 8413626-8413626 on Build GRCh38
40660 c.1203G>A p.W401* Chr.11: 8413636-8413636 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776746933 Chr.11: 8413851-8413851 on Build GRCh38
rs372369200 Chr.11: 8413463-8413463 on Build GRCh38
rs774490392 Chr.11: 8413628-8413628 on Build GRCh38
rs747954934 Chr.11: 8413691-8413691 on Build GRCh38
rs556481805 Chr.11: 8413895-8413895 on Build GRCh38
rs750020482 Chr.11: 8413669-8413669 on Build GRCh38
rs201296442 Chr.11: 8413602-8413602 on Build GRCh38
rs148957456 Chr.11: 8413473-8413473 on Build GRCh38
rs1014646604 Chr.11: 8413894-8413894 on Build GRCh38
rs879037414 Chr.11: 8413530-8413531 on Build GRCh38
rs771212761 Chr.11: 8413743-8413743 on Build GRCh38
rs145552076 Chr.11: 8413583-8413583 on Build GRCh38
rs192164307 Chr.11: 8413856-8413856 on Build GRCh38
rs755361423 Chr.11: 8413455-8413455 on Build GRCh38
rs386750506 Chr.11: 8413530-8413531 on Build GRCh38
rs1053139796 Chr.11: 8413846-8413846 on Build GRCh38
rs531517965 Chr.11: 8413745-8413745 on Build GRCh38
rs763109430 Chr.11: 8413610-8413610 on Build GRCh38
rs530194211 Chr.11: 8413639-8413639 on Build GRCh38
rs943862675 Chr.11: 8413615-8413615 on Build GRCh38
rs570768192 Chr.11: 8413771-8413771 on Build GRCh38
rs3751096 Chr.11: 8413531-8413531 on Build GRCh38
rs768279440 Chr.11: 8413503-8413503 on Build GRCh38
rs755520485 Chr.11: 8413538-8413538 on Build GRCh38
rs186878393 Chr.11: 8413854-8413854 on Build GRCh38
rs564944697 Chr.11: 8413545-8413545 on Build GRCh38
rs774242804 Chr.11: 8413744-8413744 on Build GRCh38
rs769773362 Chr.11: 8413705-8413705 on Build GRCh38
rs143683529 Chr.11: 8413546-8413546 on Build GRCh38
rs752060364 Chr.11: 8413537-8413537 on Build GRCh38
rs758267793 Chr.11: 8413624-8413625 on Build GRCh38
rs771722375 Chr.11: 8413515-8413515 on Build GRCh38
rs753038178 Chr.11: 8413654-8413654 on Build GRCh38
rs754853163 Chr.11: 8413681-8413681 on Build GRCh38
rs376836438 Chr.11: 8413597-8413597 on Build GRCh38
rs758076205 Chr.11: 8413674-8413674 on Build GRCh38
rs779632517 Chr.11: 8413582-8413582 on Build GRCh38
rs745404119 Chr.11: 8413476-8413476 on Build GRCh38
rs752884359 Chr.11: 8413490-8413493 on Build GRCh38
rs748638696 Chr.11: 8413449-8413449 on Build GRCh38
rs569005061 Chr.11: 8413735-8413735 on Build GRCh38
rs773408144 Chr.11: 8413529-8413529 on Build GRCh38
rs746808447 Chr.11: 8413493-8413493 on Build GRCh38
rs779391532 Chr.11: 8413675-8413675 on Build GRCh38
rs371820251 Chr.11: 8413566-8413566 on Build GRCh38
rs764864603 Chr.11: 8413662-8413662 on Build GRCh38
rs1031797851 Chr.11: 8413448-8413448 on Build GRCh38
rs548920572 Chr.11: 8413676-8413676 on Build GRCh38
rs774474836 Chr.11: 8413677-8413680 on Build GRCh38
rs777721140 Chr.11: 8413727-8413727 on Build GRCh38
rs571563400 Chr.11: 8413772-8413772 on Build GRCh38
rs781323502 Chr.11: 8413687-8413687 on Build GRCh38
rs776489713 Chr.11: 8413516-8413516 on Build GRCh38
rs759917299 Chr.11: 8413638-8413638 on Build GRCh38
rs148092034 Chr.11: 8413764-8413764 on Build GRCh38
rs374456505 Chr.11: 8413862-8413862 on Build GRCh38
rs533801755 Chr.11: 8413803-8413803 on Build GRCh38
rs367730444 Chr.11: 8413526-8413526 on Build GRCh38
rs770572042 Chr.11: 8413465-8413465 on Build GRCh38
rs766309071 Chr.11: 8413612-8413612 on Build GRCh38
rs996775714 Chr.11: 8413784-8413784 on Build GRCh38
rs3751095 Chr.11: 8413530-8413530 on Build GRCh38
rs934737420 Chr.11: 8413820-8413820 on Build GRCh38
rs778585247 Chr.11: 8413549-8413549 on Build GRCh38
rs759840447 Chr.11: 8413885-8413885 on Build GRCh38
rs921202669 Chr.11: 8413661-8413661 on Build GRCh38
rs906102951 Chr.11: 8413891-8413891 on Build GRCh38
rs377485276 Chr.11: 8413544-8413544 on Build GRCh38
rs927203905 Chr.11: 8413620-8413620 on Build GRCh38
rs751924380 Chr.11: 8413447-8413447 on Build GRCh38
rs775327765 Chr.11: 8413492-8413492 on Build GRCh38
rs11041911 Chr.11: 8413811-8413811 on Build GRCh38
rs149155880 Chr.11: 8413522-8413522 on Build GRCh38
rs771972595 Chr.11: 8413481-8413481 on Build GRCh38
rs1054461825 Chr.11: 8413766-8413766 on Build GRCh38
rs776243942 Chr.11: 8413588-8413588 on Build GRCh38
rs900733782 Chr.11: 8413767-8413767 on Build GRCh38
rs140349792 Chr.11: 8413553-8413553 on Build GRCh38
rs537192998 Chr.11: 8413490-8413490 on Build GRCh38
rs756509438 Chr.11: 8413657-8413657 on Build GRCh38
rs35974908 Chr.11: 8413857-8413858 on Build GRCh38
rs542889891 Chr.11: 8413462-8413462 on Build GRCh38

View Full Product Details