Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 9575702-9576199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WEE1
Entrez Gene ID
7465
Gene Name
WEE1 G2 checkpoint kinase
Gene Aliases
WEE1A, WEE1hu
UniGene
Hs.249441
Gene Chromosome Location
Chr.11: 9573681-9589767 on Build GRCh38
Transcripts
NM_001143976.1 NM_003390.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1628309 c.661G>A p.G221R Chr.11: 9575972-9575972 on Build GRCh38
40767 c.765G>A p.K255K Chr.11: 9576076-9576076 on Build GRCh38
6822475 c.711delT p.F238fs*54 Chr.11: 9576022-9576022 on Build GRCh38
1298788 c.577-1G>A p.? Chr.11: 9575887-9575887 on Build GRCh38
933647 c.732G>T p.L244F Chr.11: 9576043-9576043 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780943638 Chr.11: 9575944-9575944 on Build GRCh38
rs745790769 Chr.11: 9576125-9576125 on Build GRCh38
rs905894715 Chr.11: 9575775-9575775 on Build GRCh38
rs930733459 Chr.11: 9576052-9576052 on Build GRCh38
rs750760498 Chr.11: 9575813-9575813 on Build GRCh38
rs781682950 Chr.11: 9575750-9575750 on Build GRCh38
rs778857531 Chr.11: 9576031-9576031 on Build GRCh38
rs772824743 Chr.11: 9575966-9575966 on Build GRCh38
rs747692475 Chr.11: 9575954-9575954 on Build GRCh38
rs985733771 Chr.11: 9575906-9575906 on Build GRCh38
rs968850759 Chr.11: 9575836-9575836 on Build GRCh38
rs921003033 Chr.11: 9575789-9575789 on Build GRCh38
rs547746842 Chr.11: 9576044-9576044 on Build GRCh38
rs769467349 Chr.11: 9575960-9575960 on Build GRCh38
rs1044266068 Chr.11: 9575732-9575732 on Build GRCh38
rs771907182 Chr.11: 9576136-9576136 on Build GRCh38
rs150127351 Chr.11: 9576084-9576084 on Build GRCh38
rs570132856 Chr.11: 9575986-9575986 on Build GRCh38
rs370082104 Chr.11: 9576006-9576006 on Build GRCh38
rs749503240 Chr.11: 9575981-9575981 on Build GRCh38
rs776587965 Chr.11: 9576160-9576160 on Build GRCh38
rs756507839 Chr.11: 9575918-9575918 on Build GRCh38
rs929081168 Chr.11: 9576010-9576010 on Build GRCh38
rs774197082 Chr.11: 9575987-9575987 on Build GRCh38
rs757695369 Chr.11: 9575902-9575902 on Build GRCh38
rs752784574 Chr.11: 9575856-9575856 on Build GRCh38
rs577896829 Chr.11: 9576116-9576116 on Build GRCh38
rs753859170 Chr.11: 9576032-9576032 on Build GRCh38
rs779402741 Chr.11: 9575907-9575907 on Build GRCh38
rs759867127 Chr.11: 9575842-9575842 on Build GRCh38
rs934876910 Chr.11: 9575869-9575869 on Build GRCh38
rs1057134115 Chr.11: 9576155-9576155 on Build GRCh38
rs750823367 Chr.11: 9576066-9576066 on Build GRCh38
rs891874860 Chr.11: 9575808-9575808 on Build GRCh38
rs752960652 Chr.11: 9575997-9575997 on Build GRCh38
rs1021801660 Chr.11: 9575824-9575824 on Build GRCh38
rs34412975 Chr.11: 9575939-9575939 on Build GRCh38
rs767655770 Chr.11: 9575849-9575849 on Build GRCh38
rs189741995 Chr.11: 9576105-9576105 on Build GRCh38
rs759345408 Chr.11: 9576171-9576171 on Build GRCh38
rs375227451 Chr.11: 9575896-9575896 on Build GRCh38
rs756149856 Chr.11: 9575865-9575865 on Build GRCh38
rs1049015057 Chr.11: 9576075-9576075 on Build GRCh38
rs768899313 Chr.11: 9575994-9575994 on Build GRCh38
rs1036216855 Chr.11: 9575798-9575798 on Build GRCh38
rs199979048 Chr.11: 9576113-9576113 on Build GRCh38
rs751294907 Chr.11: 9575841-9575841 on Build GRCh38
rs780534747 Chr.11: 9576091-9576091 on Build GRCh38
rs567856051 Chr.11: 9575779-9575779 on Build GRCh38
rs757788231 Chr.11: 9576034-9576034 on Build GRCh38
rs759191432 Chr.11: 9575991-9575991 on Build GRCh38
rs770775251 Chr.11: 9576126-9576126 on Build GRCh38
rs536796841 Chr.11: 9575799-9575799 on Build GRCh38
rs976583476 Chr.11: 9575785-9575785 on Build GRCh38
rs375163810 Chr.11: 9576162-9576162 on Build GRCh38
rs548599529 Chr.11: 9575886-9575886 on Build GRCh38
rs144528960 Chr.11: 9576096-9576096 on Build GRCh38

View Full Product Details