Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 14358601-14359136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRAS2
Entrez Gene ID
22800
Gene Name
related RAS viral (r-ras) oncogene homolog 2
Gene Aliases
TC21
UniGene
Hs.502004
Gene Chromosome Location
Chr.11: 14277920-14364506 on Build GRCh38
Transcripts
NM_001177314.1 NM_012250.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6989585 c.68G>A p.G23D Chr.11: 14358803-14358803 on Build GRCh38
4722825 c.22G>A p.D8N Chr.11: 14358849-14358849 on Build GRCh38
376569 c.87G>T p.A29A Chr.11: 14358784-14358784 on Build GRCh38
4722824 c.71G>T p.G24V Chr.11: 14358800-14358800 on Build GRCh38
5749961 c.67G>T p.G23C Chr.11: 14358804-14358804 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545759157 Chr.11: 14359031-14359031 on Build GRCh38
rs943213310 Chr.11: 14359114-14359114 on Build GRCh38
rs543550319 Chr.11: 14359082-14359082 on Build GRCh38
rs782016031 Chr.11: 14358745-14358745 on Build GRCh38
rs997207680 Chr.11: 14358988-14358988 on Build GRCh38
rs539805663 Chr.11: 14358733-14358733 on Build GRCh38
rs537165957 Chr.11: 14358880-14358880 on Build GRCh38
rs782785201 Chr.11: 14358859-14358859 on Build GRCh38
rs572281354 Chr.11: 14358995-14358995 on Build GRCh38
rs375964598 Chr.11: 14358758-14358758 on Build GRCh38
rs548067518 Chr.11: 14358686-14358686 on Build GRCh38
rs971434154 Chr.11: 14359083-14359083 on Build GRCh38
rs566286071 Chr.11: 14358694-14358694 on Build GRCh38
rs568685229 Chr.11: 14358660-14358660 on Build GRCh38
rs968049193 Chr.11: 14359007-14359007 on Build GRCh38
rs782428756 Chr.11: 14358903-14358903 on Build GRCh38
rs113979773 Chr.11: 14358942-14358942 on Build GRCh38
rs554023024 Chr.11: 14358994-14358994 on Build GRCh38
rs929221154 Chr.11: 14359109-14359109 on Build GRCh38
rs782572326 Chr.11: 14358740-14358740 on Build GRCh38
rs1038837653 Chr.11: 14358973-14358973 on Build GRCh38
rs782169288 Chr.11: 14358742-14358742 on Build GRCh38
rs981426188 Chr.11: 14359084-14359084 on Build GRCh38
rs901580580 Chr.11: 14358978-14358978 on Build GRCh38
rs536114965 Chr.11: 14358667-14358667 on Build GRCh38
rs782133096 Chr.11: 14358751-14358751 on Build GRCh38
rs782599910 Chr.11: 14358886-14358886 on Build GRCh38
rs984683980 Chr.11: 14359110-14359110 on Build GRCh38
rs541703414 Chr.11: 14358956-14358956 on Build GRCh38
rs568594255 Chr.11: 14359094-14359094 on Build GRCh38
rs574297460 Chr.11: 14358891-14358891 on Build GRCh38
rs782413007 Chr.11: 14358743-14358743 on Build GRCh38
rs782647300 Chr.11: 14358731-14358731 on Build GRCh38
rs782684347 Chr.11: 14358721-14358721 on Build GRCh38
rs531400386 Chr.11: 14359075-14359075 on Build GRCh38
rs782336781 Chr.11: 14358736-14358736 on Build GRCh38
rs781902160 Chr.11: 14358842-14358842 on Build GRCh38
rs782441022 Chr.11: 14358714-14358714 on Build GRCh38
rs555901595 Chr.11: 14358890-14358890 on Build GRCh38
rs781863536 Chr.11: 14358871-14358871 on Build GRCh38
rs782710903 Chr.11: 14358841-14358841 on Build GRCh38
rs529481468 Chr.11: 14359112-14359112 on Build GRCh38
rs782151181 Chr.11: 14358858-14358858 on Build GRCh38
rs782528964 Chr.11: 14358730-14358730 on Build GRCh38
rs189969382 Chr.11: 14359047-14359047 on Build GRCh38
rs999084423 Chr.11: 14359023-14359023 on Build GRCh38
rs184872101 Chr.11: 14358781-14358781 on Build GRCh38
rs782287512 Chr.11: 14358729-14358729 on Build GRCh38

View Full Product Details