Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 14771803-14772319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE3B
Entrez Gene ID
5140
Gene Name
phosphodiesterase 3B
Gene Aliases
HcGIP1, cGIPDE1
UniGene
Hs.445711
Gene Chromosome Location
Chr.11: 14643691-14872059 on Build GRCh38
Transcripts
NM_000922.3 XM_006718249.3 XM_017017911.1 XM_017017912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5346253 c.992A>T p.D331V Chr.11: 14771950-14771950 on Build GRCh38
6308363 c.1018C>A p.P340T Chr.11: 14771976-14771976 on Build GRCh38
136820 c.997G>A p.D333N Chr.11: 14771955-14771955 on Build GRCh38
6248918 c.1010G>T p.W337L Chr.11: 14771968-14771968 on Build GRCh38
687124 c.979-2A>G p.? Chr.11: 14771935-14771935 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747816432 Chr.11: 14772067-14772067 on Build GRCh38
rs989701942 Chr.11: 14772018-14772037 on Build GRCh38
rs757073652 Chr.11: 14772000-14772000 on Build GRCh38
rs778883084 Chr.11: 14772007-14772007 on Build GRCh38
rs767103670 Chr.11: 14772015-14772018 on Build GRCh38
rs763915698 Chr.11: 14771995-14771996 on Build GRCh38
rs894252316 Chr.11: 14771871-14771871 on Build GRCh38
rs931074773 Chr.11: 14772194-14772194 on Build GRCh38
rs997511407 Chr.11: 14772019-14772022 on Build GRCh38
rs776314699 Chr.11: 14772051-14772051 on Build GRCh38
rs1015586366 Chr.11: 14772217-14772217 on Build GRCh38
rs771303467 Chr.11: 14771889-14771889 on Build GRCh38
rs1051709642 Chr.11: 14771872-14771872 on Build GRCh38
rs772061383 Chr.11: 14772019-14772019 on Build GRCh38
rs773567292 Chr.11: 14772079-14772079 on Build GRCh38
rs776974195 Chr.11: 14771921-14771921 on Build GRCh38
rs932872061 Chr.11: 14771854-14771854 on Build GRCh38
rs564021289 Chr.11: 14772253-14772253 on Build GRCh38
rs915562874 Chr.11: 14772190-14772190 on Build GRCh38
rs912775098 Chr.11: 14771884-14771884 on Build GRCh38
rs751670797 Chr.11: 14771974-14771974 on Build GRCh38
rs954273460 Chr.11: 14772287-14772287 on Build GRCh38
rs765165070 Chr.11: 14771924-14771924 on Build GRCh38
rs753667014 Chr.11: 14771991-14771991 on Build GRCh38
rs960625847 Chr.11: 14772184-14772184 on Build GRCh38
rs201526030 Chr.11: 14771910-14771910 on Build GRCh38
rs766300624 Chr.11: 14771969-14771969 on Build GRCh38
rs779046647 Chr.11: 14771890-14771890 on Build GRCh38
rs934926747 Chr.11: 14771900-14771900 on Build GRCh38
rs750876378 Chr.11: 14771937-14771937 on Build GRCh38
rs149969552 Chr.11: 14772167-14772167 on Build GRCh38
rs1055469332 Chr.11: 14771858-14771858 on Build GRCh38
rs768160980 Chr.11: 14772047-14772047 on Build GRCh38
rs763479069 Chr.11: 14771957-14771957 on Build GRCh38
rs1028941832 Chr.11: 14772020-14772020 on Build GRCh38
rs779418712 Chr.11: 14772038-14772038 on Build GRCh38
rs770100130 Chr.11: 14772068-14772068 on Build GRCh38
rs576805767 Chr.11: 14771841-14771841 on Build GRCh38
rs751189316 Chr.11: 14771999-14772000 on Build GRCh38
rs771443561 Chr.11: 14771897-14771897 on Build GRCh38
rs759539367 Chr.11: 14771923-14771924 on Build GRCh38
rs749878635 Chr.11: 14772071-14772072 on Build GRCh38
rs372601024 Chr.11: 14771886-14771886 on Build GRCh38
rs762731788 Chr.11: 14771960-14771964 on Build GRCh38
rs1021893570 Chr.11: 14772099-14772099 on Build GRCh38
rs371392469 Chr.11: 14771915-14771915 on Build GRCh38
rs887951384 Chr.11: 14772202-14772204 on Build GRCh38
rs745734144 Chr.11: 14772008-14772008 on Build GRCh38
rs1049720273 Chr.11: 14771901-14771901 on Build GRCh38
rs746594250 Chr.11: 14772044-14772044 on Build GRCh38
rs974149519 Chr.11: 14772245-14772245 on Build GRCh38
rs892155255 Chr.11: 14771942-14771942 on Build GRCh38
rs755152977 Chr.11: 14771975-14771975 on Build GRCh38
rs574518616 Chr.11: 14771984-14771984 on Build GRCh38
rs902137792 Chr.11: 14772290-14772290 on Build GRCh38
rs967207176 Chr.11: 14772145-14772145 on Build GRCh38
rs756852203 Chr.11: 14772002-14772003 on Build GRCh38
rs928578147 Chr.11: 14772161-14772161 on Build GRCh38
rs745822090 Chr.11: 14771895-14771895 on Build GRCh38
rs763308698 Chr.11: 14772087-14772087 on Build GRCh38
rs1052023532 Chr.11: 14771928-14771928 on Build GRCh38
rs189128625 Chr.11: 14772160-14772160 on Build GRCh38
rs545809950 Chr.11: 14772221-14772221 on Build GRCh38
rs776734081 Chr.11: 14771889-14771890 on Build GRCh38
rs1010607302 Chr.11: 14772042-14772042 on Build GRCh38
rs371589178 Chr.11: 14771977-14771977 on Build GRCh38
rs921317905 Chr.11: 14772256-14772256 on Build GRCh38
rs765710633 Chr.11: 14771927-14771927 on Build GRCh38
rs904948534 Chr.11: 14772143-14772143 on Build GRCh38
rs1015095953 Chr.11: 14771987-14771987 on Build GRCh38
rs938292127 Chr.11: 14771848-14771853 on Build GRCh38
rs760189258 Chr.11: 14771911-14771911 on Build GRCh38
rs775071102 Chr.11: 14771899-14771899 on Build GRCh38
rs572408706 Chr.11: 14772196-14772196 on Build GRCh38
rs775347861 Chr.11: 14771932-14771933 on Build GRCh38
rs991184065 Chr.11: 14772185-14772185 on Build GRCh38
rs1048115359 Chr.11: 14772200-14772200 on Build GRCh38

View Full Product Details