Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 18405337-18405837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LDHA
Entrez Gene ID
3939
Gene Name
lactate dehydrogenase A
Gene Aliases
GSD11, HEL-S-133P, LDHM, PIG19
UniGene
Hs.2795
Gene Chromosome Location
Chr.11: 18394389-18408218 on Build GRCh38
Transcripts
NM_001135239.1 NM_001165414.1 NM_001165415.1 NM_001165416.1 NM_005566.3 NR_028500.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6850275 c.879G>A p.M293I Chr.11: 18405530-18405530 on Build GRCh38
4602202 c.821C>T p.S274F Chr.11: 18405559-18405559 on Build GRCh38
3368321 c.755T>C p.I252T Chr.11: 18405493-18405493 on Build GRCh38
6131869 c.762C>G p.L254L Chr.11: 18405500-18405500 on Build GRCh38
4697683 c.897G>A p.V299V Chr.11: 18405548-18405548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3781641 Chr.11: 18405770-18405770 on Build GRCh38
rs759838023 Chr.11: 18405549-18405549 on Build GRCh38
rs757158178 Chr.11: 18405683-18405683 on Build GRCh38
rs769677845 Chr.11: 18405413-18405413 on Build GRCh38
rs1041471134 Chr.11: 18405396-18405396 on Build GRCh38
rs773945378 Chr.11: 18405619-18405619 on Build GRCh38
rs752788996 Chr.11: 18405428-18405428 on Build GRCh38
rs201008031 Chr.11: 18405615-18405615 on Build GRCh38
rs541062699 Chr.11: 18405427-18405427 on Build GRCh38
rs200133108 Chr.11: 18405613-18405613 on Build GRCh38
rs756225293 Chr.11: 18405565-18405565 on Build GRCh38
rs749119901 Chr.11: 18405575-18405576 on Build GRCh38
rs1016707499 Chr.11: 18405808-18405808 on Build GRCh38
rs199758475 Chr.11: 18405612-18405612 on Build GRCh38
rs559447427 Chr.11: 18405543-18405543 on Build GRCh38
rs558820257 Chr.11: 18405625-18405625 on Build GRCh38
rs566653337 Chr.11: 18405398-18405398 on Build GRCh38
rs137996815 Chr.11: 18405515-18405515 on Build GRCh38
rs367553288 Chr.11: 18405563-18405563 on Build GRCh38
rs370174954 Chr.11: 18405593-18405593 on Build GRCh38
rs766720385 Chr.11: 18405541-18405541 on Build GRCh38
rs764211566 Chr.11: 18405578-18405578 on Build GRCh38
rs762657047 Chr.11: 18405537-18405537 on Build GRCh38
rs755745849 Chr.11: 18405491-18405491 on Build GRCh38
rs200483406 Chr.11: 18405441-18405441 on Build GRCh38
rs201227737 Chr.11: 18405470-18405470 on Build GRCh38
rs201140577 Chr.11: 18405592-18405592 on Build GRCh38
rs779893062 Chr.11: 18405498-18405499 on Build GRCh38
rs961097476 Chr.11: 18405384-18405387 on Build GRCh38
rs776649952 Chr.11: 18405489-18405489 on Build GRCh38
rs905706599 Chr.11: 18405788-18405789 on Build GRCh38
rs768277742 Chr.11: 18405590-18405591 on Build GRCh38
rs747680577 Chr.11: 18405503-18405503 on Build GRCh38
rs371400978 Chr.11: 18405546-18405546 on Build GRCh38
rs758175888 Chr.11: 18405455-18405455 on Build GRCh38
rs557288314 Chr.11: 18405647-18405647 on Build GRCh38
rs918037226 Chr.11: 18405711-18405711 on Build GRCh38
rs542849128 Chr.11: 18405807-18405807 on Build GRCh38
rs981181313 Chr.11: 18405763-18405763 on Build GRCh38
rs768715402 Chr.11: 18405485-18405485 on Build GRCh38
rs749493960 Chr.11: 18405450-18405450 on Build GRCh38
rs187005834 Chr.11: 18405548-18405548 on Build GRCh38
rs754064594 Chr.11: 18405583-18405583 on Build GRCh38
rs781321565 Chr.11: 18405607-18405607 on Build GRCh38
rs746810321 Chr.11: 18405596-18405596 on Build GRCh38
rs1008320372 Chr.11: 18405438-18405438 on Build GRCh38
rs754768150 Chr.11: 18405597-18405597 on Build GRCh38
rs1016980503 Chr.11: 18405366-18405366 on Build GRCh38
rs202202736 Chr.11: 18405430-18405430 on Build GRCh38
rs746981573 Chr.11: 18405480-18405480 on Build GRCh38
rs767946655 Chr.11: 18405414-18405414 on Build GRCh38
rs200971093 Chr.11: 18405403-18405403 on Build GRCh38
rs970690263 Chr.11: 18405722-18405722 on Build GRCh38
rs879022081 Chr.11: 18405446-18405446 on Build GRCh38
rs200256380 Chr.11: 18405562-18405562 on Build GRCh38
rs936908758 Chr.11: 18405798-18405798 on Build GRCh38
rs755472354 Chr.11: 18405407-18405407 on Build GRCh38
rs35510986 Chr.11: 18405401-18405402 on Build GRCh38
rs533525174 Chr.11: 18405736-18405736 on Build GRCh38
rs772791680 Chr.11: 18405530-18405530 on Build GRCh38
rs1053023691 Chr.11: 18405682-18405686 on Build GRCh38
rs1005698286 Chr.11: 18405797-18405797 on Build GRCh38

View Full Product Details