Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65078914-65079410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCA5
Entrez Gene ID
113130
Gene Name
cell division cycle associated 5
Gene Aliases
SORORIN
UniGene
Hs.434886
Gene Chromosome Location
Chr.11: 65061009-65084143 on Build GRCh38
Transcripts
NM_080668.3 XM_005273733.3 XM_005273734.3 XM_011544743.2 XM_011544747.2 XM_011544748.2 XM_011544749.2 XM_017017158.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5808717 c.699G>A p.W233* Chr.11: 65079167-65079167 on Build GRCh38
3935572 c.671A>T p.E224V Chr.11: 65079360-65079360 on Build GRCh38
1298400 c.754G>C p.E252Q Chr.11: 65079112-65079112 on Build GRCh38
415454 c.653A>T p.K218M Chr.11: 65079378-65079378 on Build GRCh38
4442337 c.719A>G p.E240G Chr.11: 65079147-65079147 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772151897 Chr.11: 65079157-65079157 on Build GRCh38
rs757274548 Chr.11: 65079342-65079342 on Build GRCh38
rs746934875 Chr.11: 65079341-65079341 on Build GRCh38
rs753160448 Chr.11: 65079202-65079202 on Build GRCh38
rs994679427 Chr.11: 65079231-65079231 on Build GRCh38
rs776451537 Chr.11: 65079093-65079093 on Build GRCh38
rs149256940 Chr.11: 65079377-65079377 on Build GRCh38
rs781373223 Chr.11: 65079245-65079245 on Build GRCh38
rs771200184 Chr.11: 65079381-65079381 on Build GRCh38
rs1055341605 Chr.11: 65079203-65079203 on Build GRCh38
rs750531620 Chr.11: 65079319-65079319 on Build GRCh38
rs766138716 Chr.11: 65079099-65079099 on Build GRCh38
rs770967585 Chr.11: 65079306-65079306 on Build GRCh38
rs766657504 Chr.11: 65079323-65079323 on Build GRCh38
rs764857125 Chr.11: 65079195-65079196 on Build GRCh38
rs759268942 Chr.11: 65079182-65079182 on Build GRCh38
rs111891963 Chr.11: 65079246-65079246 on Build GRCh38
rs970594445 Chr.11: 65079073-65079073 on Build GRCh38
rs745491639 Chr.11: 65079279-65079279 on Build GRCh38
rs530338793 Chr.11: 65079151-65079151 on Build GRCh38
rs1031650732 Chr.11: 65079371-65079371 on Build GRCh38
rs546928568 Chr.11: 65079280-65079281 on Build GRCh38
rs767483754 Chr.11: 65079312-65079312 on Build GRCh38
rs993297833 Chr.11: 65079044-65079044 on Build GRCh38
rs987171171 Chr.11: 65078976-65078976 on Build GRCh38
rs763391704 Chr.11: 65079213-65079213 on Build GRCh38
rs747909180 Chr.11: 65079050-65079050 on Build GRCh38
rs754071624 Chr.11: 65079381-65079381 on Build GRCh38
rs752495314 Chr.11: 65079334-65079334 on Build GRCh38
rs768589748 Chr.11: 65079067-65079067 on Build GRCh38
rs372545623 Chr.11: 65079148-65079148 on Build GRCh38
rs565801632 Chr.11: 65079278-65079278 on Build GRCh38
rs973311735 Chr.11: 65078944-65078944 on Build GRCh38
rs369116079 Chr.11: 65079122-65079122 on Build GRCh38
rs748979988 Chr.11: 65079382-65079382 on Build GRCh38
rs142767058 Chr.11: 65078945-65078945 on Build GRCh38
rs779766967 Chr.11: 65079058-65079058 on Build GRCh38
rs764535684 Chr.11: 65079301-65079301 on Build GRCh38
rs565024216 Chr.11: 65079021-65079021 on Build GRCh38
rs762375035 Chr.11: 65079307-65079307 on Build GRCh38
rs767880903 Chr.11: 65079325-65079326 on Build GRCh38
rs777878652 Chr.11: 65079274-65079274 on Build GRCh38
rs756227004 Chr.11: 65079322-65079322 on Build GRCh38
rs950392978 Chr.11: 65078966-65078966 on Build GRCh38
rs781300562 Chr.11: 65079149-65079149 on Build GRCh38
rs756418450 Chr.11: 65079250-65079250 on Build GRCh38
rs117649318 Chr.11: 65079097-65079097 on Build GRCh38
rs774134289 Chr.11: 65079080-65079080 on Build GRCh38
rs779222711 Chr.11: 65079041-65079041 on Build GRCh38
rs760297100 Chr.11: 65079168-65079168 on Build GRCh38
rs765678897 Chr.11: 65079112-65079112 on Build GRCh38
rs772783951 Chr.11: 65079162-65079162 on Build GRCh38
rs757306067 Chr.11: 65079230-65079230 on Build GRCh38
rs769499015 Chr.11: 65079291-65079291 on Build GRCh38
rs758249016 Chr.11: 65079335-65079335 on Build GRCh38
rs750850005 Chr.11: 65079334-65079335 on Build GRCh38
rs759202803 Chr.11: 65079192-65079192 on Build GRCh38
rs764587018 Chr.11: 65079219-65079219 on Build GRCh38
rs202177996 Chr.11: 65079101-65079101 on Build GRCh38
rs763338557 Chr.11: 65079298-65079298 on Build GRCh38
rs552327395 Chr.11: 65079272-65079272 on Build GRCh38
rs747963159 Chr.11: 65079271-65079271 on Build GRCh38
rs561208556 Chr.11: 65078983-65078983 on Build GRCh38
rs200941468 Chr.11: 65079156-65079156 on Build GRCh38
rs780398756 Chr.11: 65079369-65079371 on Build GRCh38
rs775971340 Chr.11: 65079345-65079345 on Build GRCh38
rs775648505 Chr.11: 65079297-65079297 on Build GRCh38
rs777815769 Chr.11: 65079123-65079123 on Build GRCh38
rs943530041 Chr.11: 65078996-65078996 on Build GRCh38
rs576626558 Chr.11: 65078963-65078963 on Build GRCh38
rs751598567 Chr.11: 65079226-65079226 on Build GRCh38
rs752243827 Chr.11: 65079204-65079206 on Build GRCh38
rs759837525 Chr.11: 65079111-65079111 on Build GRCh38
rs771706050 Chr.11: 65079083-65079083 on Build GRCh38
rs908186347 Chr.11: 65079217-65079217 on Build GRCh38
rs746988176 Chr.11: 65079276-65079276 on Build GRCh38
rs756457797 Chr.11: 65079344-65079347 on Build GRCh38
rs968604550 Chr.11: 65078982-65078982 on Build GRCh38
rs926076433 Chr.11: 65079317-65079317 on Build GRCh38
rs932518282 Chr.11: 65079282-65079282 on Build GRCh38
rs777747640 Chr.11: 65079336-65079336 on Build GRCh38
rs71468688 Chr.11: 65079070-65079070 on Build GRCh38
rs545138421 Chr.11: 65079020-65079020 on Build GRCh38
rs146179402 Chr.11: 65079127-65079127 on Build GRCh38
rs753110952 Chr.11: 65079116-65079116 on Build GRCh38
rs138639197 Chr.11: 65079153-65079153 on Build GRCh38
rs749041501 Chr.11: 65079062-65079062 on Build GRCh38
rs761852107 Chr.11: 65079082-65079082 on Build GRCh38
rs770661804 Chr.11: 65079178-65079178 on Build GRCh38
rs200160379 Chr.11: 65079086-65079086 on Build GRCh38
rs943818184 Chr.11: 65079225-65079225 on Build GRCh38
rs200209838 Chr.11: 65079364-65079364 on Build GRCh38
rs570301807 Chr.11: 65079183-65079183 on Build GRCh38
rs778518102 Chr.11: 65079265-65079265 on Build GRCh38
rs780506805 Chr.11: 65079348-65079348 on Build GRCh38
rs773223782 Chr.11: 65079179-65079179 on Build GRCh38

View Full Product Details