Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65714450-65714958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAT5
Entrez Gene ID
10524
Gene Name
lysine acetyltransferase 5
Gene Aliases
ESA1, HTATIP, HTATIP1, PLIP, TIP, TIP60, ZC2HC5, cPLA2
UniGene
Hs.397010
Gene Chromosome Location
Chr.11: 65711996-65719606 on Build GRCh38
Transcripts
NM_001206833.1 NM_006388.3 NM_182709.2 NM_182710.2 XM_006718421.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2166364 c.868T>C p.L290L Chr.11: 65714672-65714672 on Build GRCh38
4694049 c.815G>A p.R272Q Chr.11: 65714619-65714619 on Build GRCh38
5692446 c.811G>T p.G271C Chr.11: 65714615-65714615 on Build GRCh38
689975 c.956G>T p.R319L Chr.11: 65714837-65714837 on Build GRCh38
6998108 c.939+7G>A p.? Chr.11: 65714750-65714750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540917689 Chr.11: 65714826-65714826 on Build GRCh38
rs758116069 Chr.11: 65714763-65714763 on Build GRCh38
rs746803643 Chr.11: 65714805-65714805 on Build GRCh38
rs754709342 Chr.11: 65714656-65714656 on Build GRCh38
rs368546143 Chr.11: 65714936-65714936 on Build GRCh38
rs1009501704 Chr.11: 65714734-65714734 on Build GRCh38
rs892470790 Chr.11: 65714726-65714726 on Build GRCh38
rs927064729 Chr.11: 65714509-65714509 on Build GRCh38
rs912789543 Chr.11: 65714602-65714602 on Build GRCh38
rs778738215 Chr.11: 65714658-65714658 on Build GRCh38
rs371854405 Chr.11: 65714569-65714569 on Build GRCh38
rs751843208 Chr.11: 65714770-65714770 on Build GRCh38
rs373064578 Chr.11: 65714812-65714812 on Build GRCh38
rs758972621 Chr.11: 65714736-65714736 on Build GRCh38
rs755643538 Chr.11: 65714665-65714667 on Build GRCh38
rs1002845271 Chr.11: 65714670-65714670 on Build GRCh38
rs769880953 Chr.11: 65714604-65714604 on Build GRCh38
rs753256055 Chr.11: 65714654-65714654 on Build GRCh38
rs374634940 Chr.11: 65714786-65714786 on Build GRCh38
rs775104942 Chr.11: 65714822-65714822 on Build GRCh38
rs779377159 Chr.11: 65714526-65714526 on Build GRCh38
rs986444927 Chr.11: 65714832-65714832 on Build GRCh38
rs944216149 Chr.11: 65714634-65714634 on Build GRCh38
rs570448126 Chr.11: 65714607-65714607 on Build GRCh38
rs200974760 Chr.11: 65714517-65714517 on Build GRCh38
rs760518156 Chr.11: 65714649-65714649 on Build GRCh38
rs773110400 Chr.11: 65714799-65714799 on Build GRCh38
rs562899965 Chr.11: 65714581-65714581 on Build GRCh38
rs1000948431 Chr.11: 65714683-65714683 on Build GRCh38
rs753586481 Chr.11: 65714527-65714527 on Build GRCh38
rs781377393 Chr.11: 65714692-65714692 on Build GRCh38
rs778427253 Chr.11: 65714578-65714578 on Build GRCh38
rs4645923 Chr.11: 65714935-65714935 on Build GRCh38
rs144349144 Chr.11: 65714635-65714635 on Build GRCh38
rs143910658 Chr.11: 65714863-65714863 on Build GRCh38
rs574430605 Chr.11: 65714530-65714530 on Build GRCh38
rs776567096 Chr.11: 65714596-65714596 on Build GRCh38
rs745952989 Chr.11: 65714701-65714701 on Build GRCh38
rs759504006 Chr.11: 65714814-65714814 on Build GRCh38
rs749348240 Chr.11: 65714925-65714925 on Build GRCh38
rs201817325 Chr.11: 65714787-65714787 on Build GRCh38
rs958777577 Chr.11: 65714707-65714707 on Build GRCh38
rs764906283 Chr.11: 65714815-65714815 on Build GRCh38
rs139506891 Chr.11: 65714600-65714600 on Build GRCh38
rs772988323 Chr.11: 65714714-65714714 on Build GRCh38
rs982429356 Chr.11: 65714937-65714937 on Build GRCh38
rs767468018 Chr.11: 65714502-65714502 on Build GRCh38
rs747460009 Chr.11: 65714579-65714579 on Build GRCh38
rs148757761 Chr.11: 65714713-65714713 on Build GRCh38
rs752331334 Chr.11: 65714660-65714660 on Build GRCh38
rs750953520 Chr.11: 65714891-65714891 on Build GRCh38
rs756611914 Chr.11: 65714895-65714895 on Build GRCh38
rs531564426 Chr.11: 65714487-65714487 on Build GRCh38
rs776444982 Chr.11: 65714813-65714813 on Build GRCh38
rs772524421 Chr.11: 65714572-65714572 on Build GRCh38
rs546757404 Chr.11: 65714835-65714835 on Build GRCh38
rs780148798 Chr.11: 65714918-65714918 on Build GRCh38
rs890857562 Chr.11: 65714480-65714480 on Build GRCh38
rs752454142 Chr.11: 65714752-65714752 on Build GRCh38
rs138358637 Chr.11: 65714810-65714810 on Build GRCh38
rs149683297 Chr.11: 65714624-65714624 on Build GRCh38
rs184003597 Chr.11: 65714479-65714479 on Build GRCh38
rs763859516 Chr.11: 65714843-65714843 on Build GRCh38
rs200620312 Chr.11: 65714619-65714619 on Build GRCh38
rs749456791 Chr.11: 65714711-65714711 on Build GRCh38
rs768836164 Chr.11: 65714795-65714795 on Build GRCh38
rs754585319 Chr.11: 65714551-65714551 on Build GRCh38
rs377190873 Chr.11: 65714929-65714929 on Build GRCh38
rs368421147 Chr.11: 65714512-65714512 on Build GRCh38
rs758845267 Chr.11: 65714671-65714671 on Build GRCh38
rs111345240 Chr.11: 65714710-65714710 on Build GRCh38
rs781584699 Chr.11: 65714773-65714773 on Build GRCh38
rs765982091 Chr.11: 65714650-65714650 on Build GRCh38
rs764480726 Chr.11: 65714816-65714816 on Build GRCh38
rs766204816 Chr.11: 65714732-65714732 on Build GRCh38
rs868643963 Chr.11: 65714585-65714585 on Build GRCh38
rs4645921 Chr.11: 65714561-65714561 on Build GRCh38
rs775645186 Chr.11: 65714606-65714606 on Build GRCh38
rs755003246 Chr.11: 65714927-65714927 on Build GRCh38
rs769882159 Chr.11: 65714705-65714705 on Build GRCh38
rs757570608 Chr.11: 65714871-65714871 on Build GRCh38
rs4645922 Chr.11: 65714930-65714930 on Build GRCh38
rs746322226 Chr.11: 65714775-65714775 on Build GRCh38
rs114058675 Chr.11: 65714672-65714672 on Build GRCh38
rs765897444 Chr.11: 65714774-65714775 on Build GRCh38
rs764591239 Chr.11: 65714767-65714767 on Build GRCh38
rs767867886 Chr.11: 65714883-65714883 on Build GRCh38
rs771308702 Chr.11: 65714680-65714680 on Build GRCh38
rs776590286 Chr.11: 65714735-65714735 on Build GRCh38
rs554450356 Chr.11: 65714524-65714524 on Build GRCh38
rs756204694 Chr.11: 65714779-65714779 on Build GRCh38
rs111512947 Chr.11: 65714825-65714825 on Build GRCh38
rs760709797 Chr.11: 65714715-65714715 on Build GRCh38
rs368796194 Chr.11: 65714677-65714677 on Build GRCh38
rs764779726 Chr.11: 65714750-65714750 on Build GRCh38
rs973907454 Chr.11: 65714808-65714808 on Build GRCh38

View Full Product Details