Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65959599-65960096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSGA10IP
Entrez Gene ID
254187
Gene Name
testis specific 10 interacting protein
Gene Aliases
FAM161C
UniGene
Hs.350671
Gene Chromosome Location
Chr.11: 65945445-65960437 on Build GRCh38
Transcripts
NM_152762.2 XM_011544887.2 XM_011544888.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5764143 c.1485G>A p.M495I Chr.11: 65959844-65959844 on Build GRCh38
5966820 c.1567G>A p.E523K Chr.11: 65959926-65959926 on Build GRCh38
6341717 c.1556G>T p.S519I Chr.11: 65959915-65959915 on Build GRCh38
5448240 c.1524C>T p.T508T Chr.11: 65959883-65959883 on Build GRCh38
5696888 c.1493C>A p.S498Y Chr.11: 65959852-65959852 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376240417 Chr.11: 65959920-65959920 on Build GRCh38
rs768999990 Chr.11: 65959950-65959950 on Build GRCh38
rs752382856 Chr.11: 65959943-65959944 on Build GRCh38
rs767599562 Chr.11: 65959882-65959882 on Build GRCh38
rs756911090 Chr.11: 65959925-65959925 on Build GRCh38
rs768807897 Chr.11: 65959859-65959859 on Build GRCh38
rs946899428 Chr.11: 65959645-65959645 on Build GRCh38
rs1054968229 Chr.11: 65959683-65959683 on Build GRCh38
rs546592209 Chr.11: 65959720-65959720 on Build GRCh38
rs779181054 Chr.11: 65959953-65959953 on Build GRCh38
rs770308687 Chr.11: 65959833-65959833 on Build GRCh38
rs1056710291 Chr.11: 65959709-65959709 on Build GRCh38
rs780606899 Chr.11: 65959821-65959821 on Build GRCh38
rs994690077 Chr.11: 65959847-65959847 on Build GRCh38
rs780552395 Chr.11: 65959943-65959943 on Build GRCh38
rs557103231 Chr.11: 65959901-65959901 on Build GRCh38
rs371897877 Chr.11: 65959981-65959981 on Build GRCh38
rs559700576 Chr.11: 65960021-65960021 on Build GRCh38
rs756787589 Chr.11: 65959820-65959820 on Build GRCh38
rs749655109 Chr.11: 65959848-65959848 on Build GRCh38
rs935613383 Chr.11: 65959703-65959703 on Build GRCh38
rs75132850 Chr.11: 65959884-65959884 on Build GRCh38
rs754910396 Chr.11: 65959776-65959776 on Build GRCh38
rs927579464 Chr.11: 65959934-65959934 on Build GRCh38
rs758257359 Chr.11: 65959907-65959907 on Build GRCh38
rs539717642 Chr.11: 65959705-65959705 on Build GRCh38
rs745400225 Chr.11: 65959927-65959927 on Build GRCh38
rs140431543 Chr.11: 65959646-65959646 on Build GRCh38
rs367820057 Chr.11: 65959883-65959883 on Build GRCh38
rs746701382 Chr.11: 65959818-65959818 on Build GRCh38
rs756544399 Chr.11: 65959930-65959930 on Build GRCh38
rs368588379 Chr.11: 65959954-65959954 on Build GRCh38
rs780705450 Chr.11: 65959926-65959926 on Build GRCh38
rs759232447 Chr.11: 65959874-65959875 on Build GRCh38
rs773279041 Chr.11: 65959962-65959962 on Build GRCh38
rs924145057 Chr.11: 65960030-65960030 on Build GRCh38
rs146564888 Chr.11: 65960062-65960062 on Build GRCh38
rs371918833 Chr.11: 65960016-65960016 on Build GRCh38
rs76654835 Chr.11: 65959844-65959844 on Build GRCh38
rs267603120 Chr.11: 65959935-65959935 on Build GRCh38
rs1027519338 Chr.11: 65959849-65959849 on Build GRCh38
rs369312564 Chr.11: 65960037-65960037 on Build GRCh38
rs491973 Chr.11: 65959830-65959830 on Build GRCh38
rs1004236052 Chr.11: 65959797-65959797 on Build GRCh38
rs542349404 Chr.11: 65960052-65960052 on Build GRCh38
rs758060674 Chr.11: 65959804-65959804 on Build GRCh38
rs758064902 Chr.11: 65959953-65959960 on Build GRCh38
rs267603121 Chr.11: 65959936-65959936 on Build GRCh38
rs764047910 Chr.11: 65959917-65959917 on Build GRCh38
rs546138370 Chr.11: 65959990-65959990 on Build GRCh38
rs1047667498 Chr.11: 65959750-65959750 on Build GRCh38
rs752574700 Chr.11: 65959802-65959802 on Build GRCh38
rs577134022 Chr.11: 65959977-65959977 on Build GRCh38
rs566306362 Chr.11: 65959807-65959807 on Build GRCh38
rs752486913 Chr.11: 65959890-65959890 on Build GRCh38
rs765100178 Chr.11: 65959801-65959801 on Build GRCh38
rs530755098 Chr.11: 65960069-65960069 on Build GRCh38
rs765273976 Chr.11: 65959889-65959889 on Build GRCh38
rs753805421 Chr.11: 65959768-65959768 on Build GRCh38
rs184743948 Chr.11: 65960061-65960061 on Build GRCh38
rs776507971 Chr.11: 65959852-65959854 on Build GRCh38
rs948038617 Chr.11: 65959826-65959826 on Build GRCh38
rs924092597 Chr.11: 65959650-65959650 on Build GRCh38
rs918139615 Chr.11: 65960065-65960065 on Build GRCh38
rs550929344 Chr.11: 65959644-65959644 on Build GRCh38
rs762003887 Chr.11: 65959869-65959869 on Build GRCh38
rs67530871 Chr.11: 65959868-65959868 on Build GRCh38
rs765024561 Chr.11: 65959923-65959928 on Build GRCh38

View Full Product Details