Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 57128266-57128785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VAMP7
Entrez Gene ID
6845
Gene Name
vesicle associated membrane protein 7
Gene Aliases
SYBL1, TI-VAMP, TIVAMP, VAMP-7
UniGene
Hs.24167
Gene Chromosome Location
Chr.Y: 57067800-57130289 on Build GRCh38
Transcripts
NM_001145149.2 NM_001185183.1 NM_005638.5 NR_033714.1 NR_033715.1 XM_011531188.1 XM_017029760.1

SNP Detail

SNP ID Chromosome Location
rs778453493 Chr.X: 155941804-155941804 on Build GRCh38
rs185857002 Chr.X: 155942100-155942100 on Build GRCh38
rs966070206 Chr.X: 155942223-155942223 on Build GRCh38
rs746891950 Chr.X: 155942011-155942011 on Build GRCh38
rs994046020 Chr.X: 155942145-155942145 on Build GRCh38
rs746803961 Chr.X: 155941883-155941883 on Build GRCh38
rs377236940 Chr.X: 155941839-155941839 on Build GRCh38
rs749893726 Chr.X: 155942200-155942200 on Build GRCh38
rs200474856 Chr.X: 155942183-155942183 on Build GRCh38
rs759887584 Chr.X: 155942077-155942077 on Build GRCh38
rs914968803 Chr.X: 155942061-155942061 on Build GRCh38
rs754209049 Chr.X: 155941990-155941990 on Build GRCh38
rs372687315 Chr.X: 155942010-155942010 on Build GRCh38
rs750865861 Chr.X: 155941974-155941974 on Build GRCh38
rs745667935 Chr.X: 155942035-155942035 on Build GRCh38
rs779764036 Chr.X: 155941868-155941868 on Build GRCh38
rs763130384 Chr.X: 155941893-155941893 on Build GRCh38
rs769961185 Chr.X: 155941977-155941977 on Build GRCh38
rs761888485 Chr.X: 155942041-155942041 on Build GRCh38
rs765397649 Chr.X: 155942211-155942211 on Build GRCh38
rs770806845 Chr.X: 155941885-155941885 on Build GRCh38
rs780886605 Chr.X: 155942024-155942024 on Build GRCh38
rs977115691 Chr.X: 155941916-155941916 on Build GRCh38
rs779177484 Chr.X: 155942043-155942044 on Build GRCh38
rs752268059 Chr.X: 155942006-155942006 on Build GRCh38
rs150795669 Chr.X: 155941907-155941907 on Build GRCh38
rs774365426 Chr.X: 155941948-155941948 on Build GRCh38
rs904905362 Chr.X: 155942096-155942096 on Build GRCh38
rs768028469 Chr.X: 155941971-155941971 on Build GRCh38
rs745430429 Chr.X: 155941805-155941805 on Build GRCh38
rs748223133 Chr.X: 155942075-155942075 on Build GRCh38
rs1027833483 Chr.X: 155942148-155942148 on Build GRCh38
rs754223645 Chr.X: 155942076-155942076 on Build GRCh38
rs376892030 Chr.X: 155941879-155941879 on Build GRCh38
rs757018318 Chr.X: 155941833-155941835 on Build GRCh38
rs781131598 Chr.X: 155941853-155941857 on Build GRCh38
rs772102935 Chr.X: 155942049-155942049 on Build GRCh38
rs200648785 Chr.X: 155941834-155941834 on Build GRCh38
rs751633839 Chr.X: 155941847-155941847 on Build GRCh38
rs200420885 Chr.X: 155941888-155941888 on Build GRCh38
rs745860800 Chr.X: 155941954-155941956 on Build GRCh38
rs758003477 Chr.X: 155942113-155942113 on Build GRCh38
rs761221796 Chr.X: 155942067-155942067 on Build GRCh38
rs760583658 Chr.X: 155942103-155942103 on Build GRCh38
rs188664444 Chr.X: 155941960-155941960 on Build GRCh38
rs768823022 Chr.X: 155942036-155942036 on Build GRCh38
rs922454198 Chr.X: 155941956-155941956 on Build GRCh38
rs761502433 Chr.X: 155942155-155942155 on Build GRCh38
rs773429537 Chr.X: 155942184-155942184 on Build GRCh38
rs757796940 Chr.X: 155941851-155941851 on Build GRCh38
rs754539784 Chr.X: 155941870-155941870 on Build GRCh38
rs772208034 Chr.X: 155942086-155942086 on Build GRCh38
rs764800652 Chr.X: 155942081-155942081 on Build GRCh38
rs1007652940 Chr.X: 155942171-155942171 on Build GRCh38
rs369252099 Chr.X: 155941869-155941869 on Build GRCh38
rs778336440 Chr.X: 155941872-155941872 on Build GRCh38
rs766605651 Chr.X: 155941978-155941978 on Build GRCh38
rs781180032 Chr.X: 155942167-155942167 on Build GRCh38
rs773379694 Chr.X: 155941995-155941995 on Build GRCh38
rs1003789587 Chr.X: 155942097-155942097 on Build GRCh38
rs753613945 Chr.X: 155942126-155942126 on Build GRCh38
rs778614692 Chr.X: 155942005-155942005 on Build GRCh38
rs764027936 Chr.X: 155941842-155941842 on Build GRCh38
rs5940625 Chr.X: 155941873-155941873 on Build GRCh38
rs952123561 Chr.X: 155941852-155941852 on Build GRCh38
rs756099145 Chr.X: 155941963-155941963 on Build GRCh38
rs777471575 Chr.X: 155942121-155942121 on Build GRCh38
rs747317445 Chr.X: 155941863-155941863 on Build GRCh38
rs768012099 Chr.X: 155941853-155941853 on Build GRCh38
rs192817973 Chr.X: 155942038-155942038 on Build GRCh38
rs768887885 Chr.X: 155941946-155941946 on Build GRCh38
rs770875892 Chr.X: 155942021-155942021 on Build GRCh38
rs763942778 Chr.X: 155942122-155942122 on Build GRCh38
rs773732812 Chr.X: 155942058-155942058 on Build GRCh38
rs182424380 Chr.X: 155941808-155941808 on Build GRCh38
rs6642256 Chr.X: 155942173-155942173 on Build GRCh38

View Full Product Details