Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 104117789-104118350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFYB
Entrez Gene ID
4801
Gene Name
nuclear transcription factor Y subunit beta
Gene Aliases
CBF-A, CBF-B, HAP3, NF-YB
UniGene
Hs.84928
Gene Chromosome Location
Chr.12: 104117080-104138262 on Build GRCh38
Transcripts
NM_006166.3 XM_005268908.4 XM_005268909.4 XM_005268910.4

SNP Detail

SNP ID Chromosome Location
rs950450822 Chr.12: 104118287-104118287 on Build GRCh38
rs891198373 Chr.12: 104118188-104118188 on Build GRCh38
rs547908346 Chr.12: 104118113-104118113 on Build GRCh38
rs911443444 Chr.12: 104118280-104118282 on Build GRCh38
rs761419831 Chr.12: 104118024-104118024 on Build GRCh38
rs895102816 Chr.12: 104118045-104118045 on Build GRCh38
rs763581367 Chr.12: 104118276-104118276 on Build GRCh38
rs142097089 Chr.12: 104118025-104118025 on Build GRCh38
rs1055445099 Chr.12: 104118042-104118042 on Build GRCh38
rs1048530661 Chr.12: 104117992-104117992 on Build GRCh38
rs1018511679 Chr.12: 104117847-104117847 on Build GRCh38
rs1013823739 Chr.12: 104118056-104118056 on Build GRCh38
rs73177982 Chr.12: 104118229-104118229 on Build GRCh38
rs918663990 Chr.12: 104117934-104117934 on Build GRCh38
rs562255551 Chr.12: 104117872-104117872 on Build GRCh38
rs966533925 Chr.12: 104118103-104118103 on Build GRCh38
rs983502392 Chr.12: 104118311-104118311 on Build GRCh38
rs962906927 Chr.12: 104117867-104117867 on Build GRCh38
rs929053131 Chr.12: 104117948-104117948 on Build GRCh38
rs756645213 Chr.12: 104118232-104118232 on Build GRCh38
rs547699419 Chr.12: 104118278-104118278 on Build GRCh38
rs985576235 Chr.12: 104117820-104117820 on Build GRCh38
rs528017465 Chr.12: 104118152-104118152 on Build GRCh38
rs770402053 Chr.12: 104118117-104118117 on Build GRCh38
rs920237231 Chr.12: 104117939-104117939 on Build GRCh38
rs935879844 Chr.12: 104118035-104118035 on Build GRCh38
rs999191189 Chr.12: 104118126-104118130 on Build GRCh38
rs763832079 Chr.12: 104117976-104117976 on Build GRCh38
rs1026306936 Chr.12: 104118284-104118284 on Build GRCh38
rs762823081 Chr.12: 104117899-104117899 on Build GRCh38
rs774018587 Chr.12: 104118013-104118013 on Build GRCh38
rs548067539 Chr.12: 104118161-104118161 on Build GRCh38
rs34781342 Chr.12: 104117940-104117941 on Build GRCh38
rs1044236473 Chr.12: 104118019-104118019 on Build GRCh38
rs568035562 Chr.12: 104118151-104118151 on Build GRCh38
rs766912899 Chr.12: 104118265-104118265 on Build GRCh38
rs1051213485 Chr.12: 104118155-104118155 on Build GRCh38
rs916981978 Chr.12: 104117980-104117980 on Build GRCh38
rs1010650264 Chr.12: 104118154-104118154 on Build GRCh38
rs888411647 Chr.12: 104117998-104117998 on Build GRCh38
rs775226968 Chr.12: 104117871-104117871 on Build GRCh38
rs571112750 Chr.12: 104118211-104118211 on Build GRCh38
rs965688381 Chr.12: 104118306-104118306 on Build GRCh38
rs1024735129 Chr.12: 104118057-104118057 on Build GRCh38
rs949885842 Chr.12: 104117996-104117996 on Build GRCh38
rs556761135 Chr.12: 104118285-104118285 on Build GRCh38
rs983252163 Chr.12: 104117957-104117957 on Build GRCh38

View Full Product Details