Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43522036-43522542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP1A
Entrez Gene ID
4130
Gene Name
microtubule associated protein 1A
Gene Aliases
MAP1L, MTAP1A
UniGene
Hs.194301
Gene Chromosome Location
Chr.15: 43510946-43531620 on Build GRCh38
Transcripts
NM_002373.5 XM_017022189.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2188376 c.740T>C p.I247T Chr.15: 43522213-43522213 on Build GRCh38
4556742 c.700G>A p.G234R Chr.15: 43522173-43522173 on Build GRCh38
3816144 c.922C>T p.L308F Chr.15: 43522395-43522395 on Build GRCh38
5920842 c.571C>T p.H191Y Chr.15: 43522044-43522044 on Build GRCh38
3501442 c.772C>T p.P258S Chr.15: 43522245-43522245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745638085 Chr.15: 43522460-43522460 on Build GRCh38
rs367933241 Chr.15: 43522420-43522420 on Build GRCh38
rs762817512 Chr.15: 43522205-43522205 on Build GRCh38
rs749180163 Chr.15: 43522340-43522340 on Build GRCh38
rs202066894 Chr.15: 43522384-43522384 on Build GRCh38
rs772286968 Chr.15: 43522171-43522171 on Build GRCh38
rs746050439 Chr.15: 43522166-43522166 on Build GRCh38
rs139538952 Chr.15: 43522459-43522459 on Build GRCh38
rs553817490 Chr.15: 43522372-43522372 on Build GRCh38
rs757704711 Chr.15: 43522383-43522383 on Build GRCh38
rs868318538 Chr.15: 43522188-43522188 on Build GRCh38
rs746245048 Chr.15: 43522269-43522269 on Build GRCh38
rs768207669 Chr.15: 43522482-43522482 on Build GRCh38
rs267604220 Chr.15: 43522279-43522279 on Build GRCh38
rs780294832 Chr.15: 43522277-43522277 on Build GRCh38
rs200513920 Chr.15: 43522504-43522504 on Build GRCh38
rs771378341 Chr.15: 43522136-43522136 on Build GRCh38
rs199517123 Chr.15: 43522193-43522193 on Build GRCh38
rs773249162 Chr.15: 43522203-43522203 on Build GRCh38
rs768004425 Chr.15: 43522462-43522462 on Build GRCh38
rs1007093669 Chr.15: 43522063-43522063 on Build GRCh38
rs200067722 Chr.15: 43522416-43522416 on Build GRCh38
rs779272678 Chr.15: 43522158-43522158 on Build GRCh38
rs766233755 Chr.15: 43522212-43522212 on Build GRCh38
rs754323568 Chr.15: 43522262-43522262 on Build GRCh38
rs965392597 Chr.15: 43522160-43522160 on Build GRCh38
rs201763786 Chr.15: 43522506-43522506 on Build GRCh38
rs774289114 Chr.15: 43522235-43522235 on Build GRCh38
rs201117585 Chr.15: 43522427-43522427 on Build GRCh38
rs759202922 Chr.15: 43522238-43522238 on Build GRCh38
rs781271816 Chr.15: 43522424-43522424 on Build GRCh38
rs769586017 Chr.15: 43522194-43522194 on Build GRCh38
rs775181769 Chr.15: 43522471-43522471 on Build GRCh38
rs576147041 Chr.15: 43522475-43522475 on Build GRCh38
rs201259084 Chr.15: 43522442-43522442 on Build GRCh38
rs763481070 Chr.15: 43522518-43522518 on Build GRCh38
rs759475015 Chr.15: 43522357-43522357 on Build GRCh38
rs776191838 Chr.15: 43522483-43522483 on Build GRCh38
rs779182721 Chr.15: 43522264-43522264 on Build GRCh38
rs767341462 Chr.15: 43522241-43522241 on Build GRCh38
rs989572023 Chr.15: 43522304-43522304 on Build GRCh38
rs775265657 Chr.15: 43522359-43522359 on Build GRCh38
rs762811768 Chr.15: 43522243-43522243 on Build GRCh38
rs767249834 Chr.15: 43522358-43522358 on Build GRCh38
rs747036548 Chr.15: 43522318-43522318 on Build GRCh38
rs34542263 Chr.15: 43522172-43522173 on Build GRCh38
rs1060935 Chr.15: 43522476-43522476 on Build GRCh38
rs755157026 Chr.15: 43522419-43522419 on Build GRCh38
rs780417652 Chr.15: 43522394-43522394 on Build GRCh38
rs145078659 Chr.15: 43522519-43522519 on Build GRCh38
rs760268968 Chr.15: 43522361-43522361 on Build GRCh38
rs778414449 Chr.15: 43522071-43522071 on Build GRCh38
rs999959286 Chr.15: 43522161-43522161 on Build GRCh38
rs576946901 Chr.15: 43522390-43522390 on Build GRCh38
rs768730706 Chr.15: 43522323-43522323 on Build GRCh38
rs201399898 Chr.15: 43522342-43522342 on Build GRCh38
rs867023856 Chr.15: 43522393-43522393 on Build GRCh38
rs1036407960 Chr.15: 43522463-43522463 on Build GRCh38
rs770990875 Chr.15: 43522428-43522428 on Build GRCh38
rs761352640 Chr.15: 43522503-43522503 on Build GRCh38
rs1060936 Chr.15: 43522480-43522480 on Build GRCh38
rs764488460 Chr.15: 43522261-43522261 on Build GRCh38
rs1020318566 Chr.15: 43522097-43522097 on Build GRCh38
rs763622955 Chr.15: 43522364-43522364 on Build GRCh38
rs749586688 Chr.15: 43522091-43522091 on Build GRCh38
rs568161996 Chr.15: 43522181-43522181 on Build GRCh38
rs755693795 Chr.15: 43522248-43522248 on Build GRCh38
rs758615612 Chr.15: 43522274-43522274 on Build GRCh38
rs757738188 Chr.15: 43522263-43522263 on Build GRCh38
rs771947278 Chr.15: 43522464-43522464 on Build GRCh38
rs776603796 Chr.15: 43522331-43522331 on Build GRCh38
rs958080607 Chr.15: 43522251-43522251 on Build GRCh38

View Full Product Details