Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 25834238-25834745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AUNIP
Entrez Gene ID
79000
Gene Name
aurora kinase A and ninein interacting protein
Gene Aliases
AIBP, C1orf135
UniGene
Hs.149305
Gene Chromosome Location
Chr.1: 25831913-25859458 on Build GRCh38
Transcripts
NM_001287490.1 NM_024037.2 XM_017002300.1

SNP Detail

SNP ID Chromosome Location
rs995677481 Chr.1: 25834666-25834666 on Build GRCh38
rs554326530 Chr.1: 25834445-25834445 on Build GRCh38
rs939262839 Chr.1: 25834662-25834662 on Build GRCh38
rs1046061496 Chr.1: 25834634-25834634 on Build GRCh38
rs796407829 Chr.1: 25834616-25834617 on Build GRCh38
rs764723271 Chr.1: 25834344-25834344 on Build GRCh38
rs966272182 Chr.1: 25834543-25834543 on Build GRCh38
rs1042203417 Chr.1: 25834356-25834356 on Build GRCh38
rs894503971 Chr.1: 25834424-25834424 on Build GRCh38
rs561995898 Chr.1: 25834567-25834567 on Build GRCh38
rs924658046 Chr.1: 25834390-25834390 on Build GRCh38
rs923864417 Chr.1: 25834381-25834381 on Build GRCh38
rs943907843 Chr.1: 25834348-25834348 on Build GRCh38
rs751327082 Chr.1: 25834571-25834571 on Build GRCh38
rs540011145 Chr.1: 25834489-25834489 on Build GRCh38
rs745803624 Chr.1: 25834691-25834691 on Build GRCh38
rs954174484 Chr.1: 25834332-25834332 on Build GRCh38
rs953745297 Chr.1: 25834281-25834281 on Build GRCh38
rs1046598547 Chr.1: 25834443-25834443 on Build GRCh38
rs115903042 Chr.1: 25834658-25834658 on Build GRCh38
rs112157382 Chr.1: 25834517-25834517 on Build GRCh38
rs986370924 Chr.1: 25834296-25834296 on Build GRCh38
rs947743059 Chr.1: 25834633-25834633 on Build GRCh38
rs1045768968 Chr.1: 25834630-25834635 on Build GRCh38
rs1037215263 Chr.1: 25834646-25834646 on Build GRCh38
rs758049464 Chr.1: 25834409-25834409 on Build GRCh38
rs558563658 Chr.1: 25834318-25834318 on Build GRCh38
rs912061286 Chr.1: 25834329-25834329 on Build GRCh38
rs139695812 Chr.1: 25834633-25834634 on Build GRCh38
rs1014784014 Chr.1: 25834486-25834486 on Build GRCh38
rs147711724 Chr.1: 25834542-25834542 on Build GRCh38
rs768489838 Chr.1: 25834719-25834719 on Build GRCh38
rs188171076 Chr.1: 25834461-25834461 on Build GRCh38
rs761683970 Chr.1: 25834340-25834340 on Build GRCh38
rs940386852 Chr.1: 25834638-25834638 on Build GRCh38
rs924814561 Chr.1: 25834554-25834554 on Build GRCh38
rs538669106 Chr.1: 25834304-25834304 on Build GRCh38
rs912687405 Chr.1: 25834550-25834550 on Build GRCh38
rs948733257 Chr.1: 25834625-25834626 on Build GRCh38
rs906283579 Chr.1: 25834636-25834636 on Build GRCh38
rs994626543 Chr.1: 25834499-25834499 on Build GRCh38
rs936617957 Chr.1: 25834588-25834588 on Build GRCh38
rs147839303 Chr.1: 25834338-25834338 on Build GRCh38
rs1015730431 Chr.1: 25834503-25834503 on Build GRCh38
rs1045042722 Chr.1: 25834459-25834460 on Build GRCh38
rs11247767 Chr.1: 25834595-25834595 on Build GRCh38
rs906356673 Chr.1: 25834467-25834467 on Build GRCh38
rs1027335565 Chr.1: 25834268-25834268 on Build GRCh38
rs916486641 Chr.1: 25834631-25834631 on Build GRCh38
rs989825421 Chr.1: 25834627-25834627 on Build GRCh38
rs977904084 Chr.1: 25834564-25834564 on Build GRCh38
rs545822080 Chr.1: 25834532-25834532 on Build GRCh38
rs370973487 Chr.1: 25834531-25834531 on Build GRCh38
rs751599793 Chr.1: 25834410-25834410 on Build GRCh38
rs574986323 Chr.1: 25834602-25834602 on Build GRCh38
rs548673990 Chr.1: 25834563-25834563 on Build GRCh38
rs985973101 Chr.1: 25834335-25834335 on Build GRCh38
rs748528267 Chr.1: 25834616-25834618 on Build GRCh38
rs575292049 Chr.1: 25834355-25834355 on Build GRCh38
rs1053470857 Chr.1: 25834400-25834400 on Build GRCh38
rs368988772 Chr.1: 25834515-25834515 on Build GRCh38
rs780340998 Chr.1: 25834617-25834618 on Build GRCh38
rs897542199 Chr.1: 25834498-25834498 on Build GRCh38
rs1011894739 Chr.1: 25834428-25834428 on Build GRCh38
rs58893037 Chr.1: 25834391-25834391 on Build GRCh38
rs191524527 Chr.1: 25834478-25834478 on Build GRCh38
rs7555012 Chr.1: 25834546-25834546 on Build GRCh38

View Full Product Details