Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26282469-26282968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBXN11
Entrez Gene ID
91544
Gene Name
UBX domain protein 11
Gene Aliases
COA-1, PP2243, SOC, SOCI, UBXD5
UniGene
Hs.145061
Gene Chromosome Location
Chr.1: 26282282-26318265 on Build GRCh38
Transcripts
NM_001077262.1 NM_145345.2 NM_183008.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
907612 c.1235T>C p.L412P Chr.1: 26282706-26282706 on Build GRCh38
5417883 c.1141G>A p.E381K Chr.1: 26282874-26282874 on Build GRCh38
907613 c.1206G>T p.K402N Chr.1: 26282735-26282735 on Build GRCh38
3804796 c.1129A>C p.T377P Chr.1: 26282886-26282886 on Build GRCh38
1341325 c.1132T>G p.L378V Chr.1: 26282883-26282883 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753366267 Chr.1: 26282489-26282489 on Build GRCh38
rs559953707 Chr.1: 26282594-26282594 on Build GRCh38
rs763509201 Chr.1: 26282539-26282539 on Build GRCh38
rs758221664 Chr.1: 26282649-26282649 on Build GRCh38
rs779408641 Chr.1: 26282494-26282494 on Build GRCh38
rs375717967 Chr.1: 26282850-26282850 on Build GRCh38
rs757493934 Chr.1: 26282779-26282780 on Build GRCh38
rs371919070 Chr.1: 26282627-26282627 on Build GRCh38
rs779159321 Chr.1: 26282802-26282802 on Build GRCh38
rs745844837 Chr.1: 26282526-26282526 on Build GRCh38
rs367697574 Chr.1: 26282498-26282498 on Build GRCh38
rs376402622 Chr.1: 26282648-26282648 on Build GRCh38
rs1022435142 Chr.1: 26282793-26282793 on Build GRCh38
rs948328554 Chr.1: 26282530-26282530 on Build GRCh38
rs773062281 Chr.1: 26282743-26282743 on Build GRCh38
rs370051543 Chr.1: 26282855-26282855 on Build GRCh38
rs201128406 Chr.1: 26282938-26282938 on Build GRCh38
rs777983942 Chr.1: 26282681-26282681 on Build GRCh38
rs752572937 Chr.1: 26282945-26282945 on Build GRCh38
rs761901021 Chr.1: 26282839-26282839 on Build GRCh38
rs754740134 Chr.1: 26282808-26282808 on Build GRCh38
rs761538125 Chr.1: 26282564-26282564 on Build GRCh38
rs185659723 Chr.1: 26282674-26282674 on Build GRCh38
rs543842002 Chr.1: 26282863-26282863 on Build GRCh38
rs779693144 Chr.1: 26282688-26282688 on Build GRCh38
rs6598952 Chr.1: 26282816-26282816 on Build GRCh38
rs776405229 Chr.1: 26282558-26282558 on Build GRCh38
rs552206395 Chr.1: 26282618-26282618 on Build GRCh38
rs756296771 Chr.1: 26282615-26282615 on Build GRCh38
rs560998848 Chr.1: 26282508-26282508 on Build GRCh38
rs748096914 Chr.1: 26282815-26282815 on Build GRCh38
rs777496692 Chr.1: 26282672-26282672 on Build GRCh38
rs747331978 Chr.1: 26282891-26282891 on Build GRCh38
rs370592440 Chr.1: 26282818-26282818 on Build GRCh38
rs568645771 Chr.1: 26282675-26282675 on Build GRCh38
rs111606040 Chr.1: 26282766-26282766 on Build GRCh38
rs771374267 Chr.1: 26282750-26282750 on Build GRCh38
rs773368071 Chr.1: 26282923-26282923 on Build GRCh38
rs765560694 Chr.1: 26282783-26282785 on Build GRCh38
rs780669820 Chr.1: 26282499-26282499 on Build GRCh38
rs76807981 Chr.1: 26282757-26282757 on Build GRCh38
rs756092849 Chr.1: 26282870-26282870 on Build GRCh38
rs372393457 Chr.1: 26282919-26282919 on Build GRCh38
rs969495893 Chr.1: 26282812-26282813 on Build GRCh38
rs976720753 Chr.1: 26282940-26282940 on Build GRCh38
rs373886308 Chr.1: 26282829-26282829 on Build GRCh38
rs765393507 Chr.1: 26282527-26282527 on Build GRCh38
rs778434703 Chr.1: 26282883-26282883 on Build GRCh38
rs989640076 Chr.1: 26282504-26282504 on Build GRCh38
rs781275230 Chr.1: 26282813-26282813 on Build GRCh38
rs151118025 Chr.1: 26282878-26282878 on Build GRCh38
rs750853098 Chr.1: 26282569-26282569 on Build GRCh38
rs766644776 Chr.1: 26282659-26282659 on Build GRCh38
rs776033967 Chr.1: 26282827-26282827 on Build GRCh38
rs201808891 Chr.1: 26282658-26282658 on Build GRCh38
rs774108310 Chr.1: 26282522-26282522 on Build GRCh38
rs746909696 Chr.1: 26282628-26282628 on Build GRCh38
rs371990639 Chr.1: 26282915-26282915 on Build GRCh38
rs750760102 Chr.1: 26282836-26282837 on Build GRCh38
rs1040607564 Chr.1: 26282941-26282941 on Build GRCh38
rs775798959 Chr.1: 26282890-26282890 on Build GRCh38
rs748449843 Chr.1: 26282610-26282610 on Build GRCh38
rs980962849 Chr.1: 26282533-26282533 on Build GRCh38
rs372018986 Chr.1: 26282682-26282682 on Build GRCh38
rs372743701 Chr.1: 26282916-26282916 on Build GRCh38
rs764147178 Chr.1: 26282769-26282769 on Build GRCh38
rs375597048 Chr.1: 26282856-26282856 on Build GRCh38
rs143743472 Chr.1: 26282509-26282509 on Build GRCh38
rs7522905 Chr.1: 26282768-26282768 on Build GRCh38
rs752163039 Chr.1: 26282671-26282671 on Build GRCh38
rs72872912 Chr.1: 26282602-26282602 on Build GRCh38
rs752007022 Chr.1: 26282929-26282931 on Build GRCh38
rs928278007 Chr.1: 26282825-26282825 on Build GRCh38
rs374388078 Chr.1: 26282942-26282942 on Build GRCh38
rs893107744 Chr.1: 26282583-26282583 on Build GRCh38
rs781138685 Chr.1: 26282735-26282735 on Build GRCh38
rs748772070 Chr.1: 26282877-26282877 on Build GRCh38
rs751329711 Chr.1: 26282807-26282807 on Build GRCh38
rs558654564 Chr.1: 26282778-26282778 on Build GRCh38
rs748311785 Chr.1: 26282512-26282512 on Build GRCh38
rs769171435 Chr.1: 26282903-26282903 on Build GRCh38
rs374453800 Chr.1: 26282528-26282528 on Build GRCh38
rs981244201 Chr.1: 26282604-26282604 on Build GRCh38
rs755926379 Chr.1: 26282946-26282946 on Build GRCh38
rs759903974 Chr.1: 26282758-26282758 on Build GRCh38
rs752734270 Chr.1: 26282866-26282866 on Build GRCh38
rs766699813 Chr.1: 26282494-26282495 on Build GRCh38
rs544213824 Chr.1: 26282503-26282503 on Build GRCh38
rs755149678 Chr.1: 26282598-26282598 on Build GRCh38
rs201777136 Chr.1: 26282738-26282738 on Build GRCh38
rs1012846171 Chr.1: 26282588-26282588 on Build GRCh38
rs761148100 Chr.1: 26282828-26282828 on Build GRCh38
rs758709571 Chr.1: 26282844-26282846 on Build GRCh38
rs768264327 Chr.1: 26282644-26282644 on Build GRCh38
rs764262264 Chr.1: 26282542-26282542 on Build GRCh38
rs747009592 Chr.1: 26282696-26282696 on Build GRCh38
rs76733027 Chr.1: 26282756-26282756 on Build GRCh38
rs765387698 Chr.1: 26282844-26282844 on Build GRCh38
rs569018795 Chr.1: 26282622-26282622 on Build GRCh38
rs759831646 Chr.1: 26282820-26282820 on Build GRCh38
rs781106308 Chr.1: 26282751-26282751 on Build GRCh38
rs995330125 Chr.1: 26282884-26282884 on Build GRCh38
rs758248476 Chr.1: 26282686-26282686 on Build GRCh38
rs745520921 Chr.1: 26282885-26282885 on Build GRCh38
rs368771477 Chr.1: 26282871-26282871 on Build GRCh38
rs772945901 Chr.1: 26282656-26282656 on Build GRCh38
rs80173211 Chr.1: 26282538-26282538 on Build GRCh38
rs772414799 Chr.1: 26282822-26282822 on Build GRCh38
rs765008329 Chr.1: 26282650-26282650 on Build GRCh38
rs189256251 Chr.1: 26282742-26282742 on Build GRCh38
rs757708291 Chr.1: 26282490-26282490 on Build GRCh38
rs757392393 Chr.1: 26282798-26282798 on Build GRCh38
rs765739835 Chr.1: 26282567-26282567 on Build GRCh38
rs754025846 Chr.1: 26282621-26282622 on Build GRCh38
rs768510187 Chr.1: 26282701-26282701 on Build GRCh38
rs767931718 Chr.1: 26282765-26282765 on Build GRCh38
rs571526240 Chr.1: 26282864-26282865 on Build GRCh38
rs772473973 Chr.1: 26282889-26282889 on Build GRCh38
rs762678285 Chr.1: 26282748-26282748 on Build GRCh38
rs753948245 Chr.1: 26282836-26282836 on Build GRCh38
rs766918963 Chr.1: 26282592-26282592 on Build GRCh38
rs373852862 Chr.1: 26282497-26282497 on Build GRCh38
rs932316360 Chr.1: 26282718-26282718 on Build GRCh38
rs377309976 Chr.1: 26282817-26282817 on Build GRCh38
rs758790286 Chr.1: 26282589-26282589 on Build GRCh38

View Full Product Details