Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27406099-27406590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WASF2
Entrez Gene ID
10163
Gene Name
WAS protein family member 2
Gene Aliases
IMD2, SCAR2, WASF4, WAVE2, dJ393P12.2
UniGene
Hs.469244
Gene Chromosome Location
Chr.1: 27404226-27490187 on Build GRCh38
Transcripts
NM_001201404.2 NM_006990.4

SNP Detail

SNP ID Chromosome Location
rs970719703 Chr.1: 27406266-27406266 on Build GRCh38
rs563102800 Chr.1: 27406204-27406204 on Build GRCh38
rs966819653 Chr.1: 27406173-27406173 on Build GRCh38
rs1050083149 Chr.1: 27406155-27406155 on Build GRCh38
rs1037829629 Chr.1: 27406125-27406125 on Build GRCh38
rs190311048 Chr.1: 27406349-27406349 on Build GRCh38
rs958486415 Chr.1: 27406433-27406433 on Build GRCh38
rs549849558 Chr.1: 27406493-27406493 on Build GRCh38
rs34170682 Chr.1: 27406347-27406348 on Build GRCh38
rs576294583 Chr.1: 27406205-27406205 on Build GRCh38
rs991346391 Chr.1: 27406254-27406254 on Build GRCh38
rs778844677 Chr.1: 27406361-27406361 on Build GRCh38
rs890156640 Chr.1: 27406164-27406164 on Build GRCh38
rs11548324 Chr.1: 27406416-27406416 on Build GRCh38
rs890082918 Chr.1: 27406364-27406364 on Build GRCh38
rs1023732113 Chr.1: 27406471-27406471 on Build GRCh38
rs940514091 Chr.1: 27406122-27406122 on Build GRCh38
rs981991271 Chr.1: 27406300-27406300 on Build GRCh38
rs747884036 Chr.1: 27406396-27406396 on Build GRCh38
rs553841460 Chr.1: 27406149-27406149 on Build GRCh38
rs940482970 Chr.1: 27406347-27406347 on Build GRCh38
rs774187102 Chr.1: 27406543-27406543 on Build GRCh38
rs547442066 Chr.1: 27406384-27406384 on Build GRCh38
rs533295728 Chr.1: 27406417-27406417 on Build GRCh38
rs368761913 Chr.1: 27406159-27406159 on Build GRCh38
rs1008527047 Chr.1: 27406367-27406367 on Build GRCh38
rs868785473 Chr.1: 27406169-27406169 on Build GRCh38
rs562238081 Chr.1: 27406328-27406328 on Build GRCh38
rs184641949 Chr.1: 27406166-27406166 on Build GRCh38
rs916582110 Chr.1: 27406261-27406261 on Build GRCh38
rs192873980 Chr.1: 27406163-27406163 on Build GRCh38
rs542583735 Chr.1: 27406206-27406206 on Build GRCh38
rs535768438 Chr.1: 27406360-27406360 on Build GRCh38
rs533509261 Chr.1: 27406148-27406148 on Build GRCh38
rs200844631 Chr.1: 27406293-27406293 on Build GRCh38
rs370965565 Chr.1: 27406307-27406307 on Build GRCh38
rs572360467 Chr.1: 27406529-27406529 on Build GRCh38
rs999715608 Chr.1: 27406202-27406202 on Build GRCh38
rs111468269 Chr.1: 27406411-27406411 on Build GRCh38
rs907899801 Chr.1: 27406567-27406567 on Build GRCh38
rs970688881 Chr.1: 27406490-27406490 on Build GRCh38
rs1019828658 Chr.1: 27406171-27406171 on Build GRCh38
rs996353710 Chr.1: 27406128-27406128 on Build GRCh38
rs898933657 Chr.1: 27406126-27406126 on Build GRCh38
rs754871087 Chr.1: 27406350-27406350 on Build GRCh38
rs1012292317 Chr.1: 27406470-27406470 on Build GRCh38
rs568753616 Chr.1: 27406510-27406510 on Build GRCh38
rs111388120 Chr.1: 27406407-27406407 on Build GRCh38

View Full Product Details