Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39995095-39995596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMD3
Entrez Gene ID
5709
Gene Name
proteasome 26S subunit, non-ATPase 3
Gene Aliases
P58, RPN3, S3, TSTA2
UniGene
Hs.12970
Gene Chromosome Location
Chr.17: 39980768-39997960 on Build GRCh38
Transcripts
NM_002809.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4796486 c.1275G>A p.K425K Chr.17: 39995482-39995482 on Build GRCh38
5464029 c.1300G>A p.A434T Chr.17: 39995507-39995507 on Build GRCh38
1177928 c.1119C>G p.A373A Chr.17: 39995198-39995198 on Build GRCh38
1222346 c.1130A>G p.Q377R Chr.17: 39995209-39995209 on Build GRCh38
6335639 c.1259C>T p.A420V Chr.17: 39995466-39995466 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778443728 Chr.17: 39995334-39995334 on Build GRCh38
rs772912884 Chr.17: 39995133-39995133 on Build GRCh38
rs772814654 Chr.17: 39995227-39995227 on Build GRCh38
rs749595717 Chr.17: 39995163-39995163 on Build GRCh38
rs377487940 Chr.17: 39995346-39995346 on Build GRCh38
rs556141280 Chr.17: 39995150-39995150 on Build GRCh38
rs774936001 Chr.17: 39995115-39995115 on Build GRCh38
rs144987407 Chr.17: 39995500-39995500 on Build GRCh38
rs182851904 Chr.17: 39995317-39995317 on Build GRCh38
rs766433096 Chr.17: 39995426-39995426 on Build GRCh38
rs759191215 Chr.17: 39995252-39995252 on Build GRCh38
rs186615693 Chr.17: 39995401-39995401 on Build GRCh38
rs757901353 Chr.17: 39995524-39995524 on Build GRCh38
rs775210346 Chr.17: 39995264-39995264 on Build GRCh38
rs753913903 Chr.17: 39995432-39995432 on Build GRCh38
rs373677696 Chr.17: 39995406-39995406 on Build GRCh38
rs761716360 Chr.17: 39995569-39995569 on Build GRCh38
rs948492594 Chr.17: 39995391-39995391 on Build GRCh38
rs561609392 Chr.17: 39995411-39995411 on Build GRCh38
rs779209857 Chr.17: 39995186-39995186 on Build GRCh38
rs746504314 Chr.17: 39995116-39995117 on Build GRCh38
rs1030967556 Chr.17: 39995365-39995365 on Build GRCh38
rs758895520 Chr.17: 39995331-39995331 on Build GRCh38
rs764572779 Chr.17: 39995515-39995515 on Build GRCh38
rs758843411 Chr.17: 39995489-39995489 on Build GRCh38
rs1029105237 Chr.17: 39995409-39995409 on Build GRCh38
rs185207884 Chr.17: 39995139-39995139 on Build GRCh38
rs901933569 Chr.17: 39995375-39995375 on Build GRCh38
rs755192717 Chr.17: 39995454-39995454 on Build GRCh38
rs765079424 Chr.17: 39995255-39995255 on Build GRCh38
rs770571878 Chr.17: 39995382-39995382 on Build GRCh38
rs769448037 Chr.17: 39995551-39995551 on Build GRCh38
rs376005242 Chr.17: 39995560-39995560 on Build GRCh38
rs780830924 Chr.17: 39995534-39995534 on Build GRCh38
rs369145353 Chr.17: 39995168-39995168 on Build GRCh38
rs140004092 Chr.17: 39995118-39995118 on Build GRCh38
rs758337354 Chr.17: 39995155-39995155 on Build GRCh38
rs781320102 Chr.17: 39995158-39995158 on Build GRCh38
rs191334485 Chr.17: 39995160-39995160 on Build GRCh38
rs779310845 Chr.17: 39995320-39995320 on Build GRCh38
rs766165401 Chr.17: 39995140-39995140 on Build GRCh38
rs761159577 Chr.17: 39995117-39995117 on Build GRCh38
rs545920644 Chr.17: 39995354-39995354 on Build GRCh38
rs753054200 Chr.17: 39995473-39995473 on Build GRCh38
rs756209157 Chr.17: 39995282-39995282 on Build GRCh38
rs771605493 Chr.17: 39995120-39995120 on Build GRCh38
rs746544593 Chr.17: 39995238-39995238 on Build GRCh38
rs748497967 Chr.17: 39995326-39995326 on Build GRCh38
rs762927208 Chr.17: 39995268-39995268 on Build GRCh38
rs903116663 Chr.17: 39995359-39995359 on Build GRCh38
rs62621374 Chr.17: 39995242-39995242 on Build GRCh38
rs563907315 Chr.17: 39995364-39995364 on Build GRCh38
rs774188003 Chr.17: 39995562-39995562 on Build GRCh38
rs201530083 Chr.17: 39995342-39995342 on Build GRCh38
rs372144959 Chr.17: 39995153-39995153 on Build GRCh38
rs766510251 Chr.17: 39995337-39995337 on Build GRCh38
rs780286845 Chr.17: 39995162-39995162 on Build GRCh38
rs774275737 Chr.17: 39995419-39995419 on Build GRCh38
rs745410697 Chr.17: 39995547-39995547 on Build GRCh38
rs775325002 Chr.17: 39995407-39995407 on Build GRCh38
rs765405766 Chr.17: 39995465-39995465 on Build GRCh38
rs776439695 Chr.17: 39995246-39995246 on Build GRCh38
rs757076787 Chr.17: 39995145-39995145 on Build GRCh38
rs947276488 Chr.17: 39995340-39995340 on Build GRCh38
rs528124445 Chr.17: 39995370-39995370 on Build GRCh38
rs374192363 Chr.17: 39995206-39995206 on Build GRCh38
rs370223398 Chr.17: 39995403-39995403 on Build GRCh38
rs925617617 Chr.17: 39995471-39995471 on Build GRCh38
rs753643353 Chr.17: 39995142-39995142 on Build GRCh38
rs761804346 Chr.17: 39995424-39995424 on Build GRCh38
rs200909296 Chr.17: 39995312-39995312 on Build GRCh38
rs750462524 Chr.17: 39995275-39995275 on Build GRCh38
rs764061023 Chr.17: 39995270-39995270 on Build GRCh38
rs766521376 Chr.17: 39995283-39995283 on Build GRCh38
rs748674701 Chr.17: 39995187-39995187 on Build GRCh38
rs779596672 Chr.17: 39995556-39995556 on Build GRCh38
rs376686096 Chr.17: 39995230-39995230 on Build GRCh38

View Full Product Details