Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40443565-40444056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGFBP4
Entrez Gene ID
3487
Gene Name
insulin like growth factor binding protein 4
Gene Aliases
BP-4, HT29-IGFBP, IBP4, IGFBP-4
UniGene
Hs.462998
Gene Chromosome Location
Chr.17: 40443424-40457730 on Build GRCh38
Transcripts
NM_001552.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6368538 c.239G>A p.C80Y Chr.17: 40443974-40443974 on Build GRCh38
3749399 c.105G>A p.L35L Chr.17: 40443840-40443840 on Build GRCh38
6409060 c.289C>T p.Q97* Chr.17: 40444024-40444024 on Build GRCh38
6431565 c.195G>T p.M65I Chr.17: 40443930-40443930 on Build GRCh38
4887922 c.242A>C p.Y81S Chr.17: 40443977-40443977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs993671824 Chr.17: 40443962-40443962 on Build GRCh38
rs149707434 Chr.17: 40443898-40443899 on Build GRCh38
rs745589702 Chr.17: 40443922-40443922 on Build GRCh38
rs895494664 Chr.17: 40443990-40443990 on Build GRCh38
rs766820217 Chr.17: 40443850-40443850 on Build GRCh38
rs746721334 Chr.17: 40443777-40443777 on Build GRCh38
rs777397826 Chr.17: 40443774-40443774 on Build GRCh38
rs578013057 Chr.17: 40443853-40443853 on Build GRCh38
rs868039406 Chr.17: 40443992-40443992 on Build GRCh38
rs1000112456 Chr.17: 40444003-40444003 on Build GRCh38
rs1040398845 Chr.17: 40443591-40443591 on Build GRCh38
rs545435670 Chr.17: 40443881-40443881 on Build GRCh38
rs9891003 Chr.17: 40443663-40443663 on Build GRCh38
rs537605722 Chr.17: 40443819-40443819 on Build GRCh38
rs922029248 Chr.17: 40443601-40443601 on Build GRCh38
rs754113478 Chr.17: 40443852-40443852 on Build GRCh38
rs1036726857 Chr.17: 40443691-40443691 on Build GRCh38
rs567448530 Chr.17: 40443687-40443687 on Build GRCh38
rs906829087 Chr.17: 40443698-40443698 on Build GRCh38
rs533817088 Chr.17: 40443628-40443628 on Build GRCh38
rs572455516 Chr.17: 40443843-40443843 on Build GRCh38
rs769638273 Chr.17: 40443963-40443963 on Build GRCh38
rs747551908 Chr.17: 40443730-40443730 on Build GRCh38
rs968653100 Chr.17: 40443722-40443722 on Build GRCh38
rs777313226 Chr.17: 40443857-40443857 on Build GRCh38
rs942237940 Chr.17: 40443891-40443891 on Build GRCh38
rs780842795 Chr.17: 40443904-40443904 on Build GRCh38
rs531134762 Chr.17: 40443900-40443900 on Build GRCh38
rs761735800 Chr.17: 40443816-40443816 on Build GRCh38
rs988133109 Chr.17: 40443845-40443845 on Build GRCh38
rs903983703 Chr.17: 40443978-40443978 on Build GRCh38
rs910722865 Chr.17: 40443868-40443868 on Build GRCh38
rs1056687123 Chr.17: 40443976-40443976 on Build GRCh38
rs1004790569 Chr.17: 40443685-40443685 on Build GRCh38
rs769567327 Chr.17: 40443798-40443798 on Build GRCh38
rs759404034 Chr.17: 40443783-40443783 on Build GRCh38
rs1002546656 Chr.17: 40443706-40443706 on Build GRCh38
rs976069773 Chr.17: 40443760-40443760 on Build GRCh38
rs775514911 Chr.17: 40443801-40443801 on Build GRCh38
rs923268369 Chr.17: 40443851-40443851 on Build GRCh38
rs749309040 Chr.17: 40444007-40444007 on Build GRCh38
rs1011353171 Chr.17: 40443718-40443718 on Build GRCh38
rs940675487 Chr.17: 40443946-40443946 on Build GRCh38
rs925420032 Chr.17: 40443810-40443810 on Build GRCh38
rs1034070017 Chr.17: 40443714-40443714 on Build GRCh38
rs958460873 Chr.17: 40443717-40443717 on Build GRCh38
rs767776913 Chr.17: 40443822-40443822 on Build GRCh38
rs887630809 Chr.17: 40443669-40443676 on Build GRCh38
rs776446714 Chr.17: 40443780-40443780 on Build GRCh38
rs1022446792 Chr.17: 40443719-40443719 on Build GRCh38
rs1021834715 Chr.17: 40443947-40443947 on Build GRCh38
rs777867036 Chr.17: 40444020-40444020 on Build GRCh38
rs599199 Chr.17: 40443860-40443860 on Build GRCh38
rs924193450 Chr.17: 40443743-40443743 on Build GRCh38
rs770686861 Chr.17: 40443778-40443778 on Build GRCh38
rs894172793 Chr.17: 40443917-40443917 on Build GRCh38
rs879762123 Chr.17: 40443906-40443906 on Build GRCh38
rs771613751 Chr.17: 40443759-40443759 on Build GRCh38
rs598892 Chr.17: 40443840-40443840 on Build GRCh38
rs940521007 Chr.17: 40443851-40443852 on Build GRCh38
rs775288523 Chr.17: 40444006-40444006 on Build GRCh38
rs554193402 Chr.17: 40443951-40443951 on Build GRCh38
rs778799414 Chr.17: 40443852-40443852 on Build GRCh38
rs772257832 Chr.17: 40444023-40444023 on Build GRCh38
rs761027365 Chr.17: 40443844-40443844 on Build GRCh38
rs758124086 Chr.17: 40443856-40443856 on Build GRCh38

View Full Product Details