Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40753727-40754224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT25
Entrez Gene ID
147183
Gene Name
keratin 25
Gene Aliases
ARWH3, KRT24IRS1, KRT25A
UniGene
Hs.55412
Gene Chromosome Location
Chr.17: 40748021-40755542 on Build GRCh38
Transcripts
NM_181534.3 XM_011524414.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5386289 c.626C>T p.T209I Chr.17: 40753903-40753903 on Build GRCh38
5924077 c.583G>A p.E195K Chr.17: 40753946-40753946 on Build GRCh38
1710224 c.637G>A p.E213K Chr.17: 40753892-40753892 on Build GRCh38
978936 c.582T>C p.D194D Chr.17: 40753947-40753947 on Build GRCh38
337470 c.622G>A p.E208K Chr.17: 40753907-40753907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755273304 Chr.17: 40753948-40753948 on Build GRCh38
rs758921574 Chr.17: 40753961-40753961 on Build GRCh38
rs56146056 Chr.17: 40754077-40754077 on Build GRCh38
rs781288231 Chr.17: 40753856-40753856 on Build GRCh38
rs769094210 Chr.17: 40753869-40753869 on Build GRCh38
rs1045978170 Chr.17: 40753989-40753989 on Build GRCh38
rs749543783 Chr.17: 40753865-40753865 on Build GRCh38
rs560835445 Chr.17: 40753938-40753938 on Build GRCh38
rs771759459 Chr.17: 40753809-40753809 on Build GRCh38
rs750444068 Chr.17: 40753928-40753928 on Build GRCh38
rs778376225 Chr.17: 40754061-40754061 on Build GRCh38
rs1001910692 Chr.17: 40753937-40753937 on Build GRCh38
rs879893603 Chr.17: 40753934-40753934 on Build GRCh38
rs765130547 Chr.17: 40753903-40753903 on Build GRCh38
rs369593497 Chr.17: 40754023-40754023 on Build GRCh38
rs761818521 Chr.17: 40754075-40754075 on Build GRCh38
rs755288740 Chr.17: 40754027-40754027 on Build GRCh38
rs770423724 Chr.17: 40753895-40753895 on Build GRCh38
rs1048224938 Chr.17: 40754079-40754079 on Build GRCh38
rs966863153 Chr.17: 40753811-40753811 on Build GRCh38
rs267604857 Chr.17: 40753868-40753868 on Build GRCh38
rs1009974763 Chr.17: 40753771-40753771 on Build GRCh38
rs758865621 Chr.17: 40754051-40754051 on Build GRCh38
rs753048825 Chr.17: 40754046-40754046 on Build GRCh38
rs892786578 Chr.17: 40754003-40754003 on Build GRCh38
rs541478715 Chr.17: 40753844-40753844 on Build GRCh38
rs867352715 Chr.17: 40753783-40753783 on Build GRCh38
rs774300352 Chr.17: 40754009-40754009 on Build GRCh38
rs199740972 Chr.17: 40753902-40753902 on Build GRCh38
rs368928911 Chr.17: 40753897-40753897 on Build GRCh38
rs770497772 Chr.17: 40753986-40753986 on Build GRCh38
rs866026516 Chr.17: 40753829-40753829 on Build GRCh38
rs778429418 Chr.17: 40753970-40753970 on Build GRCh38
rs138781615 Chr.17: 40753863-40753863 on Build GRCh38
rs150359949 Chr.17: 40753946-40753946 on Build GRCh38
rs757303994 Chr.17: 40753855-40753855 on Build GRCh38
rs920579512 Chr.17: 40753896-40753896 on Build GRCh38
rs757758848 Chr.17: 40753813-40753814 on Build GRCh38
rs906187464 Chr.17: 40753935-40753935 on Build GRCh38
rs775320636 Chr.17: 40753996-40753996 on Build GRCh38
rs762750245 Chr.17: 40753997-40753997 on Build GRCh38
rs779236179 Chr.17: 40753874-40753874 on Build GRCh38
rs765573191 Chr.17: 40754034-40754034 on Build GRCh38
rs776128576 Chr.17: 40753988-40753988 on Build GRCh38
rs1054815095 Chr.17: 40754195-40754195 on Build GRCh38
rs964936157 Chr.17: 40754120-40754120 on Build GRCh38
rs372235530 Chr.17: 40753877-40753877 on Build GRCh38
rs201621110 Chr.17: 40753891-40753891 on Build GRCh38
rs772919324 Chr.17: 40753888-40753888 on Build GRCh38
rs182168491 Chr.17: 40754103-40754103 on Build GRCh38
rs750639585 Chr.17: 40753857-40753857 on Build GRCh38
rs375239629 Chr.17: 40753825-40753825 on Build GRCh38
rs747601705 Chr.17: 40754066-40754066 on Build GRCh38
rs149543351 Chr.17: 40753990-40753990 on Build GRCh38
rs1046563714 Chr.17: 40754115-40754115 on Build GRCh38
rs565684250 Chr.17: 40754132-40754132 on Build GRCh38
rs1009046683 Chr.17: 40754092-40754092 on Build GRCh38
rs775373708 Chr.17: 40753906-40753906 on Build GRCh38
rs763948567 Chr.17: 40754000-40754000 on Build GRCh38
rs141255550 Chr.17: 40753881-40753881 on Build GRCh38
rs368305279 Chr.17: 40753860-40753860 on Build GRCh38
rs577188477 Chr.17: 40753789-40753789 on Build GRCh38
rs1002256736 Chr.17: 40754127-40754127 on Build GRCh38
rs779466976 Chr.17: 40753895-40753896 on Build GRCh38
rs779490970 Chr.17: 40753951-40753951 on Build GRCh38
rs531531024 Chr.17: 40753973-40753973 on Build GRCh38
rs752654246 Chr.17: 40753846-40753846 on Build GRCh38
rs773205178 Chr.17: 40753836-40753836 on Build GRCh38
rs370817913 Chr.17: 40753824-40753824 on Build GRCh38
rs949852479 Chr.17: 40753968-40753968 on Build GRCh38
rs146293609 Chr.17: 40754012-40754012 on Build GRCh38
rs892860587 Chr.17: 40753757-40753757 on Build GRCh38
rs930670753 Chr.17: 40753918-40753918 on Build GRCh38
rs760474930 Chr.17: 40753818-40753818 on Build GRCh38
rs762943019 Chr.17: 40753921-40753921 on Build GRCh38
rs770792292 Chr.17: 40753925-40753925 on Build GRCh38
rs769470913 Chr.17: 40753993-40753993 on Build GRCh38
rs868597746 Chr.17: 40753994-40753994 on Build GRCh38
rs759379419 Chr.17: 40753833-40753833 on Build GRCh38
rs879133136 Chr.17: 40754064-40754064 on Build GRCh38
rs138077625 Chr.17: 40753955-40753955 on Build GRCh38
rs772970432 Chr.17: 40753816-40753816 on Build GRCh38
rs746534406 Chr.17: 40753892-40753892 on Build GRCh38

View Full Product Details