Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40770120-40770637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT26
Entrez Gene ID
353288
Gene Name
keratin 26
Gene Aliases
CK26, K25, K25IRS2, K26, KRT25B
UniGene
Hs.592133
Gene Chromosome Location
Chr.17: 40766238-40772159 on Build GRCh38
Transcripts
NM_181539.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
978945 c.616G>A p.D206N Chr.17: 40770318-40770318 on Build GRCh38
6080881 c.653T>C p.L218S Chr.17: 40770281-40770281 on Build GRCh38
978946 c.598C>A p.L200M Chr.17: 40770336-40770336 on Build GRCh38
4765716 c.671_672insA p.S224fs*3 Chr.17: 40770262-40770263 on Build GRCh38
4787850 c.682G>T p.E228* Chr.17: 40770122-40770122 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747063783 Chr.17: 40770446-40770446 on Build GRCh38
rs749250027 Chr.17: 40770354-40770354 on Build GRCh38
rs1033104736 Chr.17: 40770486-40770486 on Build GRCh38
rs970174117 Chr.17: 40770208-40770208 on Build GRCh38
rs1025251322 Chr.17: 40770501-40770501 on Build GRCh38
rs757924736 Chr.17: 40770312-40770312 on Build GRCh38
rs371719816 Chr.17: 40770174-40770174 on Build GRCh38
rs763314757 Chr.17: 40770390-40770390 on Build GRCh38
rs745609975 Chr.17: 40770330-40770330 on Build GRCh38
rs751087215 Chr.17: 40770314-40770314 on Build GRCh38
rs769675226 Chr.17: 40770247-40770247 on Build GRCh38
rs143177462 Chr.17: 40770304-40770304 on Build GRCh38
rs75879256 Chr.17: 40770276-40770276 on Build GRCh38
rs773367250 Chr.17: 40770363-40770363 on Build GRCh38
rs1020546107 Chr.17: 40770540-40770540 on Build GRCh38
rs746733455 Chr.17: 40770231-40770231 on Build GRCh38
rs545470723 Chr.17: 40770558-40770558 on Build GRCh38
rs371574424 Chr.17: 40770302-40770302 on Build GRCh38
rs746137542 Chr.17: 40770454-40770454 on Build GRCh38
rs374519965 Chr.17: 40770215-40770215 on Build GRCh38
rs770072203 Chr.17: 40770461-40770461 on Build GRCh38
rs377152917 Chr.17: 40770432-40770432 on Build GRCh38
rs980830703 Chr.17: 40770200-40770200 on Build GRCh38
rs1033886016 Chr.17: 40770588-40770588 on Build GRCh38
rs986780538 Chr.17: 40770575-40770575 on Build GRCh38
rs957562318 Chr.17: 40770488-40770488 on Build GRCh38
rs184026136 Chr.17: 40770176-40770176 on Build GRCh38
rs898367219 Chr.17: 40770420-40770420 on Build GRCh38
rs148011392 Chr.17: 40770185-40770185 on Build GRCh38
rs188928120 Chr.17: 40770482-40770482 on Build GRCh38
rs372053697 Chr.17: 40770431-40770431 on Build GRCh38
rs748233427 Chr.17: 40770258-40770258 on Build GRCh38
rs200496634 Chr.17: 40770445-40770445 on Build GRCh38
rs376174988 Chr.17: 40770147-40770147 on Build GRCh38
rs759949567 Chr.17: 40770370-40770370 on Build GRCh38
rs904934616 Chr.17: 40770466-40770466 on Build GRCh38
rs550339372 Chr.17: 40770378-40770378 on Build GRCh38
rs117795478 Chr.17: 40770369-40770369 on Build GRCh38
rs148303307 Chr.17: 40770397-40770397 on Build GRCh38
rs765693900 Chr.17: 40770149-40770149 on Build GRCh38
rs753186513 Chr.17: 40770308-40770308 on Build GRCh38
rs747090105 Chr.17: 40770365-40770365 on Build GRCh38
rs138751025 Chr.17: 40770318-40770318 on Build GRCh38
rs765589041 Chr.17: 40770372-40770372 on Build GRCh38
rs115869215 Chr.17: 40770609-40770609 on Build GRCh38
rs772177649 Chr.17: 40770360-40770360 on Build GRCh38
rs777452811 Chr.17: 40770222-40770222 on Build GRCh38
rs368218339 Chr.17: 40770269-40770269 on Build GRCh38
rs755931315 Chr.17: 40770429-40770429 on Build GRCh38
rs771176678 Chr.17: 40770450-40770450 on Build GRCh38
rs371160957 Chr.17: 40770171-40770171 on Build GRCh38
rs747638576 Chr.17: 40770444-40770444 on Build GRCh38
rs767130732 Chr.17: 40770403-40770403 on Build GRCh38
rs776054590 Chr.17: 40770381-40770381 on Build GRCh38
rs1038206959 Chr.17: 40770406-40770406 on Build GRCh38
rs532826082 Chr.17: 40770275-40770275 on Build GRCh38
rs111493875 Chr.17: 40770278-40770278 on Build GRCh38
rs1051587212 Chr.17: 40770325-40770325 on Build GRCh38
rs540566287 Chr.17: 40770457-40770457 on Build GRCh38
rs374862542 Chr.17: 40770426-40770426 on Build GRCh38
rs751034030 Chr.17: 40770392-40770392 on Build GRCh38
rs1025972497 Chr.17: 40770465-40770465 on Build GRCh38
rs781562691 Chr.17: 40770453-40770453 on Build GRCh38
rs753679614 Chr.17: 40770434-40770434 on Build GRCh38
rs755909364 Chr.17: 40770348-40770348 on Build GRCh38
rs367734430 Chr.17: 40770238-40770238 on Build GRCh38
rs780850839 Chr.17: 40770319-40770319 on Build GRCh38
rs1038724586 Chr.17: 40770430-40770430 on Build GRCh38
rs554352892 Chr.17: 40770600-40770600 on Build GRCh38
rs960412364 Chr.17: 40770178-40770178 on Build GRCh38
rs775308960 Chr.17: 40770257-40770257 on Build GRCh38
rs989442286 Chr.17: 40770538-40770538 on Build GRCh38
rs896118098 Chr.17: 40770439-40770439 on Build GRCh38
rs144826934 Chr.17: 40770313-40770313 on Build GRCh38
rs781519073 Chr.17: 40770196-40770196 on Build GRCh38
rs764671039 Chr.17: 40770391-40770391 on Build GRCh38
rs780910387 Chr.17: 40770243-40770243 on Build GRCh38
rs200511150 Chr.17: 40770282-40770282 on Build GRCh38
rs911295978 Chr.17: 40770352-40770352 on Build GRCh38
rs568008639 Chr.17: 40770263-40770264 on Build GRCh38
rs140434418 Chr.17: 40770353-40770353 on Build GRCh38
rs561868723 Chr.17: 40770165-40770165 on Build GRCh38
rs772081382 Chr.17: 40770268-40770268 on Build GRCh38
rs970858840 Chr.17: 40770512-40770512 on Build GRCh38
rs370068988 Chr.17: 40770244-40770244 on Build GRCh38
rs994457538 Chr.17: 40770425-40770425 on Build GRCh38

View Full Product Details