Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41396833-41397333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT31
Entrez Gene ID
3881
Gene Name
keratin 31
Gene Aliases
HA1, Ha-1, KRTHA1, hHa1
UniGene
Hs.41696
Gene Chromosome Location
Chr.17: 41393724-41397592 on Build GRCh38
Transcripts
NM_002277.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7318110 c.423C>G p.F141L Chr.17: 41396921-41396921 on Build GRCh38
6146535 c.390G>T p.Q130H Chr.17: 41396954-41396954 on Build GRCh38
4753280 c.269A>C p.E90A Chr.17: 41397271-41397271 on Build GRCh38
3517339 c.423C>T p.F141F Chr.17: 41396921-41396921 on Build GRCh38
3517341 c.242A>T p.N81I Chr.17: 41397298-41397298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051074060 Chr.17: 41396872-41396872 on Build GRCh38
rs375281461 Chr.17: 41397008-41397008 on Build GRCh38
rs775196792 Chr.17: 41397274-41397274 on Build GRCh38
rs757585367 Chr.17: 41397254-41397254 on Build GRCh38
rs773793747 Chr.17: 41396875-41396875 on Build GRCh38
rs1031492559 Chr.17: 41397012-41397012 on Build GRCh38
rs933594108 Chr.17: 41396859-41396859 on Build GRCh38
rs771509354 Chr.17: 41397043-41397043 on Build GRCh38
rs750380476 Chr.17: 41397212-41397212 on Build GRCh38
rs755982013 Chr.17: 41397240-41397240 on Build GRCh38
rs752764312 Chr.17: 41397246-41397246 on Build GRCh38
rs139303550 Chr.17: 41397242-41397242 on Build GRCh38
rs573796512 Chr.17: 41397069-41397069 on Build GRCh38
rs371956622 Chr.17: 41397172-41397172 on Build GRCh38
rs763639152 Chr.17: 41397184-41397184 on Build GRCh38
rs774266070 Chr.17: 41397185-41397185 on Build GRCh38
rs58568023 Chr.17: 41396877-41396877 on Build GRCh38
rs769459099 Chr.17: 41397164-41397164 on Build GRCh38
rs368621857 Chr.17: 41397039-41397039 on Build GRCh38
rs914883194 Chr.17: 41397062-41397062 on Build GRCh38
rs116241763 Chr.17: 41396998-41396998 on Build GRCh38
rs751763235 Chr.17: 41397307-41397307 on Build GRCh38
rs757276506 Chr.17: 41397007-41397007 on Build GRCh38
rs376838934 Chr.17: 41397186-41397186 on Build GRCh38
rs748376848 Chr.17: 41397032-41397032 on Build GRCh38
rs767490156 Chr.17: 41397189-41397189 on Build GRCh38
rs759321355 Chr.17: 41396887-41396887 on Build GRCh38
rs764977684 Chr.17: 41396892-41396892 on Build GRCh38
rs6503627 Chr.17: 41397295-41397295 on Build GRCh38
rs868595625 Chr.17: 41397226-41397226 on Build GRCh38
rs113024681 Chr.17: 41396909-41396909 on Build GRCh38
rs149521729 Chr.17: 41397252-41397252 on Build GRCh38
rs772605005 Chr.17: 41396866-41396866 on Build GRCh38
rs1042184821 Chr.17: 41397104-41397104 on Build GRCh38
rs752862120 Chr.17: 41397297-41397297 on Build GRCh38
rs112541528 Chr.17: 41397149-41397149 on Build GRCh38
rs369861680 Chr.17: 41396863-41396863 on Build GRCh38
rs910833117 Chr.17: 41397174-41397174 on Build GRCh38
rs553792126 Chr.17: 41396951-41396951 on Build GRCh38
rs773384429 Chr.17: 41396935-41396935 on Build GRCh38
rs953817776 Chr.17: 41397110-41397110 on Build GRCh38
rs770365314 Chr.17: 41397268-41397268 on Build GRCh38
rs374329083 Chr.17: 41396899-41396899 on Build GRCh38
rs767348803 Chr.17: 41397018-41397018 on Build GRCh38
rs890189029 Chr.17: 41397109-41397109 on Build GRCh38
rs770156831 Chr.17: 41397157-41397158 on Build GRCh38
rs757750623 Chr.17: 41397135-41397135 on Build GRCh38
rs986410589 Chr.17: 41397134-41397134 on Build GRCh38
rs748743219 Chr.17: 41397275-41397275 on Build GRCh38
rs377012387 Chr.17: 41397030-41397030 on Build GRCh38
rs200375490 Chr.17: 41396931-41396931 on Build GRCh38
rs900418191 Chr.17: 41397251-41397251 on Build GRCh38
rs772935276 Chr.17: 41397294-41397294 on Build GRCh38
rs781656967 Chr.17: 41397264-41397264 on Build GRCh38
rs945127741 Chr.17: 41397065-41397066 on Build GRCh38
rs749352568 Chr.17: 41396915-41396915 on Build GRCh38
rs555244112 Chr.17: 41397049-41397049 on Build GRCh38
rs751545410 Chr.17: 41396902-41396902 on Build GRCh38
rs762679483 Chr.17: 41397178-41397178 on Build GRCh38
rs761263135 Chr.17: 41396938-41396938 on Build GRCh38
rs753954670 Chr.17: 41397022-41397022 on Build GRCh38
rs368630896 Chr.17: 41396929-41396929 on Build GRCh38
rs777381257 Chr.17: 41396901-41396901 on Build GRCh38
rs771882492 Chr.17: 41397287-41397287 on Build GRCh38
rs758495333 Chr.17: 41397034-41397034 on Build GRCh38
rs752052783 Chr.17: 41397035-41397035 on Build GRCh38
rs777257210 Chr.17: 41397141-41397141 on Build GRCh38
rs773850073 Chr.17: 41397281-41397281 on Build GRCh38
rs766304221 Chr.17: 41397296-41397296 on Build GRCh38
rs762653678 Chr.17: 41396996-41396996 on Build GRCh38
rs201366453 Chr.17: 41396969-41396969 on Build GRCh38
rs763354530 Chr.17: 41396972-41396972 on Build GRCh38
rs757074852 Chr.17: 41396906-41396906 on Build GRCh38
rs192604188 Chr.17: 41397024-41397024 on Build GRCh38
rs144128389 Chr.17: 41397292-41397292 on Build GRCh38
rs374693697 Chr.17: 41396959-41396959 on Build GRCh38
rs369687387 Chr.17: 41397305-41397305 on Build GRCh38
rs376345126 Chr.17: 41396884-41396884 on Build GRCh38
rs762921339 Chr.17: 41397304-41397304 on Build GRCh38
rs544113705 Chr.17: 41397126-41397126 on Build GRCh38
rs199832301 Chr.17: 41396961-41396961 on Build GRCh38
rs895771498 Chr.17: 41396932-41396932 on Build GRCh38
rs897936725 Chr.17: 41396905-41396905 on Build GRCh38
rs758717258 Chr.17: 41397249-41397249 on Build GRCh38
rs917430429 Chr.17: 41396966-41396966 on Build GRCh38
rs765922614 Chr.17: 41396879-41396879 on Build GRCh38
rs538647973 Chr.17: 41396945-41396945 on Build GRCh38
rs746329753 Chr.17: 41397163-41397163 on Build GRCh38
rs146229787 Chr.17: 41396930-41396930 on Build GRCh38
rs746379141 Chr.17: 41397267-41397267 on Build GRCh38
rs768391068 Chr.17: 41397276-41397276 on Build GRCh38
rs748175418 Chr.17: 41396921-41396921 on Build GRCh38
rs751255702 Chr.17: 41397000-41397000 on Build GRCh38
rs761475573 Chr.17: 41397282-41397282 on Build GRCh38
rs201840528 Chr.17: 41396874-41396874 on Build GRCh38
rs1039276522 Chr.17: 41397168-41397168 on Build GRCh38
rs752581165 Chr.17: 41396894-41396894 on Build GRCh38

View Full Product Details