Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 63533915-63534430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB7
Entrez Gene ID
84455
Gene Name
EF-hand calcium binding domain 7
Gene Aliases
-
UniGene
Hs.652324
Gene Chromosome Location
Chr.1: 63523301-63585400 on Build GRCh38
Transcripts
NM_032437.3 XM_005271274.3 XM_006710976.3 XM_006710977.1 XM_011542301.2 XM_017002548.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1687821 c.686A>G p.D229G Chr.1: 63534098-63534098 on Build GRCh38
1126963 c.742A>C p.T248P Chr.1: 63534154-63534154 on Build GRCh38
3747047 c.804+4T>C p.? Chr.1: 63534220-63534220 on Build GRCh38
1580289 c.797T>G p.L266* Chr.1: 63534209-63534209 on Build GRCh38
6902775 c.698C>T p.S233F Chr.1: 63534110-63534110 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113295948 Chr.1: 63534238-63534238 on Build GRCh38
rs770388027 Chr.1: 63534401-63534401 on Build GRCh38
rs759480026 Chr.1: 63534151-63534151 on Build GRCh38
rs775916978 Chr.1: 63534020-63534020 on Build GRCh38
rs371596905 Chr.1: 63534250-63534250 on Build GRCh38
rs201240300 Chr.1: 63534198-63534198 on Build GRCh38
rs754525455 Chr.1: 63534064-63534064 on Build GRCh38
rs914856970 Chr.1: 63534060-63534060 on Build GRCh38
rs924037202 Chr.1: 63534060-63534062 on Build GRCh38
rs956669227 Chr.1: 63534149-63534149 on Build GRCh38
rs1048952830 Chr.1: 63534241-63534241 on Build GRCh38
rs971518536 Chr.1: 63534338-63534338 on Build GRCh38
rs756105282 Chr.1: 63534054-63534054 on Build GRCh38
rs552585923 Chr.1: 63534218-63534218 on Build GRCh38
rs6657559 Chr.1: 63534220-63534220 on Build GRCh38
rs777200241 Chr.1: 63534140-63534140 on Build GRCh38
rs753835859 Chr.1: 63534210-63534210 on Build GRCh38
rs956467882 Chr.1: 63534016-63534016 on Build GRCh38
rs753441788 Chr.1: 63534237-63534238 on Build GRCh38
rs371089600 Chr.1: 63534092-63534092 on Build GRCh38
rs764466555 Chr.1: 63534181-63534181 on Build GRCh38
rs773542489 Chr.1: 63534083-63534083 on Build GRCh38
rs758033961 Chr.1: 63534219-63534219 on Build GRCh38
rs1040572600 Chr.1: 63534215-63534215 on Build GRCh38
rs778381135 Chr.1: 63534067-63534067 on Build GRCh38
rs982635049 Chr.1: 63534018-63534018 on Build GRCh38
rs763642776 Chr.1: 63534123-63534124 on Build GRCh38
rs6693255 Chr.1: 63534155-63534155 on Build GRCh38
rs6657480 Chr.1: 63534197-63534197 on Build GRCh38
rs993759901 Chr.1: 63534314-63534314 on Build GRCh38
rs778453393 Chr.1: 63534217-63534217 on Build GRCh38
rs754405749 Chr.1: 63534211-63534211 on Build GRCh38
rs1048081953 Chr.1: 63534352-63534352 on Build GRCh38
rs796417790 Chr.1: 63534006-63534006 on Build GRCh38
rs763239465 Chr.1: 63534172-63534172 on Build GRCh38
rs189834762 Chr.1: 63533961-63533961 on Build GRCh38
rs924809562 Chr.1: 63534004-63534004 on Build GRCh38
rs887718312 Chr.1: 63534278-63534278 on Build GRCh38
rs771341590 Chr.1: 63534249-63534249 on Build GRCh38
rs748675855 Chr.1: 63534057-63534057 on Build GRCh38
rs747864974 Chr.1: 63534071-63534071 on Build GRCh38
rs750286822 Chr.1: 63534046-63534046 on Build GRCh38
rs568189252 Chr.1: 63534363-63534363 on Build GRCh38
rs147915416 Chr.1: 63534153-63534153 on Build GRCh38
rs561954595 Chr.1: 63533998-63533998 on Build GRCh38
rs771751478 Chr.1: 63534079-63534079 on Build GRCh38
rs771424761 Chr.1: 63534113-63534113 on Build GRCh38
rs539929798 Chr.1: 63533971-63533971 on Build GRCh38
rs550724524 Chr.1: 63534379-63534379 on Build GRCh38
rs750236107 Chr.1: 63534186-63534186 on Build GRCh38
rs185346195 Chr.1: 63534284-63534284 on Build GRCh38
rs150584427 Chr.1: 63534372-63534372 on Build GRCh38
rs1022618018 Chr.1: 63534308-63534308 on Build GRCh38
rs756893851 Chr.1: 63534266-63534271 on Build GRCh38
rs780102569 Chr.1: 63534056-63534056 on Build GRCh38
rs547211679 Chr.1: 63534203-63534203 on Build GRCh38
rs1040744863 Chr.1: 63534282-63534282 on Build GRCh38

View Full Product Details