Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 64744942-64745441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAVER2
Entrez Gene ID
55225
Gene Name
ribonucleoprotein, PTB binding 2
Gene Aliases
-
UniGene
Hs.591443
Gene Chromosome Location
Chr.1: 64744979-64833232 on Build GRCh38
Transcripts
NM_018211.3 XM_006710738.3 XM_011541706.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3805767 c.115G>A p.E39K Chr.1: 64745287-64745287 on Build GRCh38
5832659 c.115G>A p.E39K Chr.1: 64745287-64745287 on Build GRCh38
6393178 c.178C>T p.R60W Chr.1: 64745350-64745350 on Build GRCh38
6560907 c.195G>A p.L65L Chr.1: 64745367-64745367 on Build GRCh38
6560906 c.195G>A p.L65L Chr.1: 64745367-64745367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047076934 Chr.1: 64744973-64744973 on Build GRCh38
rs551040775 Chr.1: 64745080-64745080 on Build GRCh38
rs554479993 Chr.1: 64745376-64745376 on Build GRCh38
rs969586178 Chr.1: 64745015-64745015 on Build GRCh38
rs538547077 Chr.1: 64745139-64745139 on Build GRCh38
rs1040250563 Chr.1: 64745117-64745117 on Build GRCh38
rs1038485372 Chr.1: 64745155-64745155 on Build GRCh38
rs983428846 Chr.1: 64745245-64745245 on Build GRCh38
rs757296290 Chr.1: 64745362-64745362 on Build GRCh38
rs764073733 Chr.1: 64745295-64745295 on Build GRCh38
rs916422742 Chr.1: 64745209-64745210 on Build GRCh38
rs975860876 Chr.1: 64745207-64745207 on Build GRCh38
rs896658379 Chr.1: 64745353-64745353 on Build GRCh38
rs550163082 Chr.1: 64745156-64745156 on Build GRCh38
rs903497998 Chr.1: 64745385-64745385 on Build GRCh38
rs536402716 Chr.1: 64745368-64745368 on Build GRCh38
rs867489141 Chr.1: 64745206-64745206 on Build GRCh38
rs968168303 Chr.1: 64745097-64745097 on Build GRCh38
rs960341172 Chr.1: 64745041-64745041 on Build GRCh38
rs923168964 Chr.1: 64745119-64745119 on Build GRCh38
rs572495180 Chr.1: 64745317-64745317 on Build GRCh38
rs751407200 Chr.1: 64745340-64745340 on Build GRCh38
rs938715666 Chr.1: 64745408-64745408 on Build GRCh38
rs887163615 Chr.1: 64744985-64744985 on Build GRCh38
rs929176912 Chr.1: 64745232-64745237 on Build GRCh38
rs1005644057 Chr.1: 64744986-64744986 on Build GRCh38
rs1002394577 Chr.1: 64745159-64745159 on Build GRCh38
rs967778698 Chr.1: 64745199-64745199 on Build GRCh38
rs1017482774 Chr.1: 64744997-64744997 on Build GRCh38
rs527873396 Chr.1: 64744961-64744961 on Build GRCh38
rs908533269 Chr.1: 64745265-64745265 on Build GRCh38
rs1021000472 Chr.1: 64745186-64745186 on Build GRCh38
rs538692930 Chr.1: 64745286-64745286 on Build GRCh38
rs993928032 Chr.1: 64745242-64745242 on Build GRCh38
rs979207667 Chr.1: 64745101-64745101 on Build GRCh38
rs964394564 Chr.1: 64745420-64745420 on Build GRCh38
rs1002446803 Chr.1: 64745036-64745036 on Build GRCh38
rs1035298376 Chr.1: 64745038-64745038 on Build GRCh38
rs758287229 Chr.1: 64745309-64745310 on Build GRCh38
rs1006050013 Chr.1: 64745148-64745148 on Build GRCh38
rs745385083 Chr.1: 64745363-64745363 on Build GRCh38
rs929254407 Chr.1: 64745122-64745122 on Build GRCh38
rs979578099 Chr.1: 64745205-64745205 on Build GRCh38
rs549355701 Chr.1: 64745087-64745087 on Build GRCh38
rs897843057 Chr.1: 64744983-64744983 on Build GRCh38
rs960918085 Chr.1: 64745174-64745175 on Build GRCh38
rs910382943 Chr.1: 64745104-64745104 on Build GRCh38
rs149969748 Chr.1: 64745289-64745289 on Build GRCh38
rs571688521 Chr.1: 64745269-64745269 on Build GRCh38
rs905252898 Chr.1: 64745294-64745294 on Build GRCh38
rs1035203244 Chr.1: 64745169-64745169 on Build GRCh38
rs567910619 Chr.1: 64745107-64745107 on Build GRCh38
rs1021241429 Chr.1: 64745379-64745379 on Build GRCh38
rs755815756 Chr.1: 64745397-64745397 on Build GRCh38
rs887119954 Chr.1: 64745140-64745140 on Build GRCh38
rs1056284394 Chr.1: 64745176-64745177 on Build GRCh38
rs374428681 Chr.1: 64745336-64745336 on Build GRCh38
rs938088362 Chr.1: 64745328-64745328 on Build GRCh38
rs1056610845 Chr.1: 64745332-64745332 on Build GRCh38

View Full Product Details