Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133066440-133066951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEL
Entrez Gene ID
1056
Gene Name
carboxyl ester lipase
Gene Aliases
BAL, BSDL, BSSL, CELL, CEase, FAP, FAPP, LIPA, MODY8
UniGene
Hs.533258
Gene Chromosome Location
Chr.9: 133061978-133071863 on Build GRCh38
Transcripts
NM_001807.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4829729 c.730G>A p.G244S Chr.9: 133066889-133066889 on Build GRCh38
7326215 c.678+5C>T p.? Chr.9: 133066665-133066665 on Build GRCh38
3432959 c.709C>T p.R237W Chr.9: 133066868-133066868 on Build GRCh38
5584771 c.614G>A p.G205E Chr.9: 133066596-133066596 on Build GRCh38
6550566 c.717C>T p.A239A Chr.9: 133066876-133066876 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1002291875 Chr.9: 133066856-133066856 on Build GRCh38
rs371303105 Chr.9: 133066834-133066834 on Build GRCh38
rs773306384 Chr.9: 133066514-133066514 on Build GRCh38
rs746040301 Chr.9: 133066868-133066868 on Build GRCh38
rs368024872 Chr.9: 133066825-133066825 on Build GRCh38
rs767057872 Chr.9: 133066792-133066792 on Build GRCh38
rs758530414 Chr.9: 133066675-133066675 on Build GRCh38
rs375595842 Chr.9: 133066588-133066588 on Build GRCh38
rs202216787 Chr.9: 133066648-133066648 on Build GRCh38
rs750120148 Chr.9: 133066657-133066657 on Build GRCh38
rs760031366 Chr.9: 133066818-133066818 on Build GRCh38
rs770547110 Chr.9: 133066891-133066891 on Build GRCh38
rs1032474348 Chr.9: 133066730-133066730 on Build GRCh38
rs549454090 Chr.9: 133066812-133066812 on Build GRCh38
rs759227357 Chr.9: 133066896-133066896 on Build GRCh38
rs1006641512 Chr.9: 133066519-133066519 on Build GRCh38
rs774507925 Chr.9: 133066507-133066507 on Build GRCh38
rs562193996 Chr.9: 133066594-133066594 on Build GRCh38
rs976359379 Chr.9: 133066752-133066752 on Build GRCh38
rs752352827 Chr.9: 133066788-133066788 on Build GRCh38
rs776449870 Chr.9: 133066892-133066892 on Build GRCh38
rs777677286 Chr.9: 133066679-133066679 on Build GRCh38
rs373701882 Chr.9: 133066536-133066536 on Build GRCh38
rs769282739 Chr.9: 133066595-133066595 on Build GRCh38
rs757291997 Chr.9: 133066862-133066862 on Build GRCh38
rs897814738 Chr.9: 133066710-133066710 on Build GRCh38
rs535604397 Chr.9: 133066833-133066833 on Build GRCh38
rs768469729 Chr.9: 133066869-133066869 on Build GRCh38
rs747265135 Chr.9: 133066694-133066694 on Build GRCh38
rs570015280 Chr.9: 133066742-133066742 on Build GRCh38
rs771114262 Chr.9: 133066695-133066695 on Build GRCh38
rs755657556 Chr.9: 133066663-133066663 on Build GRCh38
rs917146112 Chr.9: 133066806-133066806 on Build GRCh38
rs768416841 Chr.9: 133066701-133066701 on Build GRCh38
rs749117929 Chr.9: 133066698-133066698 on Build GRCh38
rs754393686 Chr.9: 133066594-133066595 on Build GRCh38
rs796864014 Chr.9: 133066893-133066893 on Build GRCh38
rs748585720 Chr.9: 133066673-133066673 on Build GRCh38
rs763369568 Chr.9: 133066917-133066917 on Build GRCh38
rs1014886783 Chr.9: 133066525-133066525 on Build GRCh38
rs374965842 Chr.9: 133066649-133066649 on Build GRCh38
rs543378322 Chr.9: 133066618-133066618 on Build GRCh38
rs779550536 Chr.9: 133066601-133066601 on Build GRCh38
rs559586351 Chr.9: 133066715-133066715 on Build GRCh38
rs764816823 Chr.9: 133066782-133066782 on Build GRCh38
rs8193020 Chr.9: 133066817-133066817 on Build GRCh38
rs200316440 Chr.9: 133066501-133066501 on Build GRCh38
rs200762866 Chr.9: 133066515-133066515 on Build GRCh38
rs141665160 Chr.9: 133066545-133066545 on Build GRCh38
rs755484675 Chr.9: 133066600-133066600 on Build GRCh38
rs370943954 Chr.9: 133066585-133066585 on Build GRCh38
rs764921162 Chr.9: 133066523-133066523 on Build GRCh38
rs753057752 Chr.9: 133066908-133066908 on Build GRCh38
rs764081605 Chr.9: 133066625-133066625 on Build GRCh38
rs765651548 Chr.9: 133066820-133066820 on Build GRCh38
rs537186649 Chr.9: 133066796-133066796 on Build GRCh38
rs779607545 Chr.9: 133066668-133066668 on Build GRCh38
rs766886891 Chr.9: 133066651-133066651 on Build GRCh38
rs529444912 Chr.9: 133066621-133066621 on Build GRCh38
rs202014196 Chr.9: 133066867-133066867 on Build GRCh38
rs377542125 Chr.9: 133066574-133066574 on Build GRCh38
rs762719603 Chr.9: 133066624-133066624 on Build GRCh38
rs751652936 Chr.9: 133066846-133066846 on Build GRCh38
rs199615228 Chr.9: 133066564-133066564 on Build GRCh38
rs770022400 Chr.9: 133066490-133066490 on Build GRCh38
rs747093170 Chr.9: 133066544-133066544 on Build GRCh38
rs753687010 Chr.9: 133066708-133066708 on Build GRCh38
rs939948877 Chr.9: 133066700-133066700 on Build GRCh38
rs529070571 Chr.9: 133066717-133066717 on Build GRCh38
rs1009815639 Chr.9: 133066741-133066741 on Build GRCh38
rs1025174725 Chr.9: 133066556-133066556 on Build GRCh38
rs757349759 Chr.9: 133066922-133066922 on Build GRCh38
rs372624029 Chr.9: 133066885-133066885 on Build GRCh38
rs533175753 Chr.9: 133066716-133066716 on Build GRCh38
rs773535763 Chr.9: 133066612-133066612 on Build GRCh38
rs778611627 Chr.9: 133066871-133066871 on Build GRCh38
rs762139361 Chr.9: 133066510-133066510 on Build GRCh38
rs749819142 Chr.9: 133066597-133066597 on Build GRCh38
rs953756858 Chr.9: 133066767-133066767 on Build GRCh38
rs372564944 Chr.9: 133066587-133066587 on Build GRCh38
rs888340815 Chr.9: 133066508-133066508 on Build GRCh38
rs772018012 Chr.9: 133066604-133066604 on Build GRCh38
rs752081040 Chr.9: 133066921-133066921 on Build GRCh38
rs1036803694 Chr.9: 133066705-133066705 on Build GRCh38
rs368907057 Chr.9: 133066877-133066877 on Build GRCh38
rs776824509 Chr.9: 133066697-133066697 on Build GRCh38
rs541805406 Chr.9: 133066470-133066470 on Build GRCh38
rs921019791 Chr.9: 133066764-133066764 on Build GRCh38
rs749761906 Chr.9: 133066493-133066493 on Build GRCh38
rs943825859 Chr.9: 133066620-133066620 on Build GRCh38
rs747153235 Chr.9: 133066617-133066617 on Build GRCh38
rs780658344 Chr.9: 133066492-133066492 on Build GRCh38
rs958573610 Chr.9: 133066740-133066740 on Build GRCh38
rs764545406 Chr.9: 133066919-133066919 on Build GRCh38
rs547695985 Chr.9: 133066665-133066665 on Build GRCh38
rs757967503 Chr.9: 133066805-133066805 on Build GRCh38
rs150672381 Chr.9: 133066722-133066722 on Build GRCh38
rs765704794 Chr.9: 133066900-133066900 on Build GRCh38
rs774294234 Chr.9: 133066711-133066711 on Build GRCh38
rs755390448 Chr.9: 133066667-133066667 on Build GRCh38
rs754312279 Chr.9: 133066666-133066666 on Build GRCh38
rs760144206 Chr.9: 133066889-133066889 on Build GRCh38
rs773198000 Chr.9: 133066888-133066888 on Build GRCh38
rs899669409 Chr.9: 133066485-133066485 on Build GRCh38
rs748131218 Chr.9: 133066872-133066872 on Build GRCh38
rs772396574 Chr.9: 133066512-133066512 on Build GRCh38

View Full Product Details