Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 61759918-61760438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf107
Entrez Gene ID
219621
Gene Name
chromosome 10 open reading frame 107
Gene Aliases
bA63A2.1
UniGene
Hs.673160
Gene Chromosome Location
Chr.10: 61662927-61766766 on Build GRCh38
Transcripts
NM_173554.2 XM_005269598.3 XM_005269599.4 XM_005269600.4 XM_006717690.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6013889 c.289-2A>C p.? Chr.10: 61760057-61760057 on Build GRCh38
427743 c.379G>C p.E127Q Chr.10: 61760149-61760149 on Build GRCh38
6066705 c.340G>A p.E114K Chr.10: 61760110-61760110 on Build GRCh38
6586066 c.306C>A p.V102V Chr.10: 61760076-61760076 on Build GRCh38
1283699 c.387C>A p.P129P Chr.10: 61760157-61760157 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758325979 Chr.10: 61760108-61760108 on Build GRCh38
rs192725810 Chr.10: 61760354-61760354 on Build GRCh38
rs764169830 Chr.10: 61760098-61760098 on Build GRCh38
rs187886388 Chr.10: 61760172-61760172 on Build GRCh38
rs149752966 Chr.10: 61760249-61760249 on Build GRCh38
rs188117497 Chr.10: 61760346-61760346 on Build GRCh38
rs907022325 Chr.10: 61760000-61760000 on Build GRCh38
rs776916333 Chr.10: 61759971-61759971 on Build GRCh38
rs570350422 Chr.10: 61760109-61760109 on Build GRCh38
rs769194811 Chr.10: 61760070-61760070 on Build GRCh38
rs770592192 Chr.10: 61760229-61760229 on Build GRCh38
rs777421090 Chr.10: 61760135-61760135 on Build GRCh38
rs998560872 Chr.10: 61760127-61760127 on Build GRCh38
rs753975040 Chr.10: 61760100-61760100 on Build GRCh38
rs769629521 Chr.10: 61760132-61760132 on Build GRCh38
rs767726379 Chr.10: 61760090-61760090 on Build GRCh38
rs16916605 Chr.10: 61759966-61759966 on Build GRCh38
rs369576912 Chr.10: 61760343-61760343 on Build GRCh38
rs370434652 Chr.10: 61760067-61760067 on Build GRCh38
rs746846406 Chr.10: 61760046-61760046 on Build GRCh38
rs928704398 Chr.10: 61760276-61760276 on Build GRCh38
rs137977526 Chr.10: 61760059-61760059 on Build GRCh38
rs377300945 Chr.10: 61760358-61760358 on Build GRCh38
rs780830270 Chr.10: 61760126-61760126 on Build GRCh38
rs752397070 Chr.10: 61760311-61760311 on Build GRCh38
rs912067668 Chr.10: 61760066-61760066 on Build GRCh38
rs920205493 Chr.10: 61760237-61760237 on Build GRCh38
rs370103152 Chr.10: 61760008-61760008 on Build GRCh38
rs767061850 Chr.10: 61760074-61760074 on Build GRCh38
rs528131944 Chr.10: 61760338-61760338 on Build GRCh38
rs1007310321 Chr.10: 61760392-61760392 on Build GRCh38
rs1047579856 Chr.10: 61760002-61760002 on Build GRCh38
rs374367643 Chr.10: 61760010-61760010 on Build GRCh38
rs756167463 Chr.10: 61760021-61760021 on Build GRCh38
rs763544681 Chr.10: 61760071-61760071 on Build GRCh38
rs545931904 Chr.10: 61760278-61760278 on Build GRCh38
rs751345566 Chr.10: 61760117-61760117 on Build GRCh38
rs748850180 Chr.10: 61760148-61760148 on Build GRCh38
rs772324361 Chr.10: 61760159-61760159 on Build GRCh38
rs374104809 Chr.10: 61760086-61760086 on Build GRCh38
rs147002438 Chr.10: 61760177-61760177 on Build GRCh38
rs537352428 Chr.10: 61760399-61760399 on Build GRCh38
rs61850527 Chr.10: 61760337-61760337 on Build GRCh38
rs1022508432 Chr.10: 61760031-61760031 on Build GRCh38
rs1042940698 Chr.10: 61760345-61760345 on Build GRCh38
rs986384663 Chr.10: 61760064-61760064 on Build GRCh38
rs372004989 Chr.10: 61760157-61760157 on Build GRCh38
rs779402051 Chr.10: 61760356-61760356 on Build GRCh38
rs756148887 Chr.10: 61760095-61760095 on Build GRCh38
rs1055514285 Chr.10: 61760323-61760323 on Build GRCh38
rs974337652 Chr.10: 61760235-61760235 on Build GRCh38
rs368297777 Chr.10: 61760087-61760087 on Build GRCh38
rs185083687 Chr.10: 61760371-61760371 on Build GRCh38
rs189045279 Chr.10: 61760379-61760379 on Build GRCh38
rs894331816 Chr.10: 61760325-61760325 on Build GRCh38
rs748762315 Chr.10: 61760215-61760215 on Build GRCh38
rs35286784 Chr.10: 61760402-61760403 on Build GRCh38
rs779406882 Chr.10: 61760231-61760231 on Build GRCh38
rs143081316 Chr.10: 61760152-61760152 on Build GRCh38
rs61850526 Chr.10: 61760248-61760248 on Build GRCh38
rs184427616 Chr.10: 61760306-61760306 on Build GRCh38
rs201800789 Chr.10: 61760029-61760029 on Build GRCh38
rs751458057 Chr.10: 61760184-61760184 on Build GRCh38
rs367600918 Chr.10: 61760288-61760288 on Build GRCh38
rs754746037 Chr.10: 61760191-61760191 on Build GRCh38
rs202052143 Chr.10: 61760197-61760197 on Build GRCh38
rs778777559 Chr.10: 61760025-61760025 on Build GRCh38
rs762848424 Chr.10: 61760123-61760123 on Build GRCh38
rs138612718 Chr.10: 61760154-61760154 on Build GRCh38
rs78603477 Chr.10: 61760138-61760138 on Build GRCh38
rs200962387 Chr.10: 61760055-61760055 on Build GRCh38
rs1018426808 Chr.10: 61760394-61760394 on Build GRCh38
rs761929336 Chr.10: 61760178-61760178 on Build GRCh38
rs371259865 Chr.10: 61760182-61760182 on Build GRCh38
rs191187250 Chr.10: 61760204-61760204 on Build GRCh38
rs376342452 Chr.10: 61760042-61760042 on Build GRCh38
rs1046011729 Chr.10: 61759990-61759990 on Build GRCh38
rs750985647 Chr.10: 61760186-61760186 on Build GRCh38
rs998921758 Chr.10: 61760362-61760362 on Build GRCh38
rs142705866 Chr.10: 61760171-61760171 on Build GRCh38
rs773039110 Chr.10: 61760024-61760024 on Build GRCh38
rs752606593 Chr.10: 61760194-61760194 on Build GRCh38
rs752802888 Chr.10: 61760013-61760013 on Build GRCh38
rs779329254 Chr.10: 61760032-61760039 on Build GRCh38
rs200559290 Chr.10: 61760193-61760193 on Build GRCh38
rs758129909 Chr.10: 61760041-61760041 on Build GRCh38
rs570110721 Chr.10: 61760393-61760393 on Build GRCh38
rs77805435 Chr.10: 61760006-61760006 on Build GRCh38
rs149071916 Chr.10: 61760093-61760093 on Build GRCh38

View Full Product Details