Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 63167044-63167591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JMJD1C
Entrez Gene ID
221037
Gene Name
jumonji domain containing 1C
Gene Aliases
TRIP-8, TRIP8
UniGene
Hs.413416
Gene Chromosome Location
Chr.10: 63167221-63522075 on Build GRCh38
Transcripts
NM_001282948.1 NM_001318153.1 NM_001318154.1 NM_001322252.1 NM_001322254.1 NM_001322258.1 NM_032776.2 NR_134512.1 XM_011539503.1 XM_017015897.1 XM_017015898.1 XM_017015899.1 XM_017015900.1 XM_017015901.1 XM_017015902.1 XM_017015903.1

SNP Detail

SNP ID Chromosome Location
rs765753872 Chr.10: 63167177-63167177 on Build GRCh38
rs978070662 Chr.10: 63167116-63167116 on Build GRCh38
rs957863448 Chr.10: 63167286-63167286 on Build GRCh38
rs758108475 Chr.10: 63167532-63167533 on Build GRCh38
rs763342186 Chr.10: 63167389-63167389 on Build GRCh38
rs780028068 Chr.10: 63167135-63167135 on Build GRCh38
rs376504085 Chr.10: 63167106-63167106 on Build GRCh38
rs572291445 Chr.10: 63167155-63167155 on Build GRCh38
rs1050813770 Chr.10: 63167145-63167145 on Build GRCh38
rs981855328 Chr.10: 63167343-63167343 on Build GRCh38
rs919625300 Chr.10: 63167463-63167463 on Build GRCh38
rs920410683 Chr.10: 63167510-63167510 on Build GRCh38
rs924836947 Chr.10: 63167349-63167349 on Build GRCh38
rs146409156 Chr.10: 63167219-63167219 on Build GRCh38
rs1047402385 Chr.10: 63167183-63167183 on Build GRCh38
rs750810530 Chr.10: 63167370-63167370 on Build GRCh38
rs375612489 Chr.10: 63167161-63167161 on Build GRCh38
rs930571710 Chr.10: 63167526-63167526 on Build GRCh38
rs1055086362 Chr.10: 63167070-63167070 on Build GRCh38
rs201673150 Chr.10: 63167524-63167530 on Build GRCh38
rs546113627 Chr.10: 63167515-63167515 on Build GRCh38
rs554545745 Chr.10: 63167203-63167203 on Build GRCh38
rs780701316 Chr.10: 63167236-63167236 on Build GRCh38
rs558800165 Chr.10: 63167099-63167099 on Build GRCh38
rs766922948 Chr.10: 63167446-63167446 on Build GRCh38
rs886971069 Chr.10: 63167199-63167200 on Build GRCh38
rs1047619631 Chr.10: 63167557-63167557 on Build GRCh38
rs989049681 Chr.10: 63167291-63167291 on Build GRCh38
rs987560533 Chr.10: 63167377-63167382 on Build GRCh38
rs867531139 Chr.10: 63167149-63167149 on Build GRCh38
rs924822295 Chr.10: 63167127-63167127 on Build GRCh38
rs140824389 Chr.10: 63167392-63167392 on Build GRCh38
rs760378705 Chr.10: 63167163-63167163 on Build GRCh38
rs912319233 Chr.10: 63167379-63167380 on Build GRCh38
rs78033346 Chr.10: 63167067-63167067 on Build GRCh38
rs1048204607 Chr.10: 63167150-63167150 on Build GRCh38
rs76251733 Chr.10: 63167432-63167432 on Build GRCh38
rs1035797814 Chr.10: 63167220-63167220 on Build GRCh38
rs914812256 Chr.10: 63167096-63167096 on Build GRCh38
rs958741588 Chr.10: 63167233-63167234 on Build GRCh38
rs933476400 Chr.10: 63167136-63167137 on Build GRCh38
rs796664232 Chr.10: 63167107-63167107 on Build GRCh38
rs996746032 Chr.10: 63167181-63167181 on Build GRCh38
rs942362420 Chr.10: 63167160-63167160 on Build GRCh38
rs563700359 Chr.10: 63167318-63167318 on Build GRCh38
rs772579942 Chr.10: 63167096-63167097 on Build GRCh38
rs543525486 Chr.10: 63167230-63167230 on Build GRCh38
rs1020597227 Chr.10: 63167316-63167316 on Build GRCh38
rs954892098 Chr.10: 63167373-63167373 on Build GRCh38
rs1013341400 Chr.10: 63167245-63167245 on Build GRCh38
rs927712970 Chr.10: 63167353-63167353 on Build GRCh38

View Full Product Details