Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 7063819-7064317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBL1Y
Entrez Gene ID
90665
Gene Name
transducin (beta)-like 1, Y-linked
Gene Aliases
TBL1
UniGene
Hs.664560
Gene Chromosome Location
Chr.Y: 6910686-7107154 on Build GRCh38
Transcripts
NM_033284.1 NM_134258.1 NM_134259.1 XM_005262572.3 XM_017030086.1 XM_017030087.1

SNP Detail

SNP ID Chromosome Location
rs368158489 Chr.Y: 7064128-7064128 on Build GRCh38
rs780714582 Chr.Y: 7063868-7063868 on Build GRCh38
rs764016891 Chr.Y: 7063993-7063993 on Build GRCh38
rs1037738655 Chr.Y: 7064289-7064289 on Build GRCh38
rs774740139 Chr.Y: 7063883-7063883 on Build GRCh38
rs776930645 Chr.Y: 7064071-7064071 on Build GRCh38
rs779395596 Chr.Y: 7064046-7064046 on Build GRCh38
rs751759351 Chr.Y: 7063849-7063849 on Build GRCh38
rs948597265 Chr.Y: 7063900-7063900 on Build GRCh38
rs764105766 Chr.Y: 7064142-7064142 on Build GRCh38
rs770457655 Chr.Y: 7063939-7063939 on Build GRCh38
rs768927205 Chr.Y: 7063875-7063875 on Build GRCh38
rs776986764 Chr.Y: 7063991-7063991 on Build GRCh38
rs796276338 Chr.Y: 7063966-7063966 on Build GRCh38
rs779737497 Chr.Y: 7064110-7064110 on Build GRCh38
rs766772311 Chr.Y: 7064000-7064000 on Build GRCh38
rs779272811 Chr.Y: 7063929-7063929 on Build GRCh38
rs749759552 Chr.Y: 7063872-7063872 on Build GRCh38
rs778229541 Chr.Y: 7064027-7064027 on Build GRCh38
rs777753133 Chr.Y: 7064188-7064188 on Build GRCh38
rs776184505 Chr.Y: 7064171-7064171 on Build GRCh38
rs200332530 Chr.Y: 7063916-7063916 on Build GRCh38
rs377463272 Chr.Y: 7064063-7064063 on Build GRCh38
rs928387697 Chr.Y: 7063914-7063914 on Build GRCh38
rs867351290 Chr.Y: 7064033-7064033 on Build GRCh38
rs765816816 Chr.Y: 7063901-7063901 on Build GRCh38
rs373532788 Chr.Y: 7064122-7064122 on Build GRCh38
rs751822049 Chr.Y: 7064124-7064124 on Build GRCh38
rs768212672 Chr.Y: 7064118-7064118 on Build GRCh38
rs750701720 Chr.Y: 7063953-7063953 on Build GRCh38
rs775236800 Chr.Y: 7063941-7063941 on Build GRCh38
rs780826658 Chr.Y: 7064036-7064036 on Build GRCh38
rs768221778 Chr.Y: 7063965-7063965 on Build GRCh38
rs759637144 Chr.Y: 7064074-7064074 on Build GRCh38
rs760033971 Chr.Y: 7063960-7063960 on Build GRCh38
rs778071943 Chr.Y: 7063975-7063975 on Build GRCh38
rs569505746 Chr.Y: 7064206-7064206 on Build GRCh38
rs754042482 Chr.Y: 7064001-7064001 on Build GRCh38
rs374698874 Chr.Y: 7064189-7064189 on Build GRCh38
rs770617381 Chr.Y: 7063980-7063980 on Build GRCh38
rs762058902 Chr.Y: 7063887-7063887 on Build GRCh38
rs746615136 Chr.Y: 7063938-7063938 on Build GRCh38
rs768039732 Chr.Y: 7064157-7064157 on Build GRCh38
rs35573420 Chr.Y: 7064158-7064158 on Build GRCh38
rs748046580 Chr.Y: 7064123-7064123 on Build GRCh38
rs368887682 Chr.Y: 7064197-7064197 on Build GRCh38
rs369748251 Chr.Y: 7064041-7064041 on Build GRCh38
rs753794716 Chr.Y: 7063906-7063906 on Build GRCh38
rs199659121 Chr.Y: 7063898-7063898 on Build GRCh38
rs762271041 Chr.Y: 7064154-7064154 on Build GRCh38
rs990108838 Chr.Y: 7063882-7063882 on Build GRCh38
rs749847433 Chr.Y: 7063926-7063926 on Build GRCh38
rs376720969 Chr.Y: 7063976-7063976 on Build GRCh38
rs780236370 Chr.Y: 7063924-7063924 on Build GRCh38
rs758457756 Chr.Y: 7064186-7064186 on Build GRCh38
rs781251027 Chr.Y: 7063852-7063852 on Build GRCh38
rs762812638 Chr.Y: 7063946-7063946 on Build GRCh38
rs371139535 Chr.Y: 7063921-7063921 on Build GRCh38
rs958267268 Chr.Y: 7063879-7063879 on Build GRCh38
rs779475241 Chr.Y: 7063968-7063968 on Build GRCh38
rs756367085 Chr.Y: 7063959-7063959 on Build GRCh38
rs374243244 Chr.Y: 7063919-7063919 on Build GRCh38
rs757614980 Chr.Y: 7064034-7064034 on Build GRCh38
rs778719121 Chr.Y: 7063847-7063847 on Build GRCh38
rs764654188 Chr.Y: 7064095-7064095 on Build GRCh38
rs371074156 Chr.Y: 7064175-7064175 on Build GRCh38
rs377026718 Chr.Y: 7064105-7064105 on Build GRCh38
rs769990323 Chr.Y: 7063861-7063861 on Build GRCh38
rs199525748 Chr.Y: 7064130-7064130 on Build GRCh38
rs772254940 Chr.Y: 7063933-7063933 on Build GRCh38
rs765279648 Chr.Y: 7063942-7063942 on Build GRCh38
rs775286456 Chr.Y: 7063945-7063945 on Build GRCh38
rs768766263 Chr.Y: 7064054-7064054 on Build GRCh38
rs772864460 Chr.Y: 7063936-7063936 on Build GRCh38
rs1037975631 Chr.Y: 7064021-7064021 on Build GRCh38
rs774330206 Chr.Y: 7063995-7063995 on Build GRCh38
rs748542770 Chr.Y: 7063971-7063971 on Build GRCh38
rs371282846 Chr.Y: 7063912-7063912 on Build GRCh38
rs745452130 Chr.Y: 7063984-7063984 on Build GRCh38
rs375575285 Chr.Y: 7064039-7064039 on Build GRCh38
rs201301961 Chr.Y: 7064065-7064065 on Build GRCh38
rs779019950 Chr.Y: 7064217-7064217 on Build GRCh38
rs753278778 Chr.Y: 7064020-7064020 on Build GRCh38
rs756491978 Chr.Y: 7064161-7064161 on Build GRCh38
rs373544762 Chr.Y: 7064052-7064052 on Build GRCh38
rs765181144 Chr.Y: 7064014-7064014 on Build GRCh38
rs755094751 Chr.Y: 7064005-7064005 on Build GRCh38
rs771812119 Chr.Y: 7063894-7063894 on Build GRCh38
rs763743035 Chr.Y: 7063947-7063947 on Build GRCh38
rs77259694 Chr.Y: 7064150-7064150 on Build GRCh38
rs748810727 Chr.Y: 7064116-7064116 on Build GRCh38
rs376840960 Chr.Y: 7064162-7064162 on Build GRCh38
rs868448781 Chr.Y: 7064141-7064141 on Build GRCh38
rs941719081 Chr.Y: 7064229-7064229 on Build GRCh38
rs769967204 Chr.Y: 7064125-7064125 on Build GRCh38
rs765555033 Chr.Y: 7064032-7064032 on Build GRCh38
rs757316451 Chr.Y: 7064213-7064213 on Build GRCh38
rs762450167 Chr.Y: 7064026-7064026 on Build GRCh38
rs776253653 Chr.Y: 7063983-7063983 on Build GRCh38
rs775511235 Chr.Y: 7063990-7063990 on Build GRCh38
rs774423916 Chr.Y: 7064055-7064055 on Build GRCh38
rs757540810 Chr.Y: 7063851-7063851 on Build GRCh38
rs938503313 Chr.Y: 7064168-7064168 on Build GRCh38
rs763328207 Chr.Y: 7064094-7064094 on Build GRCh38
rs772730413 Chr.Y: 7063895-7063895 on Build GRCh38
rs758812822 Chr.Y: 7063974-7063974 on Build GRCh38
rs761683569 Chr.Y: 7063999-7063999 on Build GRCh38
rs747629623 Chr.Y: 7064178-7064178 on Build GRCh38
rs752767302 Chr.Y: 7064090-7064090 on Build GRCh38
rs372699079 Chr.Y: 7064006-7064006 on Build GRCh38
rs759427209 Chr.Y: 7063909-7063909 on Build GRCh38
rs758840645 Chr.Y: 7063873-7063873 on Build GRCh38
rs375303287 Chr.Y: 7064107-7064107 on Build GRCh38
rs373338636 Chr.Y: 7064100-7064100 on Build GRCh38

View Full Product Details