Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 20507274-20507789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTTY10
Entrez Gene ID
246119
Gene Name
testis-specific transcript, Y-linked 10 (non-protein coding)
Gene Aliases
NCRNA00133, TTY10
UniGene
Hs.461175
Gene Chromosome Location
Chr.Y: 20465668-20519228 on Build GRCh38
Transcripts
NR_001542.1

SNP Detail

SNP ID Chromosome Location
rs779665025 Chr.Y: 20507616-20507616 on Build GRCh38
rs1049602017 Chr.Y: 20507447-20507447 on Build GRCh38
rs891714701 Chr.Y: 20507612-20507612 on Build GRCh38
rs868036337 Chr.Y: 20507454-20507454 on Build GRCh38
rs931038845 Chr.Y: 20507434-20507434 on Build GRCh38
rs919714657 Chr.Y: 20507375-20507375 on Build GRCh38
rs746194935 Chr.Y: 20507600-20507600 on Build GRCh38
rs779460622 Chr.Y: 20507512-20507512 on Build GRCh38
rs796544114 Chr.Y: 20507334-20507334 on Build GRCh38
rs527342852 Chr.Y: 20507552-20507552 on Build GRCh38
rs374608056 Chr.Y: 20507639-20507639 on Build GRCh38
rs1040943268 Chr.Y: 20507504-20507504 on Build GRCh38
rs1037094521 Chr.Y: 20507354-20507354 on Build GRCh38
rs373302173 Chr.Y: 20507391-20507391 on Build GRCh38
rs1008158202 Chr.Y: 20507493-20507493 on Build GRCh38
rs774946716 Chr.Y: 20507657-20507657 on Build GRCh38

View Full Product Details